Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 51913210-51913715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCUN1D4
Entrez Gene ID
23142
Gene Name
defective in cullin neddylation 1 domain containing 4
Gene Aliases
-
UniGene
Hs.605388
Gene Chromosome Location
Chr.4: 51843000-51916837 on Build GRCh38
Transcripts
NM_001040402.2 NM_001287755.1 NM_001287757.1 NM_015115.3 XM_005265731.3 XM_011534379.2 XM_011534380.1 XM_017007912.1 XM_017007913.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5703021 c.841G>A p.E281K Chr.4: 51913546-51913546 on Build GRCh38
4755320 c.938A>G p.Y313C Chr.4: 51913375-51913375 on Build GRCh38
4790601 c.915A>C p.T305T Chr.4: 51913352-51913352 on Build GRCh38
7304321 c.862G>A p.D288N Chr.4: 51913567-51913567 on Build GRCh38
4559776 c.950G>A p.G317E Chr.4: 51913387-51913387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942010609 Chr.4: 51913394-51913394 on Build GRCh38
rs377704686 Chr.4: 51913432-51913432 on Build GRCh38
rs778817128 Chr.4: 51913603-51913603 on Build GRCh38
rs368141385 Chr.4: 51913565-51913565 on Build GRCh38
rs772671011 Chr.4: 51913621-51913621 on Build GRCh38
rs772135009 Chr.4: 51913397-51913397 on Build GRCh38
rs750996854 Chr.4: 51913337-51913337 on Build GRCh38
rs868684410 Chr.4: 51913290-51913290 on Build GRCh38
rs758548535 Chr.4: 51913497-51913497 on Build GRCh38
rs781407367 Chr.4: 51913598-51913598 on Build GRCh38
rs901968738 Chr.4: 51913467-51913467 on Build GRCh38
rs577923247 Chr.4: 51913666-51913666 on Build GRCh38
rs377194143 Chr.4: 51913268-51913268 on Build GRCh38
rs960537265 Chr.4: 51913661-51913661 on Build GRCh38
rs376692521 Chr.4: 51913404-51913404 on Build GRCh38
rs762735714 Chr.4: 51913322-51913322 on Build GRCh38
rs749626371 Chr.4: 51913376-51913376 on Build GRCh38
rs61794807 Chr.4: 51913405-51913405 on Build GRCh38
rs528620753 Chr.4: 51913435-51913435 on Build GRCh38
rs757340983 Chr.4: 51913593-51913593 on Build GRCh38
rs1042994451 Chr.4: 51913447-51913447 on Build GRCh38
rs201277301 Chr.4: 51913292-51913292 on Build GRCh38
rs913983711 Chr.4: 51913366-51913366 on Build GRCh38
rs988204877 Chr.4: 51913325-51913325 on Build GRCh38
rs776569592 Chr.4: 51913270-51913270 on Build GRCh38
rs774461280 Chr.4: 51913256-51913256 on Build GRCh38
rs779124644 Chr.4: 51913492-51913492 on Build GRCh38
rs998487002 Chr.4: 51913471-51913471 on Build GRCh38
rs200190417 Chr.4: 51913347-51913347 on Build GRCh38
rs200685693 Chr.4: 51913485-51913486 on Build GRCh38
rs145441461 Chr.4: 51913491-51913491 on Build GRCh38
rs904411975 Chr.4: 51913460-51913460 on Build GRCh38
rs780101399 Chr.4: 51913503-51913503 on Build GRCh38
rs139477455 Chr.4: 51913545-51913545 on Build GRCh38
rs774088304 Chr.4: 51913422-51913422 on Build GRCh38
rs374696322 Chr.4: 51913523-51913523 on Build GRCh38
rs748234035 Chr.4: 51913389-51913389 on Build GRCh38
rs748172614 Chr.4: 51913267-51913267 on Build GRCh38
rs762403935 Chr.4: 51913613-51913613 on Build GRCh38
rs767921887 Chr.4: 51913334-51913334 on Build GRCh38
rs1016804560 Chr.4: 51913238-51913238 on Build GRCh38
rs759267380 Chr.4: 51913283-51913283 on Build GRCh38
rs797004681 Chr.4: 51913485-51913485 on Build GRCh38
rs753023556 Chr.4: 51913494-51913494 on Build GRCh38
rs771598702 Chr.4: 51913517-51913517 on Build GRCh38
rs201472874 Chr.4: 51913620-51913620 on Build GRCh38
rs550876067 Chr.4: 51913519-51913519 on Build GRCh38
rs754230709 Chr.4: 51913489-51913489 on Build GRCh38
rs765661497 Chr.4: 51913562-51913562 on Build GRCh38
rs968151536 Chr.4: 51913667-51913667 on Build GRCh38
rs189490258 Chr.4: 51913243-51913243 on Build GRCh38
rs376168943 Chr.4: 51913423-51913423 on Build GRCh38
rs760243268 Chr.4: 51913629-51913629 on Build GRCh38
rs766818060 Chr.4: 51913482-51913482 on Build GRCh38
rs1014032348 Chr.4: 51913521-51913521 on Build GRCh38
rs1046800966 Chr.4: 51913501-51913502 on Build GRCh38
rs1035352220 Chr.4: 51913631-51913631 on Build GRCh38
rs751137719 Chr.4: 51913330-51913330 on Build GRCh38
rs780942679 Chr.4: 51913247-51913247 on Build GRCh38
rs745489867 Chr.4: 51913362-51913362 on Build GRCh38
rs760026098 Chr.4: 51913546-51913546 on Build GRCh38
rs371642489 Chr.4: 51913569-51913569 on Build GRCh38
rs777809965 Chr.4: 51913575-51913575 on Build GRCh38
rs868543946 Chr.4: 51913694-51913694 on Build GRCh38
rs373757007 Chr.4: 51913384-51913384 on Build GRCh38
rs775424175 Chr.4: 51913305-51913305 on Build GRCh38
rs762788891 Chr.4: 51913414-51913414 on Build GRCh38
rs761565930 Chr.4: 51913512-51913512 on Build GRCh38
rs749100634 Chr.4: 51913504-51913504 on Build GRCh38
rs747845422 Chr.4: 51913612-51913612 on Build GRCh38
rs755213448 Chr.4: 51913377-51913377 on Build GRCh38
rs370308968 Chr.4: 51913500-51913500 on Build GRCh38
rs780504863 Chr.4: 51913375-51913375 on Build GRCh38
rs756443062 Chr.4: 51913240-51913240 on Build GRCh38
rs763871991 Chr.4: 51913419-51913419 on Build GRCh38
rs972076738 Chr.4: 51913691-51913691 on Build GRCh38
rs555373137 Chr.4: 51913563-51913563 on Build GRCh38
rs756967597 Chr.4: 51913488-51913489 on Build GRCh38
rs145055298 Chr.4: 51913585-51913585 on Build GRCh38
rs762744035 Chr.4: 51913486-51913488 on Build GRCh38
rs199758089 Chr.4: 51913411-51913411 on Build GRCh38
rs756752597 Chr.4: 51913331-51913331 on Build GRCh38
rs769792599 Chr.4: 51913501-51913501 on Build GRCh38
rs981053202 Chr.4: 51913351-51913351 on Build GRCh38
rs191084956 Chr.4: 51913602-51913602 on Build GRCh38
rs771812493 Chr.4: 51913608-51913620 on Build GRCh38
rs763818730 Chr.4: 51913327-51913327 on Build GRCh38
rs774283158 Chr.4: 51913508-51913508 on Build GRCh38
rs768832262 Chr.4: 51913250-51913250 on Build GRCh38
rs185833634 Chr.4: 51913449-51913449 on Build GRCh38
rs775475185 Chr.4: 51913413-51913413 on Build GRCh38
rs569280869 Chr.4: 51913542-51913542 on Build GRCh38
rs745523450 Chr.4: 51913486-51913491 on Build GRCh38

View Full Product Details