Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 55957138-55957646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP135
Entrez Gene ID
9662
Gene Name
centrosomal protein 135
Gene Aliases
CEP4, KIAA0635, MCPH8
UniGene
Hs.518767
Gene Chromosome Location
Chr.4: 55948808-56033363 on Build GRCh38
Transcripts
NM_025009.4 XM_005265788.3 XM_006714055.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3661138 c.521A>T p.D174V Chr.4: 55957271-55957271 on Build GRCh38
4125240 c.575A>G p.D192G Chr.4: 55957325-55957325 on Build GRCh38
6321127 c.544G>T p.V182F Chr.4: 55957294-55957294 on Build GRCh38
1618928 c.521A>T p.D174V Chr.4: 55957271-55957271 on Build GRCh38
7276129 c.503G>A p.R168H Chr.4: 55957253-55957253 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750392584 Chr.4: 55957252-55957252 on Build GRCh38
rs763632922 Chr.4: 55957315-55957315 on Build GRCh38
rs374714305 Chr.4: 55957343-55957343 on Build GRCh38
rs374576275 Chr.4: 55957399-55957399 on Build GRCh38
rs901148460 Chr.4: 55957449-55957449 on Build GRCh38
rs752703938 Chr.4: 55957380-55957380 on Build GRCh38
rs1048552678 Chr.4: 55957288-55957288 on Build GRCh38
rs747281578 Chr.4: 55957406-55957406 on Build GRCh38
rs77889786 Chr.4: 55957211-55957211 on Build GRCh38
rs1011191945 Chr.4: 55957515-55957515 on Build GRCh38
rs999622679 Chr.4: 55957467-55957467 on Build GRCh38
rs376270991 Chr.4: 55957521-55957521 on Build GRCh38
rs754959625 Chr.4: 55957494-55957494 on Build GRCh38
rs545633683 Chr.4: 55957450-55957450 on Build GRCh38
rs200668189 Chr.4: 55957386-55957386 on Build GRCh38
rs757125308 Chr.4: 55957229-55957229 on Build GRCh38
rs961717044 Chr.4: 55957583-55957583 on Build GRCh38
rs752864504 Chr.4: 55957162-55957162 on Build GRCh38
rs748235612 Chr.4: 55957187-55957189 on Build GRCh38
rs911470283 Chr.4: 55957477-55957477 on Build GRCh38
rs765813255 Chr.4: 55957205-55957205 on Build GRCh38
rs781077695 Chr.4: 55957235-55957235 on Build GRCh38
rs773474803 Chr.4: 55957326-55957326 on Build GRCh38
rs969281762 Chr.4: 55957543-55957543 on Build GRCh38
rs777824090 Chr.4: 55957392-55957392 on Build GRCh38
rs76369732 Chr.4: 55957225-55957225 on Build GRCh38
rs971759733 Chr.4: 55957258-55957258 on Build GRCh38
rs774860984 Chr.4: 55957298-55957298 on Build GRCh38
rs769946610 Chr.4: 55957193-55957193 on Build GRCh38
rs755001124 Chr.4: 55957253-55957253 on Build GRCh38
rs1013406614 Chr.4: 55957612-55957612 on Build GRCh38
rs760156022 Chr.4: 55957199-55957199 on Build GRCh38
rs972796449 Chr.4: 55957419-55957419 on Build GRCh38
rs753638567 Chr.4: 55957556-55957556 on Build GRCh38
rs760137546 Chr.4: 55957284-55957284 on Build GRCh38
rs778112036 Chr.4: 55957267-55957267 on Build GRCh38
rs762583480 Chr.4: 55957302-55957302 on Build GRCh38
rs778759306 Chr.4: 55957607-55957607 on Build GRCh38
rs753654183 Chr.4: 55957273-55957273 on Build GRCh38
rs149954755 Chr.4: 55957277-55957277 on Build GRCh38
rs367650085 Chr.4: 55957222-55957222 on Build GRCh38
rs763219938 Chr.4: 55957216-55957216 on Build GRCh38
rs756047797 Chr.4: 55957370-55957370 on Build GRCh38
rs576589552 Chr.4: 55957446-55957446 on Build GRCh38
rs561093788 Chr.4: 55957609-55957609 on Build GRCh38
rs1052195572 Chr.4: 55957560-55957560 on Build GRCh38
rs761475684 Chr.4: 55957336-55957336 on Build GRCh38
rs62308566 Chr.4: 55957563-55957563 on Build GRCh38
rs748333562 Chr.4: 55957254-55957254 on Build GRCh38
rs1023436865 Chr.4: 55957527-55957527 on Build GRCh38
rs186857421 Chr.4: 55957491-55957491 on Build GRCh38
rs777232769 Chr.4: 55957278-55957278 on Build GRCh38
rs772177312 Chr.4: 55957259-55957259 on Build GRCh38
rs769365357 Chr.4: 55957287-55957287 on Build GRCh38
rs750302631 Chr.4: 55957347-55957347 on Build GRCh38
rs760310911 Chr.4: 55957174-55957174 on Build GRCh38
rs942894930 Chr.4: 55957354-55957354 on Build GRCh38
rs1043363091 Chr.4: 55957510-55957510 on Build GRCh38
rs969325626 Chr.4: 55957166-55957166 on Build GRCh38
rs777553062 Chr.4: 55957473-55957473 on Build GRCh38
rs367901491 Chr.4: 55957377-55957377 on Build GRCh38
rs961788985 Chr.4: 55957215-55957215 on Build GRCh38
rs751265741 Chr.4: 55957333-55957333 on Build GRCh38
rs575680187 Chr.4: 55957492-55957492 on Build GRCh38
rs776236115 Chr.4: 55957195-55957195 on Build GRCh38
rs771368790 Chr.4: 55957410-55957410 on Build GRCh38
rs766004757 Chr.4: 55957180-55957180 on Build GRCh38

View Full Product Details