Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 68831575-68832076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UGT2B10
Entrez Gene ID
7365
Gene Name
UDP glucuronosyltransferase family 2 member B10
Gene Aliases
UDPGT2B10
UniGene
Hs.201634
Gene Chromosome Location
Chr.4: 68815981-68832038 on Build GRCh38
Transcripts
NM_001075.5 NM_001144767.2 NM_001290091.1 XM_017008585.1

SNP Detail

SNP ID Chromosome Location
rs775913889 Chr.4: 68831700-68831701 on Build GRCh38
rs545613497 Chr.4: 68831862-68831862 on Build GRCh38
rs556430704 Chr.4: 68831953-68831953 on Build GRCh38
rs114722061 Chr.4: 68831807-68831807 on Build GRCh38
rs954239353 Chr.4: 68831720-68831720 on Build GRCh38
rs77745587 Chr.4: 68831771-68831771 on Build GRCh38
rs116116268 Chr.4: 68831846-68831846 on Build GRCh38
rs998197944 Chr.4: 68831868-68831868 on Build GRCh38
rs185649037 Chr.4: 68831652-68831652 on Build GRCh38
rs972969412 Chr.4: 68831757-68831757 on Build GRCh38
rs763727284 Chr.4: 68831930-68831930 on Build GRCh38
rs576604197 Chr.4: 68832024-68832024 on Build GRCh38
rs547431392 Chr.4: 68831752-68831752 on Build GRCh38
rs965378969 Chr.4: 68831681-68831681 on Build GRCh38
rs1006086052 Chr.4: 68832021-68832021 on Build GRCh38
rs75446314 Chr.4: 68831749-68831749 on Build GRCh38
rs146732640 Chr.4: 68831738-68831738 on Build GRCh38
rs926297817 Chr.4: 68831826-68831826 on Build GRCh38
rs190571045 Chr.4: 68831978-68831978 on Build GRCh38
rs901065015 Chr.4: 68831867-68831867 on Build GRCh38
rs1002057902 Chr.4: 68831606-68831606 on Build GRCh38
rs1020875721 Chr.4: 68831671-68831671 on Build GRCh38
rs954051226 Chr.4: 68831992-68831992 on Build GRCh38
rs1046017567 Chr.4: 68831815-68831815 on Build GRCh38
rs762972498 Chr.4: 68831877-68831877 on Build GRCh38
rs1009368745 Chr.4: 68831857-68831857 on Build GRCh38
rs893621630 Chr.4: 68831648-68831648 on Build GRCh38
rs917461592 Chr.4: 68831804-68831804 on Build GRCh38
rs1032619241 Chr.4: 68831614-68831614 on Build GRCh38
rs7661590 Chr.4: 68831841-68831841 on Build GRCh38
rs539600765 Chr.4: 68831830-68831830 on Build GRCh38
rs369292637 Chr.4: 68831730-68831730 on Build GRCh38

View Full Product Details