Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 69999575-70000076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STATH
Entrez Gene ID
6779
Gene Name
statherin
Gene Aliases
STR
UniGene
Hs.654495
Gene Chromosome Location
Chr.4: 69995931-70002456 on Build GRCh38
Transcripts
NM_001009181.1 NM_003154.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3748845 c.71A>T p.E24V Chr.4: 69999686-69999686 on Build GRCh38
1178121 c.101G>C p.G34A Chr.4: 69999808-69999808 on Build GRCh38
3826113 c.65C>G p.S22C Chr.4: 69999680-69999680 on Build GRCh38
5780349 c.83G>A p.R28H Chr.4: 69999790-69999790 on Build GRCh38
2830007 c.75A>T p.K25N Chr.4: 69999782-69999782 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766117728 Chr.4: 69999806-69999806 on Build GRCh38
rs368038102 Chr.4: 69999825-69999825 on Build GRCh38
rs928492638 Chr.4: 70000001-70000001 on Build GRCh38
rs760500812 Chr.4: 69999804-69999804 on Build GRCh38
rs867264792 Chr.4: 69999666-69999666 on Build GRCh38
rs375160503 Chr.4: 69999824-69999824 on Build GRCh38
rs752770384 Chr.4: 69999639-69999639 on Build GRCh38
rs901316232 Chr.4: 69999936-69999936 on Build GRCh38
rs751642322 Chr.4: 69999817-69999817 on Build GRCh38
rs773047183 Chr.4: 69999713-69999713 on Build GRCh38
rs775202782 Chr.4: 69999690-69999690 on Build GRCh38
rs184817332 Chr.4: 69999672-69999672 on Build GRCh38
rs181619166 Chr.4: 69999790-69999790 on Build GRCh38
rs746372131 Chr.4: 69999675-69999675 on Build GRCh38
rs760597800 Chr.4: 69999714-69999714 on Build GRCh38
rs759404825 Chr.4: 69999813-69999813 on Build GRCh38
rs199543155 Chr.4: 69999731-69999731 on Build GRCh38
rs757484423 Chr.4: 69999819-69999819 on Build GRCh38
rs754145715 Chr.4: 70000003-70000003 on Build GRCh38
rs747664860 Chr.4: 69999857-69999857 on Build GRCh38
rs764121004 Chr.4: 69999740-69999740 on Build GRCh38
rs768470714 Chr.4: 69999702-69999702 on Build GRCh38
rs983116969 Chr.4: 70000024-70000024 on Build GRCh38
rs757595567 Chr.4: 69999650-69999650 on Build GRCh38
rs755530429 Chr.4: 69999847-69999847 on Build GRCh38
rs190082343 Chr.4: 69999705-69999705 on Build GRCh38
rs748776387 Chr.4: 69999695-69999695 on Build GRCh38
rs776585042 Chr.4: 69999807-69999807 on Build GRCh38
rs778354909 Chr.4: 69999850-69999850 on Build GRCh38
rs754399857 Chr.4: 69999831-69999831 on Build GRCh38
rs764949898 Chr.4: 69999788-69999788 on Build GRCh38
rs543985484 Chr.4: 69999758-69999758 on Build GRCh38
rs751732642 Chr.4: 69999746-69999746 on Build GRCh38
rs998348359 Chr.4: 69999957-69999957 on Build GRCh38
rs149625945 Chr.4: 70000034-70000034 on Build GRCh38
rs758664636 Chr.4: 69999640-69999640 on Build GRCh38
rs563811308 Chr.4: 69999995-69999995 on Build GRCh38
rs981287535 Chr.4: 69999724-69999724 on Build GRCh38
rs147122083 Chr.4: 69999816-69999816 on Build GRCh38
rs566115243 Chr.4: 69999706-69999706 on Build GRCh38
rs531310187 Chr.4: 70000019-70000019 on Build GRCh38
rs972437413 Chr.4: 69999648-69999648 on Build GRCh38
rs576856259 Chr.4: 70000026-70000027 on Build GRCh38
rs770317683 Chr.4: 69999688-69999688 on Build GRCh38
rs1026293001 Chr.4: 69999910-69999910 on Build GRCh38
rs1015841933 Chr.4: 70000027-70000027 on Build GRCh38
rs758498380 Chr.4: 69999737-69999737 on Build GRCh38
rs772919991 Chr.4: 69999797-69999797 on Build GRCh38
rs374363281 Chr.4: 69999729-69999729 on Build GRCh38
rs947096291 Chr.4: 70000046-70000046 on Build GRCh38
rs200332032 Chr.4: 69999711-69999711 on Build GRCh38
rs756023359 Chr.4: 69999626-69999626 on Build GRCh38
rs768311029 Chr.4: 69999780-69999780 on Build GRCh38
rs767744699 Chr.4: 69999820-69999820 on Build GRCh38
rs541755658 Chr.4: 69999865-69999865 on Build GRCh38
rs374610076 Chr.4: 69999760-69999760 on Build GRCh38
rs780683918 Chr.4: 69999771-69999771 on Build GRCh38
rs750334946 Chr.4: 69999623-69999623 on Build GRCh38
rs142984801 Chr.4: 69999751-69999751 on Build GRCh38
rs373902005 Chr.4: 69999642-69999642 on Build GRCh38
rs111880467 Chr.4: 69999649-69999649 on Build GRCh38
rs545409719 Chr.4: 69999977-69999977 on Build GRCh38
rs958559417 Chr.4: 70000004-70000004 on Build GRCh38
rs757403720 Chr.4: 69999700-69999701 on Build GRCh38
rs914256602 Chr.4: 69999783-69999783 on Build GRCh38
rs1018458812 Chr.4: 69999906-69999906 on Build GRCh38
rs549971445 Chr.4: 69999630-69999630 on Build GRCh38
rs965667017 Chr.4: 69999612-69999612 on Build GRCh38
rs138539585 Chr.4: 69999960-69999960 on Build GRCh38
rs17698678 Chr.4: 69999904-69999904 on Build GRCh38
rs757416119 Chr.4: 69999643-69999643 on Build GRCh38
rs754124875 Chr.4: 69999635-69999638 on Build GRCh38
rs977380993 Chr.4: 69999622-69999622 on Build GRCh38
rs377190127 Chr.4: 69999882-69999882 on Build GRCh38
rs750868144 Chr.4: 69999789-69999789 on Build GRCh38
rs750179437 Chr.4: 69999810-69999811 on Build GRCh38

View Full Product Details