Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87170310-87170807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL8
Entrez Gene ID
57563
Gene Name
kelch like family member 8
Gene Aliases
-
UniGene
Hs.106601
Gene Chromosome Location
Chr.4: 87160103-87220608 on Build GRCh38
Transcripts
NM_001292003.1 NM_001292006.1 NM_001292007.1 NM_020803.4 NR_120447.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
124009 c.1211G>A p.R404Q Chr.4: 87170613-87170613 on Build GRCh38
1058589 c.1246T>C p.Y416H Chr.4: 87170578-87170578 on Build GRCh38
6447077 c.1370C>T p.A457V Chr.4: 87170454-87170454 on Build GRCh38
1212687 c.1243A>G p.I415V Chr.4: 87170581-87170581 on Build GRCh38
5425842 c.1377+10A>T p.? Chr.4: 87170437-87170437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547537190 Chr.4: 87170559-87170559 on Build GRCh38
rs898399629 Chr.4: 87170582-87170582 on Build GRCh38
rs57080086 Chr.4: 87170437-87170437 on Build GRCh38
rs1043449262 Chr.4: 87170340-87170340 on Build GRCh38
rs920999844 Chr.4: 87170351-87170351 on Build GRCh38
rs755087098 Chr.4: 87170454-87170454 on Build GRCh38
rs1045657023 Chr.4: 87170595-87170595 on Build GRCh38
rs1025568359 Chr.4: 87170614-87170614 on Build GRCh38
rs775332249 Chr.4: 87170414-87170414 on Build GRCh38
rs765955558 Chr.4: 87170727-87170727 on Build GRCh38
rs764444844 Chr.4: 87170633-87170633 on Build GRCh38
rs775598413 Chr.4: 87170663-87170663 on Build GRCh38
rs960784472 Chr.4: 87170654-87170654 on Build GRCh38
rs532263956 Chr.4: 87170536-87170536 on Build GRCh38
rs757169631 Chr.4: 87170635-87170635 on Build GRCh38
rs773858482 Chr.4: 87170549-87170549 on Build GRCh38
rs761165036 Chr.4: 87170572-87170572 on Build GRCh38
rs1023383568 Chr.4: 87170753-87170753 on Build GRCh38
rs769397900 Chr.4: 87170410-87170410 on Build GRCh38
rs548356822 Chr.4: 87170653-87170653 on Build GRCh38
rs768368543 Chr.4: 87170541-87170541 on Build GRCh38
rs1050899491 Chr.4: 87170475-87170475 on Build GRCh38
rs750599744 Chr.4: 87170711-87170711 on Build GRCh38
rs150094972 Chr.4: 87170462-87170462 on Build GRCh38
rs751903687 Chr.4: 87170367-87170367 on Build GRCh38
rs760070920 Chr.4: 87170588-87170588 on Build GRCh38
rs925513831 Chr.4: 87170373-87170373 on Build GRCh38
rs777533085 Chr.4: 87170748-87170749 on Build GRCh38
rs766697068 Chr.4: 87170575-87170575 on Build GRCh38
rs994467110 Chr.4: 87170591-87170591 on Build GRCh38
rs781737612 Chr.4: 87170521-87170521 on Build GRCh38
rs773628185 Chr.4: 87170438-87170438 on Build GRCh38
rs901977208 Chr.4: 87170449-87170449 on Build GRCh38
rs781158236 Chr.4: 87170647-87170647 on Build GRCh38
rs1056467678 Chr.4: 87170677-87170677 on Build GRCh38
rs768042496 Chr.4: 87170660-87170660 on Build GRCh38
rs946889172 Chr.4: 87170479-87170479 on Build GRCh38
rs745780837 Chr.4: 87170503-87170503 on Build GRCh38
rs781319440 Chr.4: 87170495-87170495 on Build GRCh38
rs1004074492 Chr.4: 87170766-87170766 on Build GRCh38
rs368547120 Chr.4: 87170399-87170399 on Build GRCh38
rs181224269 Chr.4: 87170648-87170648 on Build GRCh38
rs757114495 Chr.4: 87170490-87170490 on Build GRCh38
rs200768283 Chr.4: 87170627-87170627 on Build GRCh38
rs955315047 Chr.4: 87170352-87170352 on Build GRCh38
rs777803997 Chr.4: 87170488-87170488 on Build GRCh38
rs751701752 Chr.4: 87170634-87170634 on Build GRCh38
rs969157391 Chr.4: 87170768-87170768 on Build GRCh38
rs758653635 Chr.4: 87170466-87170466 on Build GRCh38
rs112151312 Chr.4: 87170403-87170403 on Build GRCh38
rs17854115 Chr.4: 87170603-87170603 on Build GRCh38
rs767966938 Chr.4: 87170519-87170520 on Build GRCh38
rs746882488 Chr.4: 87170489-87170489 on Build GRCh38
rs200861123 Chr.4: 87170625-87170625 on Build GRCh38
rs754335030 Chr.4: 87170444-87170444 on Build GRCh38
rs777168344 Chr.4: 87170581-87170581 on Build GRCh38
rs375509977 Chr.4: 87170509-87170509 on Build GRCh38
rs760712758 Chr.4: 87170663-87170664 on Build GRCh38
rs745440147 Chr.4: 87170400-87170400 on Build GRCh38
rs200101206 Chr.4: 87170439-87170439 on Build GRCh38
rs1016246462 Chr.4: 87170554-87170554 on Build GRCh38
rs765274896 Chr.4: 87170455-87170455 on Build GRCh38
rs992045229 Chr.4: 87170708-87170708 on Build GRCh38

View Full Product Details