Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99313679-99314208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH1B
Entrez Gene ID
125
Gene Name
alcohol dehydrogenase 1B (class I), beta polypeptide
Gene Aliases
ADH2, HEL-S-117
UniGene
Hs.4
Gene Chromosome Location
Chr.4: 99306387-99321442 on Build GRCh38
Transcripts
NM_000668.5 NM_001286650.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5918333 c.824C>T p.T275I Chr.4: 99313825-99313825 on Build GRCh38
4554818 c.631G>A p.G211S Chr.4: 99314018-99314018 on Build GRCh38
1159748 c.811G>A p.G271S Chr.4: 99313838-99313838 on Build GRCh38
7238882 c.666G>T p.A222A Chr.4: 99313983-99313983 on Build GRCh38
7240506 c.645T>C p.A215A Chr.4: 99314004-99314004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78984460 Chr.4: 99314097-99314097 on Build GRCh38
rs1051106197 Chr.4: 99313816-99313816 on Build GRCh38
rs764414006 Chr.4: 99314041-99314041 on Build GRCh38
rs911259035 Chr.4: 99314085-99314085 on Build GRCh38
rs780390061 Chr.4: 99314101-99314101 on Build GRCh38
rs773852995 Chr.4: 99313928-99313928 on Build GRCh38
rs770039399 Chr.4: 99314158-99314158 on Build GRCh38
rs759891440 Chr.4: 99313861-99313861 on Build GRCh38
rs774743141 Chr.4: 99314115-99314115 on Build GRCh38
rs767946339 Chr.4: 99313779-99313779 on Build GRCh38
rs79236485 Chr.4: 99314098-99314098 on Build GRCh38
rs557448763 Chr.4: 99314065-99314065 on Build GRCh38
rs758475277 Chr.4: 99313803-99313803 on Build GRCh38
rs755890908 Chr.4: 99314010-99314010 on Build GRCh38
rs370516154 Chr.4: 99313851-99313851 on Build GRCh38
rs577040077 Chr.4: 99313703-99313703 on Build GRCh38
rs773483740 Chr.4: 99314120-99314120 on Build GRCh38
rs773553129 Chr.4: 99314160-99314160 on Build GRCh38
rs747643331 Chr.4: 99313932-99313932 on Build GRCh38
rs181946861 Chr.4: 99313776-99313776 on Build GRCh38
rs769438704 Chr.4: 99313771-99313771 on Build GRCh38
rs371001544 Chr.4: 99314024-99314024 on Build GRCh38
rs748906738 Chr.4: 99314015-99314015 on Build GRCh38
rs377123975 Chr.4: 99313815-99313815 on Build GRCh38
rs28626993 Chr.4: 99314037-99314037 on Build GRCh38
rs760601299 Chr.4: 99314062-99314062 on Build GRCh38
rs189894519 Chr.4: 99313749-99313749 on Build GRCh38
rs755948847 Chr.4: 99313909-99313909 on Build GRCh38
rs777623976 Chr.4: 99313804-99313804 on Build GRCh38
rs768162102 Chr.4: 99313835-99313835 on Build GRCh38
rs745641952 Chr.4: 99313817-99313817 on Build GRCh38
rs779592631 Chr.4: 99313913-99313913 on Build GRCh38
rs757937623 Chr.4: 99314086-99314086 on Build GRCh38
rs762059310 Chr.4: 99314116-99314116 on Build GRCh38
rs149386430 Chr.4: 99313892-99313892 on Build GRCh38
rs539249951 Chr.4: 99314049-99314049 on Build GRCh38
rs532844712 Chr.4: 99313783-99313783 on Build GRCh38
rs918576208 Chr.4: 99313814-99313814 on Build GRCh38
rs750335943 Chr.4: 99314001-99314001 on Build GRCh38
rs777147821 Chr.4: 99314031-99314031 on Build GRCh38
rs758125644 Chr.4: 99313890-99313890 on Build GRCh38
rs755136035 Chr.4: 99313796-99313796 on Build GRCh38
rs900197737 Chr.4: 99314155-99314155 on Build GRCh38
rs113103450 Chr.4: 99313758-99313758 on Build GRCh38
rs186117499 Chr.4: 99313797-99313797 on Build GRCh38
rs201999694 Chr.4: 99314042-99314042 on Build GRCh38
rs748930079 Chr.4: 99313916-99313916 on Build GRCh38
rs561969097 Chr.4: 99313705-99313705 on Build GRCh38
rs535041064 Chr.4: 99313982-99313982 on Build GRCh38
rs77258305 Chr.4: 99314078-99314078 on Build GRCh38
rs78681053 Chr.4: 99313926-99313926 on Build GRCh38
rs904388509 Chr.4: 99313839-99313839 on Build GRCh38
rs748144394 Chr.4: 99314134-99314134 on Build GRCh38
rs775870751 Chr.4: 99314039-99314039 on Build GRCh38
rs150495372 Chr.4: 99313834-99313834 on Build GRCh38
rs751751640 Chr.4: 99313984-99313984 on Build GRCh38
rs772290764 Chr.4: 99314119-99314119 on Build GRCh38
rs757377587 Chr.4: 99313995-99313995 on Build GRCh38
rs766744276 Chr.4: 99313843-99313843 on Build GRCh38
rs551035498 Chr.4: 99313795-99313795 on Build GRCh38
rs763329813 Chr.4: 99314167-99314167 on Build GRCh38
rs995852183 Chr.4: 99314163-99314163 on Build GRCh38
rs74451421 Chr.4: 99313838-99313838 on Build GRCh38
rs141384054 Chr.4: 99313882-99313882 on Build GRCh38
rs148100209 Chr.4: 99313936-99313936 on Build GRCh38
rs201571406 Chr.4: 99313875-99313875 on Build GRCh38
rs774916432 Chr.4: 99313774-99313774 on Build GRCh38
rs754717581 Chr.4: 99314020-99314020 on Build GRCh38
rs529217285 Chr.4: 99313716-99313716 on Build GRCh38
rs752300177 Chr.4: 99314077-99314077 on Build GRCh38
rs904448712 Chr.4: 99313711-99313711 on Build GRCh38
rs1031333865 Chr.4: 99313719-99313719 on Build GRCh38
rs775609473 Chr.4: 99313957-99313957 on Build GRCh38
rs75973598 Chr.4: 99313931-99313931 on Build GRCh38
rs770886223 Chr.4: 99313809-99313809 on Build GRCh38
rs955719465 Chr.4: 99313725-99313725 on Build GRCh38
rs761103181 Chr.4: 99313841-99313841 on Build GRCh38
rs923754714 Chr.4: 99313787-99313798 on Build GRCh38
rs768851539 Chr.4: 99314105-99314105 on Build GRCh38
rs770395174 Chr.4: 99314099-99314099 on Build GRCh38
rs376719170 Chr.4: 99313792-99313792 on Build GRCh38
rs911321037 Chr.4: 99313826-99313826 on Build GRCh38
rs1038744555 Chr.4: 99313741-99313741 on Build GRCh38
rs367750195 Chr.4: 99313788-99313788 on Build GRCh38
rs373672745 Chr.4: 99313905-99313905 on Build GRCh38
rs78574385 Chr.4: 99313886-99313886 on Build GRCh38
rs575649813 Chr.4: 99314140-99314140 on Build GRCh38
rs770061958 Chr.4: 99314038-99314038 on Build GRCh38
rs750389182 Chr.4: 99314053-99314053 on Build GRCh38
rs775173450 Chr.4: 99313831-99313831 on Build GRCh38
rs754699804 Chr.4: 99314071-99314071 on Build GRCh38
rs908790517 Chr.4: 99313718-99313718 on Build GRCh38
rs769635325 Chr.4: 99313819-99313819 on Build GRCh38
rs780464594 Chr.4: 99313743-99313743 on Build GRCh38
rs778587185 Chr.4: 99314075-99314075 on Build GRCh38
rs759944524 Chr.4: 99313944-99313944 on Build GRCh38
rs752789670 Chr.4: 99313800-99313800 on Build GRCh38
rs76667147 Chr.4: 99314006-99314006 on Build GRCh38
rs760667354 Chr.4: 99314129-99314129 on Build GRCh38
rs753732945 Chr.4: 99314067-99314067 on Build GRCh38
rs779831454 Chr.4: 99314011-99314011 on Build GRCh38
rs2018417 Chr.4: 99313983-99313983 on Build GRCh38
rs749707610 Chr.4: 99314104-99314104 on Build GRCh38
rs367812131 Chr.4: 99313997-99313997 on Build GRCh38
rs1789882 Chr.4: 99313896-99313896 on Build GRCh38
rs765419231 Chr.4: 99313869-99313869 on Build GRCh38
rs753698775 Chr.4: 99314133-99314133 on Build GRCh38
rs763218108 Chr.4: 99314040-99314040 on Build GRCh38
rs766381253 Chr.4: 99314131-99314131 on Build GRCh38
rs199777080 Chr.4: 99313807-99313807 on Build GRCh38
rs1024076752 Chr.4: 99313745-99313745 on Build GRCh38

View Full Product Details