Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 100187498-100187992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDIT4L
Entrez Gene ID
115265
Gene Name
DNA damage inducible transcript 4 like
Gene Aliases
REDD2, Rtp801L
UniGene
Hs.480378
Gene Chromosome Location
Chr.4: 100185870-100190498 on Build GRCh38
Transcripts
NM_145244.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2951639 c.475C>T p.R159C Chr.4: 100187784-100187784 on Build GRCh38
4787616 c.460T>G p.F154V Chr.4: 100187799-100187799 on Build GRCh38
260375 c.523C>T p.R175* Chr.4: 100187736-100187736 on Build GRCh38
1683999 c.535delA p.K180fs*5 Chr.4: 100187724-100187724 on Build GRCh38
1618197 c.295G>C p.E99Q Chr.4: 100187964-100187964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758099451 Chr.4: 100187947-100187947 on Build GRCh38
rs553786082 Chr.4: 100187757-100187757 on Build GRCh38
rs145506344 Chr.4: 100187767-100187767 on Build GRCh38
rs201938125 Chr.4: 100187955-100187955 on Build GRCh38
rs770929183 Chr.4: 100187740-100187741 on Build GRCh38
rs775177490 Chr.4: 100187904-100187907 on Build GRCh38
rs1007966863 Chr.4: 100187594-100187594 on Build GRCh38
rs750516670 Chr.4: 100187893-100187893 on Build GRCh38
rs771710780 Chr.4: 100187831-100187831 on Build GRCh38
rs200154511 Chr.4: 100187730-100187730 on Build GRCh38
rs371837784 Chr.4: 100187936-100187936 on Build GRCh38
rs146614946 Chr.4: 100187950-100187950 on Build GRCh38
rs777634074 Chr.4: 100187665-100187665 on Build GRCh38
rs781368736 Chr.4: 100187696-100187696 on Build GRCh38
rs978276114 Chr.4: 100187556-100187556 on Build GRCh38
rs887569801 Chr.4: 100187794-100187794 on Build GRCh38
rs779779103 Chr.4: 100187724-100187724 on Build GRCh38
rs556541575 Chr.4: 100187789-100187789 on Build GRCh38
rs897010877 Chr.4: 100187612-100187613 on Build GRCh38
rs746599140 Chr.4: 100187954-100187954 on Build GRCh38
rs867286833 Chr.4: 100187869-100187869 on Build GRCh38
rs745495312 Chr.4: 100187639-100187639 on Build GRCh38
rs777145731 Chr.4: 100187626-100187626 on Build GRCh38
rs760841229 Chr.4: 100187927-100187927 on Build GRCh38
rs768339013 Chr.4: 100187735-100187735 on Build GRCh38
rs58706659 Chr.4: 100187719-100187719 on Build GRCh38
rs752478332 Chr.4: 100187944-100187944 on Build GRCh38
rs770239719 Chr.4: 100187852-100187852 on Build GRCh38
rs140082687 Chr.4: 100187821-100187821 on Build GRCh38
rs750339383 Chr.4: 100187709-100187709 on Build GRCh38
rs778623824 Chr.4: 100187830-100187830 on Build GRCh38
rs538207535 Chr.4: 100187783-100187783 on Build GRCh38
rs760029984 Chr.4: 100187759-100187759 on Build GRCh38
rs1006907464 Chr.4: 100187894-100187894 on Build GRCh38
rs770325787 Chr.4: 100187766-100187766 on Build GRCh38
rs1022340614 Chr.4: 100187913-100187913 on Build GRCh38
rs997463567 Chr.4: 100187867-100187867 on Build GRCh38
rs767707745 Chr.4: 100187790-100187790 on Build GRCh38
rs754844589 Chr.4: 100187937-100187937 on Build GRCh38
rs144358521 Chr.4: 100187736-100187736 on Build GRCh38
rs369312562 Chr.4: 100187956-100187956 on Build GRCh38
rs201713115 Chr.4: 100187720-100187720 on Build GRCh38
rs771881964 Chr.4: 100187774-100187776 on Build GRCh38
rs767838916 Chr.4: 100187891-100187891 on Build GRCh38
rs374448501 Chr.4: 100187878-100187878 on Build GRCh38
rs200392653 Chr.4: 100187868-100187868 on Build GRCh38
rs889559109 Chr.4: 100187560-100187560 on Build GRCh38
rs993963265 Chr.4: 100187718-100187718 on Build GRCh38
rs111601345 Chr.4: 100187801-100187801 on Build GRCh38
rs923987695 Chr.4: 100187817-100187817 on Build GRCh38
rs199896226 Chr.4: 100187704-100187704 on Build GRCh38
rs762190497 Chr.4: 100187890-100187890 on Build GRCh38
rs201805787 Chr.4: 100187582-100187582 on Build GRCh38
rs754789622 Chr.4: 100187822-100187822 on Build GRCh38
rs867300369 Chr.4: 100187591-100187591 on Build GRCh38
rs746510127 Chr.4: 100187840-100187840 on Build GRCh38
rs1031208980 Chr.4: 100187962-100187962 on Build GRCh38
rs866882589 Chr.4: 100187562-100187562 on Build GRCh38
rs971106686 Chr.4: 100187782-100187782 on Build GRCh38
rs772714803 Chr.4: 100187758-100187758 on Build GRCh38
rs796856504 Chr.4: 100187724-100187724 on Build GRCh38
rs542290168 Chr.4: 100187808-100187808 on Build GRCh38
rs760034691 Chr.4: 100187627-100187627 on Build GRCh38
rs56763427 Chr.4: 100187719-100187719 on Build GRCh38
rs753105356 Chr.4: 100187662-100187662 on Build GRCh38
rs1049405609 Chr.4: 100187532-100187532 on Build GRCh38
rs141348282 Chr.4: 100187965-100187965 on Build GRCh38
rs11553154 Chr.4: 100187777-100187777 on Build GRCh38
rs750509500 Chr.4: 100187791-100187791 on Build GRCh38
rs745323788 Chr.4: 100187768-100187768 on Build GRCh38
rs201313366 Chr.4: 100187835-100187835 on Build GRCh38
rs565851948 Chr.4: 100187601-100187601 on Build GRCh38
rs577739259 Chr.4: 100187647-100187647 on Build GRCh38
rs375394393 Chr.4: 100187586-100187586 on Build GRCh38
rs200928469 Chr.4: 100187862-100187862 on Build GRCh38
rs112236091 Chr.4: 100187933-100187933 on Build GRCh38
rs763553051 Chr.4: 100187570-100187570 on Build GRCh38
rs751725054 Chr.4: 100187677-100187677 on Build GRCh38
rs201201560 Chr.4: 100187960-100187960 on Build GRCh38
rs142511472 Chr.4: 100187806-100187806 on Build GRCh38
rs796921367 Chr.4: 100187719-100187720 on Build GRCh38
rs368682316 Chr.4: 100187932-100187932 on Build GRCh38
rs776091624 Chr.4: 100187853-100187853 on Build GRCh38
rs140725424 Chr.4: 100187931-100187931 on Build GRCh38
rs1004724636 Chr.4: 100187803-100187803 on Build GRCh38
rs183155282 Chr.4: 100187966-100187966 on Build GRCh38
rs764385244 Chr.4: 100187666-100187666 on Build GRCh38
rs201592671 Chr.4: 100187663-100187663 on Build GRCh38
rs374315461 Chr.4: 100187685-100187685 on Build GRCh38
rs774012796 Chr.4: 100187747-100187748 on Build GRCh38
rs768990716 Chr.4: 100187871-100187871 on Build GRCh38
rs751851511 Chr.4: 100187784-100187784 on Build GRCh38
rs747752742 Chr.4: 100187748-100187748 on Build GRCh38
rs557451955 Chr.4: 100187721-100187721 on Build GRCh38
rs376338276 Chr.4: 100187729-100187729 on Build GRCh38
rs144488942 Chr.4: 100187940-100187940 on Build GRCh38
rs561838544 Chr.4: 100187903-100187904 on Build GRCh38
rs954060552 Chr.4: 100187872-100187872 on Build GRCh38

View Full Product Details