Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112187356-112187864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf32
Entrez Gene ID
132720
Gene Name
chromosome 4 open reading frame 32
Gene Aliases
-
UniGene
Hs.23439
Gene Chromosome Location
Chr.4: 112145397-112189081 on Build GRCh38
Transcripts
NM_152400.2 XM_017007732.1

SNP Detail

SNP ID Chromosome Location
rs571917132 Chr.4: 112187457-112187457 on Build GRCh38
rs996239428 Chr.4: 112187476-112187476 on Build GRCh38
rs537668481 Chr.4: 112187804-112187804 on Build GRCh38
rs182275214 Chr.4: 112187739-112187739 on Build GRCh38
rs148469467 Chr.4: 112187429-112187429 on Build GRCh38
rs1020240266 Chr.4: 112187560-112187560 on Build GRCh38
rs894514824 Chr.4: 112187597-112187597 on Build GRCh38
rs1011540792 Chr.4: 112187600-112187600 on Build GRCh38
rs929693566 Chr.4: 112187519-112187519 on Build GRCh38
rs145521720 Chr.4: 112187646-112187647 on Build GRCh38
rs963701725 Chr.4: 112187448-112187448 on Build GRCh38
rs78815360 Chr.4: 112187649-112187650 on Build GRCh38
rs942257425 Chr.4: 112187718-112187718 on Build GRCh38
rs992449253 Chr.4: 112187833-112187833 on Build GRCh38
rs1055636115 Chr.4: 112187585-112187585 on Build GRCh38
rs958333952 Chr.4: 112187832-112187832 on Build GRCh38
rs142418043 Chr.4: 112187767-112187768 on Build GRCh38
rs973785311 Chr.4: 112187751-112187751 on Build GRCh38
rs187277445 Chr.4: 112187786-112187786 on Build GRCh38
rs1043410058 Chr.4: 112187610-112187610 on Build GRCh38
rs571772206 Chr.4: 112187466-112187466 on Build GRCh38
rs547767542 Chr.4: 112187580-112187580 on Build GRCh38
rs1017879720 Chr.4: 112187735-112187735 on Build GRCh38
rs1027039781 Chr.4: 112187769-112187769 on Build GRCh38
rs1051810311 Chr.4: 112187752-112187752 on Build GRCh38
rs926936282 Chr.4: 112187587-112187587 on Build GRCh38
rs918086674 Chr.4: 112187614-112187614 on Build GRCh38
rs1030526669 Chr.4: 112187693-112187693 on Build GRCh38
rs564456205 Chr.4: 112187405-112187405 on Build GRCh38
rs999314345 Chr.4: 112187655-112187655 on Build GRCh38
rs986365440 Chr.4: 112187436-112187436 on Build GRCh38
rs910912851 Chr.4: 112187439-112187439 on Build GRCh38
rs567097578 Chr.4: 112187797-112187797 on Build GRCh38
rs982922579 Chr.4: 112187792-112187792 on Build GRCh38
rs565982840 Chr.4: 112187699-112187699 on Build GRCh38
rs202090416 Chr.4: 112187646-112187646 on Build GRCh38
rs908145921 Chr.4: 112187677-112187677 on Build GRCh38
rs928445786 Chr.4: 112187563-112187563 on Build GRCh38
rs1048351038 Chr.4: 112187671-112187671 on Build GRCh38
rs903285030 Chr.4: 112187613-112187613 on Build GRCh38
rs919689094 Chr.4: 112187509-112187509 on Build GRCh38
rs762918054 Chr.4: 112187413-112187413 on Build GRCh38
rs890543209 Chr.4: 112187753-112187753 on Build GRCh38
rs954493215 Chr.4: 112187546-112187546 on Build GRCh38
rs969032035 Chr.4: 112187561-112187561 on Build GRCh38
rs761282088 Chr.4: 112187770-112187770 on Build GRCh38
rs1007851965 Chr.4: 112187726-112187726 on Build GRCh38
rs1010161297 Chr.4: 112187554-112187554 on Build GRCh38
rs12511775 Chr.4: 112187524-112187524 on Build GRCh38
rs949591906 Chr.4: 112187648-112187648 on Build GRCh38
rs1000500624 Chr.4: 112187802-112187802 on Build GRCh38

View Full Product Details