Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 121907221-121907728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPC3
Entrez Gene ID
7222
Gene Name
transient receptor potential cation channel subfamily C member 3
Gene Aliases
SCA41, TRP3
UniGene
Hs.150981
Gene Chromosome Location
Chr.4: 121879028-121952060 on Build GRCh38
Transcripts
NM_001130698.1 NM_003305.2 XM_011532217.2 XM_011532218.2 XM_017008578.1 XM_017008579.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1594243 c.1853C>A p.A618D Chr.4: 121907507-121907507 on Build GRCh38
6098995 c.1858G>T p.V620L Chr.4: 121907502-121907502 on Build GRCh38
5351041 c.1611A>G p.I537M Chr.4: 121907530-121907530 on Build GRCh38
4472836 c.1590C>T p.L530L Chr.4: 121907551-121907551 on Build GRCh38
6098996 c.1639G>T p.V547L Chr.4: 121907502-121907502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55712749 Chr.4: 121907440-121907440 on Build GRCh38
rs750724885 Chr.4: 121907630-121907630 on Build GRCh38
rs774191611 Chr.4: 121907429-121907429 on Build GRCh38
rs751981744 Chr.4: 121907531-121907531 on Build GRCh38
rs200853037 Chr.4: 121907392-121907392 on Build GRCh38
rs750768607 Chr.4: 121907298-121907298 on Build GRCh38
rs535357089 Chr.4: 121907248-121907248 on Build GRCh38
rs1044006116 Chr.4: 121907283-121907283 on Build GRCh38
rs760512827 Chr.4: 121907608-121907608 on Build GRCh38
rs938373017 Chr.4: 121907320-121907320 on Build GRCh38
rs973557839 Chr.4: 121907650-121907650 on Build GRCh38
rs899152105 Chr.4: 121907636-121907636 on Build GRCh38
rs768836131 Chr.4: 121907611-121907611 on Build GRCh38
rs35995428 Chr.4: 121907662-121907663 on Build GRCh38
rs202011680 Chr.4: 121907469-121907469 on Build GRCh38
rs891551044 Chr.4: 121907466-121907466 on Build GRCh38
rs545557485 Chr.4: 121907619-121907619 on Build GRCh38
rs745733011 Chr.4: 121907595-121907595 on Build GRCh38
rs760849828 Chr.4: 121907472-121907472 on Build GRCh38
rs145337107 Chr.4: 121907597-121907597 on Build GRCh38
rs770631116 Chr.4: 121907580-121907580 on Build GRCh38
rs202172262 Chr.4: 121907302-121907302 on Build GRCh38
rs747700292 Chr.4: 121907327-121907327 on Build GRCh38
rs777612960 Chr.4: 121907573-121907573 on Build GRCh38
rs767852687 Chr.4: 121907572-121907573 on Build GRCh38
rs199896927 Chr.4: 121907384-121907384 on Build GRCh38
rs771817585 Chr.4: 121907449-121907449 on Build GRCh38
rs530705810 Chr.4: 121907649-121907649 on Build GRCh38
rs201266386 Chr.4: 121907285-121907285 on Build GRCh38
rs372025790 Chr.4: 121907329-121907329 on Build GRCh38
rs374475823 Chr.4: 121907291-121907291 on Build GRCh38
rs202161034 Chr.4: 121907607-121907607 on Build GRCh38
rs750951273 Chr.4: 121907511-121907511 on Build GRCh38
rs770970191 Chr.4: 121907419-121907419 on Build GRCh38
rs765770767 Chr.4: 121907492-121907492 on Build GRCh38
rs778895001 Chr.4: 121907583-121907583 on Build GRCh38
rs532663355 Chr.4: 121907666-121907666 on Build GRCh38
rs145710040 Chr.4: 121907579-121907579 on Build GRCh38
rs575087117 Chr.4: 121907437-121907437 on Build GRCh38
rs951974 Chr.4: 121907639-121907639 on Build GRCh38
rs148779272 Chr.4: 121907647-121907647 on Build GRCh38
rs377005731 Chr.4: 121907623-121907623 on Build GRCh38
rs777376339 Chr.4: 121907335-121907335 on Build GRCh38
rs761921081 Chr.4: 121907487-121907487 on Build GRCh38
rs748829521 Chr.4: 121907352-121907352 on Build GRCh38
rs1003390981 Chr.4: 121907652-121907652 on Build GRCh38
rs371940806 Chr.4: 121907479-121907479 on Build GRCh38
rs199969572 Chr.4: 121907276-121907276 on Build GRCh38
rs374961402 Chr.4: 121907512-121907512 on Build GRCh38
rs780214352 Chr.4: 121907521-121907521 on Build GRCh38
rs570535867 Chr.4: 121907659-121907659 on Build GRCh38
rs544037886 Chr.4: 121907536-121907536 on Build GRCh38
rs969563484 Chr.4: 121907694-121907694 on Build GRCh38
rs780885360 Chr.4: 121907312-121907312 on Build GRCh38
rs180961013 Chr.4: 121907632-121907632 on Build GRCh38
rs200377323 Chr.4: 121907613-121907613 on Build GRCh38
rs200357045 Chr.4: 121907475-121907475 on Build GRCh38
rs375144966 Chr.4: 121907454-121907454 on Build GRCh38
rs566087204 Chr.4: 121907701-121907701 on Build GRCh38
rs550786362 Chr.4: 121907653-121907653 on Build GRCh38
rs948310639 Chr.4: 121907278-121907278 on Build GRCh38
rs904403121 Chr.4: 121907600-121907600 on Build GRCh38
rs201225851 Chr.4: 121907598-121907598 on Build GRCh38
rs552357201 Chr.4: 121907674-121907674 on Build GRCh38

View Full Product Details