Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 128287934-128288440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGRMC2
Entrez Gene ID
10424
Gene Name
progesterone receptor membrane component 2
Gene Aliases
DG6, PMBP
UniGene
Hs.507910
Gene Chromosome Location
Chr.4: 128269237-128288829 on Build GRCh38
Transcripts
NM_006320.4 XM_011531533.1

SNP Detail

SNP ID Chromosome Location
rs533436539 Chr.4: 128288290-128288290 on Build GRCh38
rs546456515 Chr.4: 128288340-128288340 on Build GRCh38
rs574295224 Chr.4: 128288247-128288247 on Build GRCh38
rs567012130 Chr.4: 128288316-128288316 on Build GRCh38
rs935431289 Chr.4: 128288018-128288018 on Build GRCh38
rs979622683 Chr.4: 128287983-128287983 on Build GRCh38
rs557396229 Chr.4: 128288232-128288232 on Build GRCh38
rs569761231 Chr.4: 128288061-128288061 on Build GRCh38
rs1010744490 Chr.4: 128288153-128288153 on Build GRCh38
rs922218814 Chr.4: 128287981-128287981 on Build GRCh38
rs996979649 Chr.4: 128288279-128288279 on Build GRCh38
rs962293507 Chr.4: 128288200-128288200 on Build GRCh38
rs562280778 Chr.4: 128287955-128287955 on Build GRCh38
rs1043786337 Chr.4: 128288157-128288157 on Build GRCh38
rs1028083703 Chr.4: 128288202-128288202 on Build GRCh38
rs1010567678 Chr.4: 128287959-128287959 on Build GRCh38
rs928109561 Chr.4: 128288216-128288216 on Build GRCh38
rs745640883 Chr.4: 128288327-128288327 on Build GRCh38
rs187649117 Chr.4: 128288294-128288294 on Build GRCh38
rs892416768 Chr.4: 128288146-128288146 on Build GRCh38
rs2893244 Chr.4: 128288295-128288295 on Build GRCh38
rs896489016 Chr.4: 128288030-128288030 on Build GRCh38
rs969446030 Chr.4: 128288207-128288207 on Build GRCh38
rs886919898 Chr.4: 128288130-128288130 on Build GRCh38
rs1010216349 Chr.4: 128288412-128288412 on Build GRCh38
rs949376929 Chr.4: 128288055-128288055 on Build GRCh38
rs548634681 Chr.4: 128288134-128288134 on Build GRCh38
rs552888334 Chr.4: 128288170-128288170 on Build GRCh38
rs536732499 Chr.4: 128288087-128288087 on Build GRCh38
rs557192654 Chr.4: 128288395-128288395 on Build GRCh38
rs970861238 Chr.4: 128288317-128288317 on Build GRCh38
rs577756120 Chr.4: 128288249-128288249 on Build GRCh38
rs948156042 Chr.4: 128288105-128288105 on Build GRCh38
rs946653365 Chr.4: 128288239-128288239 on Build GRCh38
rs995401440 Chr.4: 128288282-128288282 on Build GRCh38
rs1052217669 Chr.4: 128288136-128288136 on Build GRCh38
rs960665102 Chr.4: 128288406-128288406 on Build GRCh38
rs1004377100 Chr.4: 128288154-128288154 on Build GRCh38
rs201667613 Chr.4: 128288292-128288292 on Build GRCh38
rs571549261 Chr.4: 128288366-128288366 on Build GRCh38
rs979654806 Chr.4: 128288312-128288312 on Build GRCh38
rs554076250 Chr.4: 128288315-128288315 on Build GRCh38
rs960778875 Chr.4: 128288297-128288297 on Build GRCh38
rs973659210 Chr.4: 128288201-128288201 on Build GRCh38
rs28522979 Chr.4: 128288160-128288160 on Build GRCh38
rs988636310 Chr.4: 128288213-128288213 on Build GRCh38
rs41298547 Chr.4: 128288159-128288159 on Build GRCh38
rs146352849 Chr.4: 128288323-128288323 on Build GRCh38
rs538572766 Chr.4: 128288383-128288383 on Build GRCh38
rs897969626 Chr.4: 128288280-128288280 on Build GRCh38
rs999905045 Chr.4: 128288368-128288368 on Build GRCh38
rs41298551 Chr.4: 128287959-128287959 on Build GRCh38
rs996568737 Chr.4: 128287976-128287976 on Build GRCh38
rs953631533 Chr.4: 128288174-128288174 on Build GRCh38
rs925422246 Chr.4: 128288000-128288000 on Build GRCh38
rs951163487 Chr.4: 128287958-128287958 on Build GRCh38
rs949494599 Chr.4: 128288228-128288228 on Build GRCh38
rs755435949 Chr.4: 128288211-128288211 on Build GRCh38
rs1026799944 Chr.4: 128288094-128288094 on Build GRCh38
rs904193603 Chr.4: 128288365-128288365 on Build GRCh38
rs755587115 Chr.4: 128288262-128288262 on Build GRCh38
rs760650783 Chr.4: 128288344-128288344 on Build GRCh38
rs1049687672 Chr.4: 128288355-128288355 on Build GRCh38
rs892749798 Chr.4: 128288397-128288397 on Build GRCh38
rs1015121498 Chr.4: 128288276-128288276 on Build GRCh38
rs931248988 Chr.4: 128288242-128288242 on Build GRCh38
rs995056901 Chr.4: 128288352-128288352 on Build GRCh38
rs995371415 Chr.4: 128288091-128288091 on Build GRCh38
rs939212346 Chr.4: 128288125-128288125 on Build GRCh38
rs932213354 Chr.4: 128287982-128287982 on Build GRCh38
rs1029440969 Chr.4: 128288163-128288163 on Build GRCh38
rs41297943 Chr.4: 128288272-128288272 on Build GRCh38
rs1052503688 Chr.4: 128288022-128288022 on Build GRCh38
rs920569386 Chr.4: 128288066-128288066 on Build GRCh38
rs772043616 Chr.4: 128288336-128288336 on Build GRCh38
rs539849415 Chr.4: 128288196-128288196 on Build GRCh38
rs1041267942 Chr.4: 128288269-128288269 on Build GRCh38
rs557848513 Chr.4: 128288225-128288225 on Build GRCh38
rs1048270865 Chr.4: 128288245-128288245 on Build GRCh38
rs1048378432 Chr.4: 128288328-128288328 on Build GRCh38
rs551089951 Chr.4: 128288035-128288035 on Build GRCh38
rs996610597 Chr.4: 128288346-128288346 on Build GRCh38
rs1027976341 Chr.4: 128288171-128288171 on Build GRCh38
rs532986486 Chr.4: 128288027-128288027 on Build GRCh38
rs751804101 Chr.4: 128288194-128288194 on Build GRCh38
rs543953017 Chr.4: 128288236-128288236 on Build GRCh38
rs559757521 Chr.4: 128288001-128288001 on Build GRCh38
rs988363231 Chr.4: 128288305-128288305 on Build GRCh38
rs939918631 Chr.4: 128288337-128288337 on Build GRCh38
rs996560295 Chr.4: 128288166-128288166 on Build GRCh38
rs1049470760 Chr.4: 128288351-128288351 on Build GRCh38
rs41298545 Chr.4: 128288238-128288238 on Build GRCh38
rs577856189 Chr.4: 128288180-128288180 on Build GRCh38
rs1019717916 Chr.4: 128287962-128287962 on Build GRCh38
rs957177114 Chr.4: 128288303-128288303 on Build GRCh38
rs1045044930 Chr.4: 128288056-128288056 on Build GRCh38
rs913749527 Chr.4: 128288309-128288309 on Build GRCh38
rs1043619897 Chr.4: 128288240-128288240 on Build GRCh38
rs965040310 Chr.4: 128287974-128287974 on Build GRCh38
rs1015069140 Chr.4: 128288199-128288199 on Build GRCh38
rs928027415 Chr.4: 128288116-128288116 on Build GRCh38
rs905451265 Chr.4: 128288364-128288364 on Build GRCh38
rs1040904369 Chr.4: 128288338-128288338 on Build GRCh38
rs748848640 Chr.4: 128288274-128288274 on Build GRCh38
rs925453364 Chr.4: 128288209-128288209 on Build GRCh38
rs41298549 Chr.4: 128287977-128287977 on Build GRCh38
rs980799777 Chr.4: 128288319-128288319 on Build GRCh38

View Full Product Details