Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 147638668-147639180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRMT9
Entrez Gene ID
90826
Gene Name
protein arginine methyltransferase 9
Gene Aliases
PRMT10
UniGene
Hs.591692
Gene Chromosome Location
Chr.4: 147635626-147684230 on Build GRCh38
Transcripts
NM_001304458.1 NM_138364.3 XM_017008823.1 XM_017008824.1 XM_017008825.1 XM_017008826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4934709 c.2388A>G p.E796E Chr.4: 147638682-147638682 on Build GRCh38
257811 c.2386G>T p.E796* Chr.4: 147638684-147638684 on Build GRCh38
4915942 c.2399T>C p.L800S Chr.4: 147638671-147638671 on Build GRCh38
5635793 c.2206A>G p.I736V Chr.4: 147639076-147639076 on Build GRCh38
273153 c.2288C>T p.T763I Chr.4: 147638994-147638994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373065313 Chr.4: 147638788-147638788 on Build GRCh38
rs767833597 Chr.4: 147638985-147638985 on Build GRCh38
rs539730089 Chr.4: 147638751-147638751 on Build GRCh38
rs780864233 Chr.4: 147639055-147639055 on Build GRCh38
rs970665081 Chr.4: 147639088-147639088 on Build GRCh38
rs776481526 Chr.4: 147638916-147638916 on Build GRCh38
rs978125968 Chr.4: 147638807-147638807 on Build GRCh38
rs751309669 Chr.4: 147638950-147638950 on Build GRCh38
rs111550773 Chr.4: 147639097-147639097 on Build GRCh38
rs959637698 Chr.4: 147639064-147639064 on Build GRCh38
rs200951430 Chr.4: 147639051-147639051 on Build GRCh38
rs577054395 Chr.4: 147639136-147639136 on Build GRCh38
rs142221362 Chr.4: 147639046-147639046 on Build GRCh38
rs765966573 Chr.4: 147638927-147638930 on Build GRCh38
rs374848916 Chr.4: 147639029-147639029 on Build GRCh38
rs34852471 Chr.4: 147639059-147639059 on Build GRCh38
rs1000606159 Chr.4: 147638875-147638875 on Build GRCh38
rs765305520 Chr.4: 147639125-147639125 on Build GRCh38
rs753035624 Chr.4: 147638990-147638990 on Build GRCh38
rs752006546 Chr.4: 147638724-147638724 on Build GRCh38
rs907913003 Chr.4: 147639010-147639010 on Build GRCh38
rs760207279 Chr.4: 147638767-147638767 on Build GRCh38
rs77210874 Chr.4: 147638692-147638692 on Build GRCh38
rs138669035 Chr.4: 147639072-147639072 on Build GRCh38
rs769798180 Chr.4: 147638931-147638931 on Build GRCh38
rs547212861 Chr.4: 147639121-147639121 on Build GRCh38
rs201431944 Chr.4: 147638933-147638933 on Build GRCh38
rs759209520 Chr.4: 147638959-147638959 on Build GRCh38
rs983858347 Chr.4: 147638996-147638996 on Build GRCh38
rs11557361 Chr.4: 147639042-147639042 on Build GRCh38
rs747929266 Chr.4: 147639056-147639056 on Build GRCh38
rs199739695 Chr.4: 147638925-147638925 on Build GRCh38
rs1000553842 Chr.4: 147639139-147639139 on Build GRCh38
rs902269265 Chr.4: 147638873-147638873 on Build GRCh38
rs746072069 Chr.4: 147638789-147638789 on Build GRCh38
rs751200776 Chr.4: 147639012-147639012 on Build GRCh38
rs759111616 Chr.4: 147639086-147639086 on Build GRCh38
rs1054064428 Chr.4: 147638844-147638844 on Build GRCh38
rs372543562 Chr.4: 147638737-147638737 on Build GRCh38
rs770627113 Chr.4: 147639082-147639082 on Build GRCh38
rs756590512 Chr.4: 147639021-147639021 on Build GRCh38
rs773903910 Chr.4: 147639084-147639084 on Build GRCh38
rs765959937 Chr.4: 147638945-147638945 on Build GRCh38
rs771551945 Chr.4: 147638722-147638722 on Build GRCh38
rs758138750 Chr.4: 147638698-147638698 on Build GRCh38
rs754390577 Chr.4: 147639101-147639101 on Build GRCh38
rs1057510390 Chr.4: 147638750-147638750 on Build GRCh38
rs990763061 Chr.4: 147638756-147638756 on Build GRCh38
rs949400754 Chr.4: 147638803-147638803 on Build GRCh38
rs767137695 Chr.4: 147639089-147639089 on Build GRCh38
rs754697158 Chr.4: 147639112-147639117 on Build GRCh38
rs764720937 Chr.4: 147638696-147638696 on Build GRCh38
rs76148346 Chr.4: 147638747-147638747 on Build GRCh38
rs936484667 Chr.4: 147638837-147638837 on Build GRCh38
rs768393895 Chr.4: 147638791-147638791 on Build GRCh38
rs750461228 Chr.4: 147639133-147639133 on Build GRCh38
rs924033574 Chr.4: 147638829-147638829 on Build GRCh38
rs978072139 Chr.4: 147639093-147639093 on Build GRCh38
rs900396841 Chr.4: 147638929-147638929 on Build GRCh38
rs766083386 Chr.4: 147638718-147638718 on Build GRCh38
rs771669448 Chr.4: 147639145-147639145 on Build GRCh38
rs753023087 Chr.4: 147639091-147639091 on Build GRCh38
rs772271661 Chr.4: 147639053-147639053 on Build GRCh38
rs77293186 Chr.4: 147638741-147638741 on Build GRCh38
rs199696814 Chr.4: 147638934-147638934 on Build GRCh38
rs762965104 Chr.4: 147639075-147639075 on Build GRCh38
rs749305882 Chr.4: 147638786-147638786 on Build GRCh38
rs758203300 Chr.4: 147638717-147638720 on Build GRCh38
rs17675537 Chr.4: 147638697-147638697 on Build GRCh38
rs567253337 Chr.4: 147638843-147638843 on Build GRCh38
rs753948433 Chr.4: 147639036-147639036 on Build GRCh38
rs753198549 Chr.4: 147638742-147638742 on Build GRCh38

View Full Product Details