Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 155208526-155209023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPY2R
Entrez Gene ID
4887
Gene Name
neuropeptide Y receptor Y2
Gene Aliases
NPY2-R
UniGene
Hs.37125
Gene Chromosome Location
Chr.4: 155173514-155217078 on Build GRCh38
Transcripts
NM_000910.3 XM_005263033.4 XM_005263034.4

SNP Detail

SNP ID Chromosome Location
rs913560957 Chr.4: 155208900-155208900 on Build GRCh38
rs1026232171 Chr.4: 155208760-155208760 on Build GRCh38
rs548078543 Chr.4: 155208996-155208996 on Build GRCh38
rs756390949 Chr.4: 155208925-155208925 on Build GRCh38
rs532417469 Chr.4: 155208625-155208625 on Build GRCh38
rs986113299 Chr.4: 155208566-155208566 on Build GRCh38
rs949740162 Chr.4: 155208880-155208880 on Build GRCh38
rs941272069 Chr.4: 155208911-155208911 on Build GRCh38
rs184295299 Chr.4: 155208578-155208578 on Build GRCh38
rs925679139 Chr.4: 155208612-155208612 on Build GRCh38
rs576470334 Chr.4: 155208830-155208830 on Build GRCh38
rs1016796765 Chr.4: 155208719-155208719 on Build GRCh38
rs557683990 Chr.4: 155208812-155208812 on Build GRCh38
rs1008976522 Chr.4: 155208811-155208811 on Build GRCh38
rs771203027 Chr.4: 155208959-155208959 on Build GRCh38
rs1007277806 Chr.4: 155208960-155208960 on Build GRCh38
rs968288012 Chr.4: 155208778-155208778 on Build GRCh38
rs1018923172 Chr.4: 155208771-155208771 on Build GRCh38
rs570646284 Chr.4: 155208599-155208599 on Build GRCh38
rs1047032768 Chr.4: 155208682-155208682 on Build GRCh38
rs541782772 Chr.4: 155208923-155208923 on Build GRCh38
rs920953395 Chr.4: 155208549-155208549 on Build GRCh38
rs1005372331 Chr.4: 155208714-155208714 on Build GRCh38
rs539470659 Chr.4: 155208758-155208758 on Build GRCh38
rs913663774 Chr.4: 155208611-155208611 on Build GRCh38
rs994661996 Chr.4: 155208757-155208757 on Build GRCh38
rs961425502 Chr.4: 155208751-155208751 on Build GRCh38
rs946871480 Chr.4: 155208579-155208579 on Build GRCh38
rs72972775 Chr.4: 155208953-155208953 on Build GRCh38
rs934374743 Chr.4: 155208833-155208833 on Build GRCh38
rs750361634 Chr.4: 155208629-155208629 on Build GRCh38
rs930893022 Chr.4: 155208955-155208955 on Build GRCh38
rs895476218 Chr.4: 155208654-155208654 on Build GRCh38
rs968235492 Chr.4: 155208977-155208977 on Build GRCh38
rs979538446 Chr.4: 155208780-155208780 on Build GRCh38
rs965223988 Chr.4: 155208825-155208825 on Build GRCh38
rs985506759 Chr.4: 155208763-155208763 on Build GRCh38
rs1018241080 Chr.4: 155208970-155208970 on Build GRCh38
rs543891392 Chr.4: 155208859-155208859 on Build GRCh38
rs796119959 Chr.4: 155208680-155208682 on Build GRCh38
rs113342908 Chr.4: 155208982-155208982 on Build GRCh38
rs955547990 Chr.4: 155208762-155208762 on Build GRCh38
rs1019895812 Chr.4: 155208813-155208813 on Build GRCh38
rs1044698510 Chr.4: 155208667-155208667 on Build GRCh38
rs574157658 Chr.4: 155208921-155208921 on Build GRCh38
rs1043894851 Chr.4: 155208610-155208610 on Build GRCh38
rs1055667037 Chr.4: 155208948-155208948 on Build GRCh38
rs533618101 Chr.4: 155208972-155208972 on Build GRCh38
rs905756661 Chr.4: 155208899-155208899 on Build GRCh38
rs927677435 Chr.4: 155208944-155208944 on Build GRCh38
rs1044091744 Chr.4: 155208898-155208898 on Build GRCh38
rs938206594 Chr.4: 155208947-155208947 on Build GRCh38
rs555733239 Chr.4: 155208915-155208915 on Build GRCh38
rs767637348 Chr.4: 155208922-155208922 on Build GRCh38
rs934422286 Chr.4: 155208622-155208622 on Build GRCh38
rs1013962119 Chr.4: 155208656-155208656 on Build GRCh38
rs988966603 Chr.4: 155208853-155208853 on Build GRCh38
rs533860510 Chr.4: 155208757-155208758 on Build GRCh38
rs905807559 Chr.4: 155208713-155208713 on Build GRCh38
rs886566385 Chr.4: 155208807-155208807 on Build GRCh38
rs1053422523 Chr.4: 155208607-155208607 on Build GRCh38
rs752739739 Chr.4: 155208924-155208924 on Build GRCh38

View Full Product Details