Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 155215698-155216195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPY2R
Entrez Gene ID
4887
Gene Name
neuropeptide Y receptor Y2
Gene Aliases
NPY2-R
UniGene
Hs.37125
Gene Chromosome Location
Chr.4: 155173514-155217078 on Build GRCh38
Transcripts
NM_000910.3 XM_005263033.4 XM_005263034.4

SNP Detail

SNP ID Chromosome Location
rs925524819 Chr.4: 155215806-155215806 on Build GRCh38
rs535190682 Chr.4: 155215976-155215976 on Build GRCh38
rs567084084 Chr.4: 155216051-155216051 on Build GRCh38
rs551457701 Chr.4: 155215724-155215724 on Build GRCh38
rs1013793116 Chr.4: 155215878-155215878 on Build GRCh38
rs936721318 Chr.4: 155216097-155216097 on Build GRCh38
rs147914972 Chr.4: 155216025-155216025 on Build GRCh38
rs930641897 Chr.4: 155215760-155215760 on Build GRCh38
rs146356210 Chr.4: 155215956-155215956 on Build GRCh38
rs549211966 Chr.4: 155216169-155216169 on Build GRCh38
rs542311949 Chr.4: 155216160-155216160 on Build GRCh38
rs866567221 Chr.4: 155216047-155216047 on Build GRCh38
rs112557709 Chr.4: 155215884-155215884 on Build GRCh38
rs755020232 Chr.4: 155215961-155215961 on Build GRCh38
rs553709267 Chr.4: 155216010-155216010 on Build GRCh38
rs1040123166 Chr.4: 155215787-155215787 on Build GRCh38
rs912618527 Chr.4: 155216049-155216049 on Build GRCh38
rs746152653 Chr.4: 155216144-155216144 on Build GRCh38
rs748036375 Chr.4: 155216013-155216013 on Build GRCh38
rs570148100 Chr.4: 155215807-155215807 on Build GRCh38
rs751304885 Chr.4: 155215900-155215900 on Build GRCh38
rs1012377175 Chr.4: 155216011-155216011 on Build GRCh38
rs1003234866 Chr.4: 155215978-155215978 on Build GRCh38
rs900991478 Chr.4: 155215846-155215846 on Build GRCh38
rs376216837 Chr.4: 155215830-155215830 on Build GRCh38
rs560789363 Chr.4: 155216170-155216170 on Build GRCh38
rs778812506 Chr.4: 155216113-155216113 on Build GRCh38
rs893692115 Chr.4: 155215909-155215909 on Build GRCh38
rs932487437 Chr.4: 155216147-155216147 on Build GRCh38
rs1034431834 Chr.4: 155215915-155215915 on Build GRCh38
rs986832479 Chr.4: 155215741-155215741 on Build GRCh38
rs975401654 Chr.4: 155216074-155216074 on Build GRCh38
rs1025544550 Chr.4: 155216003-155216003 on Build GRCh38
rs912669383 Chr.4: 155215766-155215766 on Build GRCh38
rs556022388 Chr.4: 155215898-155215898 on Build GRCh38
rs545006214 Chr.4: 155216069-155216069 on Build GRCh38
rs766234231 Chr.4: 155215839-155215839 on Build GRCh38
rs918415507 Chr.4: 155215738-155215738 on Build GRCh38
rs921984772 Chr.4: 155216145-155216145 on Build GRCh38
rs754166816 Chr.4: 155215726-155215727 on Build GRCh38
rs369043698 Chr.4: 155215911-155215911 on Build GRCh38
rs141658548 Chr.4: 155216100-155216100 on Build GRCh38
rs116026747 Chr.4: 155216101-155216101 on Build GRCh38
rs193243702 Chr.4: 155215924-155215924 on Build GRCh38
rs945045469 Chr.4: 155216152-155216152 on Build GRCh38
rs949510683 Chr.4: 155216127-155216127 on Build GRCh38
rs1040921878 Chr.4: 155215844-155215844 on Build GRCh38
rs757447204 Chr.4: 155216091-155216091 on Build GRCh38
rs757963258 Chr.4: 155215775-155215775 on Build GRCh38
rs750111636 Chr.4: 155215740-155215740 on Build GRCh38
rs936923235 Chr.4: 155215821-155215821 on Build GRCh38

View Full Product Details