Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 155344001-155344549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP9
Entrez Gene ID
79884
Gene Name
microtubule associated protein 9
Gene Aliases
ASAP
UniGene
Hs.61271
Gene Chromosome Location
Chr.4: 155342658-155377118 on Build GRCh38
Transcripts
NM_001039580.1 XM_006714306.3 XM_011532253.1 XM_011532254.2 XM_011532255.2 XM_017008616.1 XM_017008617.1

SNP Detail

SNP ID Chromosome Location
rs1038397167 Chr.4: 155344205-155344205 on Build GRCh38
rs1044295870 Chr.4: 155344173-155344173 on Build GRCh38
rs916914673 Chr.4: 155344149-155344149 on Build GRCh38
rs201024823 Chr.4: 155344482-155344483 on Build GRCh38
rs567762706 Chr.4: 155344433-155344434 on Build GRCh38
rs990196512 Chr.4: 155344354-155344354 on Build GRCh38
rs769272203 Chr.4: 155344332-155344332 on Build GRCh38
rs557104641 Chr.4: 155344027-155344027 on Build GRCh38
rs897273507 Chr.4: 155344214-155344214 on Build GRCh38
rs947744087 Chr.4: 155344510-155344510 on Build GRCh38
rs747360418 Chr.4: 155344326-155344326 on Build GRCh38
rs1007108810 Chr.4: 155344273-155344273 on Build GRCh38
rs916885858 Chr.4: 155344339-155344339 on Build GRCh38
rs934302986 Chr.4: 155344126-155344126 on Build GRCh38
rs949975248 Chr.4: 155344154-155344154 on Build GRCh38
rs977853646 Chr.4: 155344376-155344377 on Build GRCh38
rs916423864 Chr.4: 155344496-155344496 on Build GRCh38
rs947817998 Chr.4: 155344169-155344169 on Build GRCh38
rs939185353 Chr.4: 155344114-155344114 on Build GRCh38
rs994670205 Chr.4: 155344233-155344233 on Build GRCh38
rs549115790 Chr.4: 155344444-155344444 on Build GRCh38
rs979620773 Chr.4: 155344322-155344322 on Build GRCh38
rs562363336 Chr.4: 155344427-155344427 on Build GRCh38
rs746116504 Chr.4: 155344171-155344171 on Build GRCh38
rs946597739 Chr.4: 155344064-155344064 on Build GRCh38
rs72960496 Chr.4: 155344376-155344376 on Build GRCh38
rs1005377953 Chr.4: 155344186-155344186 on Build GRCh38
rs777007082 Chr.4: 155344340-155344340 on Build GRCh38
rs902118038 Chr.4: 155344267-155344267 on Build GRCh38
rs554703447 Chr.4: 155344196-155344196 on Build GRCh38
rs762321644 Chr.4: 155344407-155344407 on Build GRCh38
rs374346404 Chr.4: 155344280-155344280 on Build GRCh38
rs1056724008 Chr.4: 155344127-155344127 on Build GRCh38
rs953430828 Chr.4: 155344346-155344346 on Build GRCh38
rs968205541 Chr.4: 155344319-155344319 on Build GRCh38
rs979981720 Chr.4: 155344359-155344359 on Build GRCh38
rs6817266 Chr.4: 155344257-155344257 on Build GRCh38
rs75174989 Chr.4: 155344184-155344184 on Build GRCh38
rs941183363 Chr.4: 155344420-155344420 on Build GRCh38
rs925731758 Chr.4: 155344102-155344102 on Build GRCh38
rs186043503 Chr.4: 155344406-155344406 on Build GRCh38
rs955786598 Chr.4: 155344330-155344330 on Build GRCh38
rs760051305 Chr.4: 155344258-155344258 on Build GRCh38
rs573107259 Chr.4: 155344241-155344241 on Build GRCh38
rs1052818476 Chr.4: 155344134-155344134 on Build GRCh38
rs1029638655 Chr.4: 155344254-155344254 on Build GRCh38
rs17032851 Chr.4: 155344087-155344087 on Build GRCh38

View Full Product Details