Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 163345509-163346010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPY5R
Entrez Gene ID
4889
Gene Name
neuropeptide Y receptor Y5
Gene Aliases
NPY5-R, NPYR5, NPYY5-R
UniGene
Hs.519058
Gene Chromosome Location
Chr.4: 163336968-163352277 on Build GRCh38
Transcripts
NM_001317091.1 NM_001317092.1 NM_006174.3 XM_005263038.3 XM_011532015.2 XM_011532017.2 XM_017008255.1 XM_017008256.1

SNP Detail

SNP ID Chromosome Location
rs78488414 Chr.4: 163345742-163345742 on Build GRCh38
rs1015272771 Chr.4: 163345700-163345700 on Build GRCh38
rs1018724703 Chr.4: 163345750-163345750 on Build GRCh38
rs957915177 Chr.4: 163345932-163345932 on Build GRCh38
rs865842400 Chr.4: 163345842-163345842 on Build GRCh38
rs899209327 Chr.4: 163345954-163345954 on Build GRCh38
rs191239515 Chr.4: 163345593-163345593 on Build GRCh38
rs182514099 Chr.4: 163345944-163345944 on Build GRCh38
rs562801332 Chr.4: 163345778-163345778 on Build GRCh38
rs991527858 Chr.4: 163345829-163345829 on Build GRCh38
rs1041771313 Chr.4: 163345538-163345538 on Build GRCh38
rs1028872998 Chr.4: 163345677-163345677 on Build GRCh38
rs925670112 Chr.4: 163345804-163345804 on Build GRCh38
rs142892956 Chr.4: 163345808-163345808 on Build GRCh38
rs924663405 Chr.4: 163345864-163345864 on Build GRCh38
rs953255187 Chr.4: 163345739-163345739 on Build GRCh38
rs980640803 Chr.4: 163345921-163345921 on Build GRCh38
rs994848362 Chr.4: 163345959-163345959 on Build GRCh38
rs962366673 Chr.4: 163345741-163345741 on Build GRCh38
rs4632602 Chr.4: 163345977-163345977 on Build GRCh38
rs554504797 Chr.4: 163345567-163345567 on Build GRCh38
rs971146520 Chr.4: 163345637-163345637 on Build GRCh38
rs11100493 Chr.4: 163345607-163345607 on Build GRCh38
rs903168845 Chr.4: 163345552-163345552 on Build GRCh38
rs944798584 Chr.4: 163345810-163345810 on Build GRCh38
rs36227920 Chr.4: 163345910-163345910 on Build GRCh38
rs995224719 Chr.4: 163345676-163345676 on Build GRCh38
rs367904521 Chr.4: 163345952-163345952 on Build GRCh38
rs936943270 Chr.4: 163345559-163345559 on Build GRCh38
rs894685567 Chr.4: 163345574-163345574 on Build GRCh38
rs548175741 Chr.4: 163345898-163345898 on Build GRCh38
rs928105481 Chr.4: 163345934-163345934 on Build GRCh38
rs935988657 Chr.4: 163345867-163345867 on Build GRCh38
rs898211666 Chr.4: 163345983-163345983 on Build GRCh38
rs1046562934 Chr.4: 163345584-163345584 on Build GRCh38
rs921184211 Chr.4: 163345748-163345748 on Build GRCh38
rs36227919 Chr.4: 163345565-163345565 on Build GRCh38
rs1054457554 Chr.4: 163345897-163345897 on Build GRCh38
rs894751760 Chr.4: 163345908-163345908 on Build GRCh38
rs1032645159 Chr.4: 163345926-163345926 on Build GRCh38
rs540304233 Chr.4: 163345594-163345594 on Build GRCh38
rs552507302 Chr.4: 163345652-163345652 on Build GRCh38
rs977523498 Chr.4: 163345831-163345831 on Build GRCh38
rs953985670 Chr.4: 163345753-163345753 on Build GRCh38
rs912389787 Chr.4: 163345796-163345796 on Build GRCh38
rs1000175280 Chr.4: 163345554-163345554 on Build GRCh38
rs1054060487 Chr.4: 163345562-163345562 on Build GRCh38
rs12503365 Chr.4: 163345751-163345751 on Build GRCh38
rs201933290 Chr.4: 163345944-163345944 on Build GRCh38
rs200253139 Chr.4: 163345575-163345575 on Build GRCh38
rs940797105 Chr.4: 163345940-163345940 on Build GRCh38
rs527401235 Chr.4: 163345911-163345911 on Build GRCh38
rs987073092 Chr.4: 163345763-163345763 on Build GRCh38

View Full Product Details