Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 163545372-163545873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARCH1
Entrez Gene ID
55016
Gene Name
membrane associated ring-CH-type finger 1
Gene Aliases
MARCH-I, RNF171
UniGene
Hs.592804
Gene Chromosome Location
Chr.4: 163524298-164384050 on Build GRCh38
Transcripts
NM_001166373.1 NM_017923.3 XM_011532055.2 XM_017008334.1 XM_017008335.1 XM_017008336.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6845944 c.535C>T p.R179W Chr.4: 163545632-163545632 on Build GRCh38
1154324 c.486C>T p.H162H Chr.4: 163545681-163545681 on Build GRCh38
3335298 c.496G>A p.E166K Chr.4: 163545620-163545620 on Build GRCh38
321587 c.491C>A p.A164E Chr.4: 163545625-163545625 on Build GRCh38
1137158 c.428A>G p.E143G Chr.4: 163545739-163545739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770229511 Chr.4: 163545575-163545575 on Build GRCh38
rs938875099 Chr.4: 163545675-163545675 on Build GRCh38
rs754567412 Chr.4: 163545669-163545669 on Build GRCh38
rs964011851 Chr.4: 163545607-163545607 on Build GRCh38
rs758248171 Chr.4: 163545631-163545631 on Build GRCh38
rs544879154 Chr.4: 163545826-163545826 on Build GRCh38
rs188779015 Chr.4: 163545459-163545459 on Build GRCh38
rs752377581 Chr.4: 163545784-163545784 on Build GRCh38
rs555884097 Chr.4: 163545806-163545806 on Build GRCh38
rs760683696 Chr.4: 163545751-163545751 on Build GRCh38
rs774779525 Chr.4: 163545592-163545592 on Build GRCh38
rs746651742 Chr.4: 163545561-163545561 on Build GRCh38
rs752184817 Chr.4: 163545680-163545680 on Build GRCh38
rs1017287841 Chr.4: 163545567-163545567 on Build GRCh38
rs767084970 Chr.4: 163545778-163545778 on Build GRCh38
rs925965781 Chr.4: 163545467-163545467 on Build GRCh38
rs766055729 Chr.4: 163545452-163545452 on Build GRCh38
rs926176131 Chr.4: 163545797-163545797 on Build GRCh38
rs887481542 Chr.4: 163545837-163545837 on Build GRCh38
rs754584625 Chr.4: 163545564-163545564 on Build GRCh38
rs917294055 Chr.4: 163545844-163545844 on Build GRCh38
rs779396035 Chr.4: 163545738-163545738 on Build GRCh38
rs13130399 Chr.4: 163545672-163545672 on Build GRCh38
rs937364288 Chr.4: 163545479-163545479 on Build GRCh38
rs576850511 Chr.4: 163545765-163545765 on Build GRCh38
rs762848402 Chr.4: 163545767-163545767 on Build GRCh38
rs761659279 Chr.4: 163545693-163545694 on Build GRCh38
rs749804634 Chr.4: 163545726-163545726 on Build GRCh38
rs760767827 Chr.4: 163545620-163545620 on Build GRCh38
rs779695649 Chr.4: 163545553-163545553 on Build GRCh38
rs1055819217 Chr.4: 163545494-163545494 on Build GRCh38
rs11726828 Chr.4: 163545818-163545818 on Build GRCh38
rs1014349052 Chr.4: 163545510-163545510 on Build GRCh38
rs913397218 Chr.4: 163545674-163545674 on Build GRCh38
rs371533234 Chr.4: 163545772-163545772 on Build GRCh38
rs759340258 Chr.4: 163545776-163545776 on Build GRCh38
rs778478425 Chr.4: 163545715-163545715 on Build GRCh38
rs997429282 Chr.4: 163545544-163545544 on Build GRCh38
rs758121722 Chr.4: 163545548-163545548 on Build GRCh38
rs200342233 Chr.4: 163545434-163545434 on Build GRCh38
rs753716249 Chr.4: 163545509-163545509 on Build GRCh38
rs886972560 Chr.4: 163545551-163545551 on Build GRCh38
rs375468000 Chr.4: 163545591-163545591 on Build GRCh38
rs768709767 Chr.4: 163545758-163545758 on Build GRCh38
rs368577075 Chr.4: 163545729-163545729 on Build GRCh38
rs775452411 Chr.4: 163545604-163545604 on Build GRCh38
rs751267508 Chr.4: 163545656-163545656 on Build GRCh38
rs765164081 Chr.4: 163545624-163545624 on Build GRCh38
rs772212250 Chr.4: 163545746-163545746 on Build GRCh38
rs149548665 Chr.4: 163545641-163545641 on Build GRCh38
rs955086758 Chr.4: 163545650-163545650 on Build GRCh38
rs764044725 Chr.4: 163545558-163545575 on Build GRCh38
rs753576734 Chr.4: 163545587-163545587 on Build GRCh38
rs1046926650 Chr.4: 163545550-163545550 on Build GRCh38
rs770669436 Chr.4: 163545840-163545840 on Build GRCh38
rs759643413 Chr.4: 163545597-163545597 on Build GRCh38
rs767683546 Chr.4: 163545603-163545603 on Build GRCh38
rs755588226 Chr.4: 163545681-163545681 on Build GRCh38
rs780539451 Chr.4: 163545571-163545571 on Build GRCh38
rs766214272 Chr.4: 163545771-163545771 on Build GRCh38
rs754785558 Chr.4: 163545625-163545625 on Build GRCh38
rs757565191 Chr.4: 163545449-163545449 on Build GRCh38
rs773819124 Chr.4: 163545584-163545584 on Build GRCh38
rs555303958 Chr.4: 163545831-163545831 on Build GRCh38
rs1056160316 Chr.4: 163545824-163545824 on Build GRCh38
rs757843719 Chr.4: 163545749-163545749 on Build GRCh38
rs1005957610 Chr.4: 163545839-163545839 on Build GRCh38
rs1047433413 Chr.4: 163545404-163545404 on Build GRCh38
rs763508847 Chr.4: 163545630-163545630 on Build GRCh38
rs1017447348 Chr.4: 163545492-163545492 on Build GRCh38
rs539547997 Chr.4: 163545791-163545791 on Build GRCh38
rs200399363 Chr.4: 163545572-163545572 on Build GRCh38
rs749680101 Chr.4: 163545588-163545588 on Build GRCh38

View Full Product Details