Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 168162394-168162939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA10
Entrez Gene ID
11199
Gene Name
annexin A10
Gene Aliases
ANX14
UniGene
Hs.188401
Gene Chromosome Location
Chr.4: 168092525-168187742 on Build GRCh38
Transcripts
NM_007193.4 XM_011531571.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4123354 c.196-1G>A p.? Chr.4: 168162527-168162527 on Build GRCh38
5934857 c.217G>A p.E73K Chr.4: 168162549-168162549 on Build GRCh38
5673720 c.309+1G>T p.? Chr.4: 168162642-168162642 on Build GRCh38
149755 c.211A>C p.M71L Chr.4: 168162543-168162543 on Build GRCh38
1236381 c.250G>T p.A84S Chr.4: 168162582-168162582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002450618 Chr.4: 168162429-168162429 on Build GRCh38
rs886451325 Chr.4: 168162825-168162825 on Build GRCh38
rs1046415330 Chr.4: 168162624-168162624 on Build GRCh38
rs760581323 Chr.4: 168162519-168162519 on Build GRCh38
rs779995435 Chr.4: 168162551-168162551 on Build GRCh38
rs750500036 Chr.4: 168162806-168162806 on Build GRCh38
rs748462667 Chr.4: 168162566-168162566 on Build GRCh38
rs12506651 Chr.4: 168162767-168162767 on Build GRCh38
rs149761708 Chr.4: 168162839-168162839 on Build GRCh38
rs773975383 Chr.4: 168162506-168162506 on Build GRCh38
rs572486143 Chr.4: 168162486-168162486 on Build GRCh38
rs959519186 Chr.4: 168162842-168162842 on Build GRCh38
rs565472425 Chr.4: 168162826-168162826 on Build GRCh38
rs997189306 Chr.4: 168162537-168162537 on Build GRCh38
rs760489040 Chr.4: 168162615-168162615 on Build GRCh38
rs775305529 Chr.4: 168162610-168162610 on Build GRCh38
rs749752027 Chr.4: 168162591-168162591 on Build GRCh38
rs759152398 Chr.4: 168162431-168162431 on Build GRCh38
rs1047691591 Chr.4: 168162778-168162778 on Build GRCh38
rs199709153 Chr.4: 168162574-168162574 on Build GRCh38
rs762886219 Chr.4: 168162605-168162607 on Build GRCh38
rs959548297 Chr.4: 168162461-168162461 on Build GRCh38
rs866431224 Chr.4: 168162912-168162912 on Build GRCh38
rs752513554 Chr.4: 168162518-168162518 on Build GRCh38
rs773715316 Chr.4: 168162593-168162593 on Build GRCh38
rs1054026468 Chr.4: 168162764-168162764 on Build GRCh38
rs771523675 Chr.4: 168162592-168162592 on Build GRCh38
rs938285013 Chr.4: 168162733-168162733 on Build GRCh38
rs755072363 Chr.4: 168162670-168162670 on Build GRCh38
rs1009758114 Chr.4: 168162867-168162867 on Build GRCh38
rs773726818 Chr.4: 168162489-168162489 on Build GRCh38
rs751552468 Chr.4: 168162654-168162654 on Build GRCh38
rs994496500 Chr.4: 168162773-168162774 on Build GRCh38
rs770153355 Chr.4: 168162576-168162576 on Build GRCh38
rs759101033 Chr.4: 168162514-168162514 on Build GRCh38
rs149422799 Chr.4: 168162637-168162637 on Build GRCh38
rs942540006 Chr.4: 168162740-168162740 on Build GRCh38
rs375945140 Chr.4: 168162559-168162559 on Build GRCh38
rs946377102 Chr.4: 168162490-168162490 on Build GRCh38
rs776557451 Chr.4: 168162636-168162636 on Build GRCh38
rs766241479 Chr.4: 168162608-168162609 on Build GRCh38
rs114905136 Chr.4: 168162606-168162606 on Build GRCh38
rs372783431 Chr.4: 168162505-168162505 on Build GRCh38
rs6836994 Chr.4: 168162543-168162543 on Build GRCh38
rs762462705 Chr.4: 168162497-168162497 on Build GRCh38
rs763986667 Chr.4: 168162623-168162623 on Build GRCh38
rs541343416 Chr.4: 168162873-168162873 on Build GRCh38
rs752101271 Chr.4: 168162575-168162575 on Build GRCh38
rs898457169 Chr.4: 168162770-168162770 on Build GRCh38
rs770245234 Chr.4: 168162487-168162487 on Build GRCh38
rs753733601 Chr.4: 168162532-168162532 on Build GRCh38
rs765257405 Chr.4: 168162648-168162648 on Build GRCh38
rs543694474 Chr.4: 168162741-168162741 on Build GRCh38
rs373516528 Chr.4: 168162482-168162482 on Build GRCh38
rs767331211 Chr.4: 168162516-168162516 on Build GRCh38
rs556757096 Chr.4: 168162651-168162651 on Build GRCh38
rs751709412 Chr.4: 168162555-168162555 on Build GRCh38
rs781423027 Chr.4: 168162560-168162560 on Build GRCh38
rs778182459 Chr.4: 168162590-168162590 on Build GRCh38
rs777178434 Chr.4: 168162539-168162539 on Build GRCh38
rs764076161 Chr.4: 168162528-168162528 on Build GRCh38
rs967098139 Chr.4: 168162535-168162535 on Build GRCh38
rs1020056764 Chr.4: 168162526-168162526 on Build GRCh38
rs1042543429 Chr.4: 168162896-168162896 on Build GRCh38

View Full Product Details