Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 176131303-176131812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR17
Entrez Gene ID
116966
Gene Name
WD repeat domain 17
Gene Aliases
-
UniGene
Hs.532056
Gene Chromosome Location
Chr.4: 176065834-176182828 on Build GRCh38
Transcripts
NM_170710.4 NM_181265.3 XM_005262725.3 XM_011531582.2 XM_017007686.1 XM_017007687.1 XM_017007688.1 XM_017007689.1 XM_017007690.1 XM_017007691.1 XM_017007692.1 XM_017007693.1 XM_017007694.1 XM_017007695.1 XM_017007696.1 XM_017007697.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1618609 c.1032C>T p.S344S Chr.4: 176131600-176131600 on Build GRCh38
3940814 c.1132G>C p.D378H Chr.4: 176131700-176131700 on Build GRCh38
420109 c.1165G>C p.D389H Chr.4: 176131733-176131733 on Build GRCh38
1618608 c.986-9T>C p.? Chr.4: 176131545-176131545 on Build GRCh38
4403030 c.1022C>T p.S341F Chr.4: 176131590-176131590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757951281 Chr.4: 176131613-176131613 on Build GRCh38
rs531861423 Chr.4: 176131596-176131596 on Build GRCh38
rs766623832 Chr.4: 176131646-176131646 on Build GRCh38
rs74761018 Chr.4: 176131475-176131475 on Build GRCh38
rs150847957 Chr.4: 176131564-176131564 on Build GRCh38
rs534257571 Chr.4: 176131365-176131365 on Build GRCh38
rs763049246 Chr.4: 176131745-176131745 on Build GRCh38
rs199859479 Chr.4: 176131512-176131512 on Build GRCh38
rs148806126 Chr.4: 176131713-176131713 on Build GRCh38
rs993782508 Chr.4: 176131420-176131420 on Build GRCh38
rs202010584 Chr.4: 176131752-176131752 on Build GRCh38
rs202009749 Chr.4: 176131528-176131528 on Build GRCh38
rs536441769 Chr.4: 176131781-176131781 on Build GRCh38
rs779894613 Chr.4: 176131412-176131412 on Build GRCh38
rs926732133 Chr.4: 176131442-176131442 on Build GRCh38
rs951944214 Chr.4: 176131701-176131701 on Build GRCh38
rs770489051 Chr.4: 176131516-176131516 on Build GRCh38
rs778533672 Chr.4: 176131532-176131533 on Build GRCh38
rs774346585 Chr.4: 176131520-176131520 on Build GRCh38
rs768683694 Chr.4: 176131739-176131739 on Build GRCh38
rs188518085 Chr.4: 176131515-176131515 on Build GRCh38
rs1019534405 Chr.4: 176131491-176131491 on Build GRCh38
rs542851993 Chr.4: 176131551-176131551 on Build GRCh38
rs918113322 Chr.4: 176131746-176131746 on Build GRCh38
rs749978593 Chr.4: 176131353-176131353 on Build GRCh38
rs200095015 Chr.4: 176131720-176131720 on Build GRCh38
rs368563812 Chr.4: 176131689-176131689 on Build GRCh38
rs776397874 Chr.4: 176131741-176131741 on Build GRCh38
rs984819918 Chr.4: 176131540-176131540 on Build GRCh38
rs573863627 Chr.4: 176131431-176131431 on Build GRCh38
rs758713034 Chr.4: 176131546-176131546 on Build GRCh38
rs978271665 Chr.4: 176131760-176131760 on Build GRCh38
rs999965360 Chr.4: 176131361-176131365 on Build GRCh38
rs986435608 Chr.4: 176131479-176131479 on Build GRCh38
rs781512974 Chr.4: 176131757-176131760 on Build GRCh38
rs965996524 Chr.4: 176131419-176131419 on Build GRCh38
rs781684936 Chr.4: 176131571-176131571 on Build GRCh38
rs771945312 Chr.4: 176131605-176131605 on Build GRCh38
rs773398896 Chr.4: 176131620-176131620 on Build GRCh38
rs753428302 Chr.4: 176131702-176131702 on Build GRCh38
rs370197137 Chr.4: 176131601-176131601 on Build GRCh38
rs58826384 Chr.4: 176131339-176131339 on Build GRCh38
rs548704530 Chr.4: 176131587-176131587 on Build GRCh38
rs1026332097 Chr.4: 176131427-176131427 on Build GRCh38
rs145503980 Chr.4: 176131619-176131619 on Build GRCh38
rs774093217 Chr.4: 176131710-176131710 on Build GRCh38
rs372649389 Chr.4: 176131541-176131541 on Build GRCh38
rs566412840 Chr.4: 176131581-176131581 on Build GRCh38
rs762915565 Chr.4: 176131639-176131639 on Build GRCh38
rs1009775918 Chr.4: 176131393-176131393 on Build GRCh38
rs936906139 Chr.4: 176131775-176131775 on Build GRCh38
rs945613576 Chr.4: 176131338-176131338 on Build GRCh38
rs116240732 Chr.4: 176131535-176131535 on Build GRCh38
rs760592732 Chr.4: 176131618-176131618 on Build GRCh38
rs556495752 Chr.4: 176131437-176131437 on Build GRCh38
rs199774879 Chr.4: 176131660-176131660 on Build GRCh38
rs781620231 Chr.4: 176131635-176131635 on Build GRCh38
rs1019694423 Chr.4: 176131726-176131726 on Build GRCh38
rs979873865 Chr.4: 176131771-176131771 on Build GRCh38
rs139987645 Chr.4: 176131610-176131610 on Build GRCh38
rs888856082 Chr.4: 176131373-176131373 on Build GRCh38
rs959444869 Chr.4: 176131474-176131475 on Build GRCh38
rs753004304 Chr.4: 176131664-176131664 on Build GRCh38
rs779966222 Chr.4: 176131733-176131733 on Build GRCh38
rs267600097 Chr.4: 176131557-176131557 on Build GRCh38
rs191468896 Chr.4: 176131344-176131344 on Build GRCh38
rs904001389 Chr.4: 176131651-176131651 on Build GRCh38
rs375973862 Chr.4: 176131736-176131736 on Build GRCh38
rs201581345 Chr.4: 176131776-176131776 on Build GRCh38
rs930755860 Chr.4: 176131538-176131538 on Build GRCh38
rs778121595 Chr.4: 176131695-176131695 on Build GRCh38
rs745691060 Chr.4: 176131603-176131603 on Build GRCh38
rs1028015585 Chr.4: 176131448-176131448 on Build GRCh38
rs10017197 Chr.4: 176131432-176131432 on Build GRCh38
rs377267907 Chr.4: 176131568-176131568 on Build GRCh38
rs112001577 Chr.4: 176131441-176131441 on Build GRCh38
rs751737915 Chr.4: 176131659-176131659 on Build GRCh38
rs953710881 Chr.4: 176131469-176131469 on Build GRCh38
rs756789949 Chr.4: 176131704-176131704 on Build GRCh38
rs1019185447 Chr.4: 176131394-176131394 on Build GRCh38
rs973046199 Chr.4: 176131742-176131742 on Build GRCh38
rs750614653 Chr.4: 176131545-176131545 on Build GRCh38
rs758087370 Chr.4: 176131386-176131386 on Build GRCh38
rs770757528 Chr.4: 176131600-176131600 on Build GRCh38
rs7699831 Chr.4: 176131774-176131774 on Build GRCh38
rs992375025 Chr.4: 176131475-176131475 on Build GRCh38
rs369270280 Chr.4: 176131768-176131768 on Build GRCh38
rs113765271 Chr.4: 176131370-176131370 on Build GRCh38
rs756705356 Chr.4: 176131666-176131666 on Build GRCh38
rs759382985 Chr.4: 176131527-176131527 on Build GRCh38
rs749951494 Chr.4: 176131533-176131533 on Build GRCh38
rs748695070 Chr.4: 176131774-176131775 on Build GRCh38
rs200906152 Chr.4: 176131532-176131532 on Build GRCh38
rs778731861 Chr.4: 176131707-176131707 on Build GRCh38

View Full Product Details