Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 184398837-184399341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF2
Entrez Gene ID
3660
Gene Name
interferon regulatory factor 2
Gene Aliases
IRF-2
UniGene
Hs.654566
Gene Chromosome Location
Chr.4: 184387722-184474572 on Build GRCh38
Transcripts
NM_002199.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4934179 c.571G>T p.E191* Chr.4: 184399038-184399038 on Build GRCh38
4426517 c.679G>A p.V227M Chr.4: 184398930-184398930 on Build GRCh38
6166917 c.622G>A p.E208K Chr.4: 184398987-184398987 on Build GRCh38
1328594 c.541_549delTCCCATCTG p.S181_L183delSHL Chr.4: 184399060-184399068 on Build GRCh38
3602435 c.676C>G p.P226A Chr.4: 184398933-184398933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539768956 Chr.4: 184399279-184399279 on Build GRCh38
rs766729867 Chr.4: 184398868-184398868 on Build GRCh38
rs201688327 Chr.4: 184399075-184399075 on Build GRCh38
rs201790611 Chr.4: 184398877-184398877 on Build GRCh38
rs566046035 Chr.4: 184399114-184399114 on Build GRCh38
rs374977236 Chr.4: 184399126-184399126 on Build GRCh38
rs759210054 Chr.4: 184399223-184399223 on Build GRCh38
rs150575776 Chr.4: 184399021-184399021 on Build GRCh38
rs781140860 Chr.4: 184399119-184399119 on Build GRCh38
rs73005101 Chr.4: 184398955-184398955 on Build GRCh38
rs200173412 Chr.4: 184398958-184398958 on Build GRCh38
rs113848358 Chr.4: 184398876-184398876 on Build GRCh38
rs746919993 Chr.4: 184398973-184398973 on Build GRCh38
rs905713681 Chr.4: 184399209-184399209 on Build GRCh38
rs1019243853 Chr.4: 184399321-184399321 on Build GRCh38
rs542706545 Chr.4: 184399242-184399242 on Build GRCh38
rs371796553 Chr.4: 184398892-184398892 on Build GRCh38
rs772772400 Chr.4: 184398903-184398903 on Build GRCh38
rs768030620 Chr.4: 184398919-184398919 on Build GRCh38
rs74960894 Chr.4: 184399102-184399102 on Build GRCh38
rs946555540 Chr.4: 184399139-184399139 on Build GRCh38
rs548043238 Chr.4: 184399095-184399095 on Build GRCh38
rs905081357 Chr.4: 184399173-184399173 on Build GRCh38
rs935085112 Chr.4: 184398874-184398874 on Build GRCh38
rs994984585 Chr.4: 184399232-184399232 on Build GRCh38
rs777730413 Chr.4: 184398985-184398985 on Build GRCh38
rs753118859 Chr.4: 184398930-184398930 on Build GRCh38
rs201848919 Chr.4: 184398971-184398971 on Build GRCh38
rs187952756 Chr.4: 184398939-184398939 on Build GRCh38
rs1029591703 Chr.4: 184399262-184399262 on Build GRCh38
rs200380327 Chr.4: 184398982-184398982 on Build GRCh38
rs560034190 Chr.4: 184398873-184398873 on Build GRCh38
rs753321388 Chr.4: 184399080-184399080 on Build GRCh38
rs987882074 Chr.4: 184399115-184399115 on Build GRCh38
rs757183928 Chr.4: 184398888-184398888 on Build GRCh38
rs112725705 Chr.4: 184398870-184398870 on Build GRCh38
rs983120470 Chr.4: 184398911-184398911 on Build GRCh38
rs758246792 Chr.4: 184398970-184398970 on Build GRCh38
rs748859954 Chr.4: 184398886-184398886 on Build GRCh38
rs574343604 Chr.4: 184399233-184399233 on Build GRCh38
rs550442580 Chr.4: 184399149-184399149 on Build GRCh38
rs765031173 Chr.4: 184399094-184399094 on Build GRCh38
rs545600776 Chr.4: 184398902-184398902 on Build GRCh38
rs1036429785 Chr.4: 184399221-184399221 on Build GRCh38
rs374816043 Chr.4: 184399239-184399239 on Build GRCh38
rs375169477 Chr.4: 184398908-184398908 on Build GRCh38
rs775243165 Chr.4: 184398895-184398895 on Build GRCh38
rs772168170 Chr.4: 184398894-184398894 on Build GRCh38
rs953614580 Chr.4: 184399265-184399265 on Build GRCh38
rs755491625 Chr.4: 184398871-184398871 on Build GRCh38
rs375952283 Chr.4: 184399117-184399117 on Build GRCh38
rs748308993 Chr.4: 184398900-184398900 on Build GRCh38
rs779705186 Chr.4: 184399104-184399104 on Build GRCh38
rs367602178 Chr.4: 184399258-184399258 on Build GRCh38
rs761360407 Chr.4: 184399081-184399081 on Build GRCh38
rs146966318 Chr.4: 184398931-184398931 on Build GRCh38
rs141757669 Chr.4: 184398997-184398997 on Build GRCh38
rs1004878505 Chr.4: 184399219-184399219 on Build GRCh38
rs563312207 Chr.4: 184399030-184399030 on Build GRCh38
rs758338927 Chr.4: 184399103-184399103 on Build GRCh38
rs576581600 Chr.4: 184398885-184398885 on Build GRCh38
rs771170897 Chr.4: 184399002-184399002 on Build GRCh38
rs182599605 Chr.4: 184399208-184399208 on Build GRCh38
rs370266524 Chr.4: 184399049-184399049 on Build GRCh38
rs199920647 Chr.4: 184398909-184398909 on Build GRCh38
rs1057023155 Chr.4: 184399193-184399193 on Build GRCh38
rs148252735 Chr.4: 184399174-184399174 on Build GRCh38
rs776132166 Chr.4: 184399045-184399045 on Build GRCh38
rs749389460 Chr.4: 184399123-184399123 on Build GRCh38
rs776452465 Chr.4: 184399222-184399222 on Build GRCh38
rs745593135 Chr.4: 184398893-184398893 on Build GRCh38
rs756982090 Chr.4: 184398952-184398952 on Build GRCh38
rs568951049 Chr.4: 184399164-184399164 on Build GRCh38
rs774504500 Chr.4: 184399019-184399019 on Build GRCh38
rs72701789 Chr.4: 184399317-184399317 on Build GRCh38
rs987109894 Chr.4: 184399022-184399022 on Build GRCh38
rs191230059 Chr.4: 184399131-184399131 on Build GRCh38

View Full Product Details