Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 1063962-1064459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC12A7
Entrez Gene ID
10723
Gene Name
solute carrier family 12 member 7
Gene Aliases
KCC4
UniGene
Hs.172613
Gene Chromosome Location
Chr.5: 1050374-1155887 on Build GRCh38
Transcripts
NM_006598.2 XM_005248231.3 XM_011513939.2 XM_011513940.2 XM_011513941.2 XM_017008958.1 XM_017008959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3918429 c.2585C>T p.P862L Chr.5: 1064105-1064105 on Build GRCh38
315216 c.2515G>T p.E839* Chr.5: 1064175-1064175 on Build GRCh38
3381072 c.2512C>A p.Q838K Chr.5: 1064178-1064178 on Build GRCh38
1225897 c.2560G>A p.D854N Chr.5: 1064130-1064130 on Build GRCh38
1432137 c.2562C>T p.D854D Chr.5: 1064128-1064128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534639626 Chr.5: 1064228-1064228 on Build GRCh38
rs371922462 Chr.5: 1064023-1064023 on Build GRCh38
rs982434029 Chr.5: 1064348-1064348 on Build GRCh38
rs746812366 Chr.5: 1064039-1064039 on Build GRCh38
rs747489820 Chr.5: 1064127-1064127 on Build GRCh38
rs749533085 Chr.5: 1064109-1064109 on Build GRCh38
rs761510641 Chr.5: 1064071-1064071 on Build GRCh38
rs1040136789 Chr.5: 1064355-1064355 on Build GRCh38
rs574433513 Chr.5: 1064094-1064094 on Build GRCh38
rs967060009 Chr.5: 1064075-1064075 on Build GRCh38
rs996451071 Chr.5: 1064437-1064437 on Build GRCh38
rs147157876 Chr.5: 1064137-1064137 on Build GRCh38
rs372764592 Chr.5: 1064256-1064256 on Build GRCh38
rs772643043 Chr.5: 1064060-1064060 on Build GRCh38
rs200262061 Chr.5: 1064196-1064196 on Build GRCh38
rs751647966 Chr.5: 1064226-1064226 on Build GRCh38
rs751598195 Chr.5: 1064148-1064148 on Build GRCh38
rs536477577 Chr.5: 1064028-1064028 on Build GRCh38
rs900847311 Chr.5: 1064313-1064313 on Build GRCh38
rs376157680 Chr.5: 1064271-1064271 on Build GRCh38
rs923685769 Chr.5: 1064352-1064352 on Build GRCh38
rs548379644 Chr.5: 1064191-1064191 on Build GRCh38
rs558593640 Chr.5: 1064054-1064054 on Build GRCh38
rs761534585 Chr.5: 1064266-1064266 on Build GRCh38
rs778169891 Chr.5: 1064047-1064047 on Build GRCh38
rs756576351 Chr.5: 1064299-1064299 on Build GRCh38
rs143444431 Chr.5: 1064131-1064131 on Build GRCh38
rs375637356 Chr.5: 1064167-1064167 on Build GRCh38
rs201925892 Chr.5: 1064192-1064192 on Build GRCh38
rs200952889 Chr.5: 1064076-1064076 on Build GRCh38
rs1043197627 Chr.5: 1064387-1064387 on Build GRCh38
rs575704893 Chr.5: 1064303-1064303 on Build GRCh38
rs200353015 Chr.5: 1064034-1064034 on Build GRCh38
rs755916299 Chr.5: 1064402-1064402 on Build GRCh38
rs143746308 Chr.5: 1064149-1064149 on Build GRCh38
rs369427526 Chr.5: 1064275-1064275 on Build GRCh38
rs769063291 Chr.5: 1064128-1064128 on Build GRCh38
rs772093101 Chr.5: 1064121-1064121 on Build GRCh38
rs776412060 Chr.5: 1064257-1064257 on Build GRCh38
rs199829222 Chr.5: 1064035-1064035 on Build GRCh38
rs201538075 Chr.5: 1064072-1064072 on Build GRCh38
rs537355176 Chr.5: 1064194-1064194 on Build GRCh38
rs781114978 Chr.5: 1064097-1064097 on Build GRCh38
rs755001086 Chr.5: 1064227-1064227 on Build GRCh38
rs141189884 Chr.5: 1064248-1064248 on Build GRCh38
rs188889163 Chr.5: 1064199-1064199 on Build GRCh38
rs146540406 Chr.5: 1064243-1064243 on Build GRCh38
rs749496217 Chr.5: 1064050-1064050 on Build GRCh38
rs369218517 Chr.5: 1064241-1064241 on Build GRCh38
rs1010737300 Chr.5: 1064029-1064029 on Build GRCh38
rs755811025 Chr.5: 1064156-1064156 on Build GRCh38
rs757205489 Chr.5: 1064035-1064036 on Build GRCh38
rs555423907 Chr.5: 1064416-1064416 on Build GRCh38
rs749017763 Chr.5: 1064233-1064233 on Build GRCh38
rs748566611 Chr.5: 1064173-1064173 on Build GRCh38
rs947553603 Chr.5: 1064384-1064384 on Build GRCh38
rs767486873 Chr.5: 1064151-1064151 on Build GRCh38
rs761054117 Chr.5: 1064063-1064063 on Build GRCh38
rs971416228 Chr.5: 1064321-1064321 on Build GRCh38
rs903736661 Chr.5: 1064361-1064361 on Build GRCh38
rs199981006 Chr.5: 1064122-1064122 on Build GRCh38
rs770967010 Chr.5: 1064053-1064053 on Build GRCh38
rs528322353 Chr.5: 1064338-1064338 on Build GRCh38
rs376700762 Chr.5: 1064068-1064068 on Build GRCh38
rs113356251 Chr.5: 1064032-1064032 on Build GRCh38
rs749400800 Chr.5: 1063993-1063993 on Build GRCh38
rs760067571 Chr.5: 1064006-1064006 on Build GRCh38
rs55792623 Chr.5: 1063985-1063985 on Build GRCh38
rs369998225 Chr.5: 1064069-1064069 on Build GRCh38
rs751739887 Chr.5: 1064274-1064274 on Build GRCh38
rs748443484 Chr.5: 1063984-1063984 on Build GRCh38
rs745978080 Chr.5: 1064116-1064116 on Build GRCh38
rs76539639 Chr.5: 1064152-1064152 on Build GRCh38
rs375034450 Chr.5: 1064224-1064224 on Build GRCh38
rs781490697 Chr.5: 1064172-1064172 on Build GRCh38
rs55717514 Chr.5: 1064388-1064388 on Build GRCh38
rs779222783 Chr.5: 1064163-1064163 on Build GRCh38
rs760966428 Chr.5: 1064018-1064018 on Build GRCh38
rs754858146 Chr.5: 1064096-1064096 on Build GRCh38
rs762554473 Chr.5: 1064146-1064146 on Build GRCh38
rs562235109 Chr.5: 1064179-1064179 on Build GRCh38
rs1008791294 Chr.5: 1064045-1064045 on Build GRCh38
rs139195330 Chr.5: 1064197-1064197 on Build GRCh38
rs903404713 Chr.5: 1064289-1064289 on Build GRCh38
rs754769892 Chr.5: 1064046-1064046 on Build GRCh38
rs928654414 Chr.5: 1064353-1064353 on Build GRCh38
rs192946976 Chr.5: 1064345-1064345 on Build GRCh38
rs149966134 Chr.5: 1064242-1064242 on Build GRCh38
rs573972499 Chr.5: 1064354-1064355 on Build GRCh38
rs1043719233 Chr.5: 1064283-1064283 on Build GRCh38
rs184953727 Chr.5: 1064164-1064164 on Build GRCh38
rs568203509 Chr.5: 1064026-1064026 on Build GRCh38
rs148964142 Chr.5: 1064036-1064036 on Build GRCh38
rs900875488 Chr.5: 1064418-1064418 on Build GRCh38
rs771000583 Chr.5: 1063995-1063995 on Build GRCh38
rs770624765 Chr.5: 1064186-1064186 on Build GRCh38
rs775831595 Chr.5: 1064011-1064011 on Build GRCh38
rs555900198 Chr.5: 1064177-1064177 on Build GRCh38
rs202124307 Chr.5: 1064229-1064229 on Build GRCh38
rs758516630 Chr.5: 1064166-1064166 on Build GRCh38
rs200015076 Chr.5: 1064185-1064185 on Build GRCh38
rs991592719 Chr.5: 1064364-1064364 on Build GRCh38
rs372136981 Chr.5: 1064059-1064059 on Build GRCh38
rs778277736 Chr.5: 1064301-1064301 on Build GRCh38
rs983947438 Chr.5: 1064339-1064339 on Build GRCh38
rs116788523 Chr.5: 1064112-1064112 on Build GRCh38
rs138300444 Chr.5: 1064093-1064093 on Build GRCh38
rs745587860 Chr.5: 1064100-1064100 on Build GRCh38
rs763752467 Chr.5: 1064019-1064019 on Build GRCh38
rs187890112 Chr.5: 1063999-1063999 on Build GRCh38
rs375569381 Chr.5: 1064232-1064232 on Build GRCh38
rs773510223 Chr.5: 1063990-1063990 on Build GRCh38
rs778911381 Chr.5: 1064047-1064047 on Build GRCh38
rs938648785 Chr.5: 1064355-1064355 on Build GRCh38
rs112178884 Chr.5: 1064326-1064326 on Build GRCh38
rs763494805 Chr.5: 1064247-1064247 on Build GRCh38
rs750713099 Chr.5: 1064200-1064200 on Build GRCh38
rs534347051 Chr.5: 1064024-1064024 on Build GRCh38
rs762012139 Chr.5: 1064136-1064136 on Build GRCh38
rs758611973 Chr.5: 1064205-1064205 on Build GRCh38
rs1005416958 Chr.5: 1064012-1064012 on Build GRCh38
rs748319707 Chr.5: 1064335-1064335 on Build GRCh38
rs540252418 Chr.5: 1064347-1064347 on Build GRCh38
rs757789370 Chr.5: 1064078-1064078 on Build GRCh38
rs137938985 Chr.5: 1064125-1064125 on Build GRCh38
rs753076014 Chr.5: 1064240-1064240 on Build GRCh38
rs775921517 Chr.5: 1064061-1064061 on Build GRCh38
rs779280701 Chr.5: 1064329-1064329 on Build GRCh38
rs771447442 Chr.5: 1064244-1064244 on Build GRCh38
rs545986241 Chr.5: 1064314-1064314 on Build GRCh38
rs377477211 Chr.5: 1064230-1064230 on Build GRCh38
rs934970097 Chr.5: 1064408-1064408 on Build GRCh38
rs915972620 Chr.5: 1064383-1064383 on Build GRCh38
rs753303615 Chr.5: 1064298-1064298 on Build GRCh38
rs144435869 Chr.5: 1064115-1064115 on Build GRCh38
rs931811450 Chr.5: 1064360-1064360 on Build GRCh38
rs150783997 Chr.5: 1064171-1064171 on Build GRCh38
rs975189608 Chr.5: 1064101-1064101 on Build GRCh38

View Full Product Details