Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 6633125-6633638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NSUN2
Entrez Gene ID
54888
Gene Name
NOP2/Sun RNA methyltransferase family member 2
Gene Aliases
MISU, MRT5, SAKI, TRM4
UniGene
Hs.481526
Gene Chromosome Location
Chr.5: 6599239-6633360 on Build GRCh38
Transcripts
NM_001193455.1 NM_017755.5 NR_037947.1 NM_001047.3 NM_001324322.1 NM_001324323.1 NR_136739.1

Gene Symbol
SRD5A1
Entrez Gene ID
6715
Gene Name
steroid 5 alpha-reductase 1
Gene Aliases
S5AR 1
UniGene
Hs.552
Gene Chromosome Location
Chr.5: 6633322-6669562 on Build GRCh38
Transcripts
NM_001193455.1 NM_017755.5 NR_037947.1 NM_001047.3 NM_001324322.1 NM_001324323.1 NR_136739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6416079 c.48G>A p.W16* Chr.5: 6633598-6633598 on Build GRCh38
7317087 c.26G>A p.R9Q Chr.5: 6633576-6633576 on Build GRCh38
6416078 c.22G>A p.A8T Chr.5: 6633598-6633598 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1000117715 Chr.5: 6633562-6633562 on Build GRCh38
rs1021065627 Chr.5: 6633449-6633450 on Build GRCh38
rs562817249 Chr.5: 6633200-6633201 on Build GRCh38
rs911757364 Chr.5: 6633393-6633393 on Build GRCh38
rs1028687957 Chr.5: 6633599-6633599 on Build GRCh38
rs895146530 Chr.5: 6633472-6633472 on Build GRCh38
rs773515911 Chr.5: 6633590-6633590 on Build GRCh38
rs553765471 Chr.5: 6633425-6633425 on Build GRCh38
rs886060730 Chr.5: 6633260-6633260 on Build GRCh38
rs775242539 Chr.5: 6633618-6633618 on Build GRCh38
rs1031127266 Chr.5: 6633584-6633584 on Build GRCh38
rs8192124 Chr.5: 6633195-6633195 on Build GRCh38
rs768308608 Chr.5: 6633561-6633561 on Build GRCh38
rs965234927 Chr.5: 6633510-6633510 on Build GRCh38
rs1055442322 Chr.5: 6633406-6633406 on Build GRCh38
rs890291814 Chr.5: 6633232-6633232 on Build GRCh38
rs1012308268 Chr.5: 6633480-6633480 on Build GRCh38
rs1002516510 Chr.5: 6633264-6633264 on Build GRCh38
rs940613768 Chr.5: 6633233-6633233 on Build GRCh38
rs898381211 Chr.5: 6633263-6633263 on Build GRCh38
rs759531023 Chr.5: 6633601-6633601 on Build GRCh38
rs1013686446 Chr.5: 6633302-6633302 on Build GRCh38
rs975399086 Chr.5: 6633328-6633328 on Build GRCh38
rs989129672 Chr.5: 6633149-6633149 on Build GRCh38
rs930542647 Chr.5: 6633440-6633440 on Build GRCh38
rs540541359 Chr.5: 6633415-6633415 on Build GRCh38
rs1021013304 Chr.5: 6633307-6633308 on Build GRCh38
rs920910653 Chr.5: 6633207-6633207 on Build GRCh38
rs542586533 Chr.5: 6633497-6633497 on Build GRCh38
rs1032551251 Chr.5: 6633476-6633476 on Build GRCh38
rs921592232 Chr.5: 6633352-6633352 on Build GRCh38
rs899054676 Chr.5: 6633253-6633253 on Build GRCh38
rs771322690 Chr.5: 6633592-6633592 on Build GRCh38
rs774981039 Chr.5: 6633593-6633593 on Build GRCh38
rs1025000469 Chr.5: 6633481-6633481 on Build GRCh38
rs1038192733 Chr.5: 6633381-6633381 on Build GRCh38
rs966835298 Chr.5: 6633158-6633158 on Build GRCh38
rs958288370 Chr.5: 6633322-6633322 on Build GRCh38
rs902631204 Chr.5: 6633467-6633467 on Build GRCh38
rs891462737 Chr.5: 6633410-6633410 on Build GRCh38
rs939869834 Chr.5: 6633374-6633374 on Build GRCh38
rs544403469 Chr.5: 6633591-6633591 on Build GRCh38
rs8192126 Chr.5: 6633392-6633392 on Build GRCh38
rs8192123 Chr.5: 6633172-6633172 on Build GRCh38
rs931761666 Chr.5: 6633360-6633360 on Build GRCh38
rs886949286 Chr.5: 6633226-6633227 on Build GRCh38
rs997295040 Chr.5: 6633469-6633483 on Build GRCh38
rs1050795695 Chr.5: 6633218-6633218 on Build GRCh38
rs546579970 Chr.5: 6633152-6633152 on Build GRCh38
rs531700189 Chr.5: 6633560-6633560 on Build GRCh38
rs970850944 Chr.5: 6633316-6633316 on Build GRCh38
rs995931064 Chr.5: 6633271-6633271 on Build GRCh38
rs908276419 Chr.5: 6633359-6633359 on Build GRCh38
rs8192127 Chr.5: 6633401-6633401 on Build GRCh38
rs8192122 Chr.5: 6633168-6633168 on Build GRCh38
rs769376823 Chr.5: 6633434-6633434 on Build GRCh38
rs562686165 Chr.5: 6633521-6633521 on Build GRCh38
rs533040317 Chr.5: 6633150-6633150 on Build GRCh38
rs778200969 Chr.5: 6633284-6633284 on Build GRCh38
rs1036676322 Chr.5: 6633238-6633238 on Build GRCh38
rs8192125 Chr.5: 6633334-6633334 on Build GRCh38
rs1026948051 Chr.5: 6633276-6633276 on Build GRCh38
rs375270250 Chr.5: 6633437-6633437 on Build GRCh38
rs933796392 Chr.5: 6633214-6633214 on Build GRCh38
rs573730972 Chr.5: 6633494-6633494 on Build GRCh38
rs745515015 Chr.5: 6633418-6633418 on Build GRCh38
rs746596830 Chr.5: 6633559-6633559 on Build GRCh38
rs368667028 Chr.5: 6633297-6633297 on Build GRCh38
rs1048908651 Chr.5: 6633454-6633454 on Build GRCh38
rs569790049 Chr.5: 6633321-6633321 on Build GRCh38
rs975135660 Chr.5: 6633201-6633201 on Build GRCh38
rs933617257 Chr.5: 6633335-6633335 on Build GRCh38
rs1025136831 Chr.5: 6633312-6633312 on Build GRCh38
rs147269098 Chr.5: 6633319-6633319 on Build GRCh38

View Full Product Details