Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 6737419-6737915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAPD7
Entrez Gene ID
11044
Gene Name
PAP associated domain containing 7
Gene Aliases
LAK-1, LAK1, POLK, POLS, TRF4, TRF4-1, TRF41, TUTASE5
UniGene
Hs.481542
Gene Chromosome Location
Chr.5: 6713952-6757048 on Build GRCh38
Transcripts
NM_001171805.1 NM_006999.4 XM_005248234.3 XM_011513945.1 XM_017008988.1 XM_017008989.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4909874 c.80C>G p.P27R Chr.5: 6737623-6737623 on Build GRCh38
2149209 c.60_61delTG p.V21fs*9 Chr.5: 6737603-6737604 on Build GRCh38
6737402 c.55G>T p.E19* Chr.5: 6737598-6737598 on Build GRCh38
1620478 c.67A>G p.K23E Chr.5: 6737610-6737610 on Build GRCh38
7423231 c.35G>A p.R12K Chr.5: 6737578-6737578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772959682 Chr.5: 6737576-6737576 on Build GRCh38
rs878957608 Chr.5: 6737467-6737467 on Build GRCh38
rs2279656 Chr.5: 6737463-6737463 on Build GRCh38
rs983601862 Chr.5: 6737809-6737809 on Build GRCh38
rs116527147 Chr.5: 6737675-6737675 on Build GRCh38
rs956693910 Chr.5: 6737549-6737549 on Build GRCh38
rs768687895 Chr.5: 6737501-6737501 on Build GRCh38
rs765977463 Chr.5: 6737480-6737480 on Build GRCh38
rs145475863 Chr.5: 6737627-6737627 on Build GRCh38
rs751030736 Chr.5: 6737666-6737666 on Build GRCh38
rs949510163 Chr.5: 6737735-6737735 on Build GRCh38
rs902634762 Chr.5: 6737891-6737891 on Build GRCh38
rs954616223 Chr.5: 6737648-6737648 on Build GRCh38
rs756305305 Chr.5: 6737668-6737668 on Build GRCh38
rs1017786458 Chr.5: 6737822-6737822 on Build GRCh38
rs577717766 Chr.5: 6737866-6737866 on Build GRCh38
rs1042435760 Chr.5: 6737890-6737890 on Build GRCh38
rs973076191 Chr.5: 6737840-6737840 on Build GRCh38
rs1032543438 Chr.5: 6737763-6737763 on Build GRCh38
rs966174882 Chr.5: 6737826-6737826 on Build GRCh38
rs867711160 Chr.5: 6737782-6737782 on Build GRCh38
rs369940829 Chr.5: 6737662-6737662 on Build GRCh38
rs571317210 Chr.5: 6737625-6737625 on Build GRCh38
rs775977163 Chr.5: 6737656-6737657 on Build GRCh38
rs945941469 Chr.5: 6737881-6737881 on Build GRCh38
rs760201324 Chr.5: 6737611-6737611 on Build GRCh38
rs963023951 Chr.5: 6737838-6737838 on Build GRCh38
rs745741016 Chr.5: 6737677-6737677 on Build GRCh38
rs1010085675 Chr.5: 6737569-6737569 on Build GRCh38
rs964830939 Chr.5: 6737618-6737618 on Build GRCh38
rs555247924 Chr.5: 6737778-6737778 on Build GRCh38
rs375314494 Chr.5: 6737624-6737624 on Build GRCh38
rs201903547 Chr.5: 6737591-6737591 on Build GRCh38
rs566908256 Chr.5: 6737706-6737706 on Build GRCh38
rs371018590 Chr.5: 6737886-6737886 on Build GRCh38
rs778563360 Chr.5: 6737634-6737637 on Build GRCh38
rs1042485191 Chr.5: 6737736-6737736 on Build GRCh38
rs758893026 Chr.5: 6737626-6737626 on Build GRCh38
rs369641293 Chr.5: 6737744-6737744 on Build GRCh38
rs1030923768 Chr.5: 6737801-6737801 on Build GRCh38
rs535671695 Chr.5: 6737717-6737717 on Build GRCh38
rs775083489 Chr.5: 6737600-6737600 on Build GRCh38
rs1021378258 Chr.5: 6737460-6737460 on Build GRCh38
rs748261070 Chr.5: 6737538-6737538 on Build GRCh38
rs781084776 Chr.5: 6737665-6737665 on Build GRCh38
rs937164179 Chr.5: 6737692-6737692 on Build GRCh38
rs1056288275 Chr.5: 6737757-6737757 on Build GRCh38
rs1000845322 Chr.5: 6737758-6737758 on Build GRCh38
rs201401259 Chr.5: 6737580-6737580 on Build GRCh38
rs766876723 Chr.5: 6737598-6737598 on Build GRCh38
rs182397859 Chr.5: 6737671-6737671 on Build GRCh38
rs781338394 Chr.5: 6737505-6737505 on Build GRCh38
rs538156050 Chr.5: 6737814-6737814 on Build GRCh38
rs774207262 Chr.5: 6737676-6737676 on Build GRCh38
rs372514240 Chr.5: 6737597-6737597 on Build GRCh38
rs755883791 Chr.5: 6737694-6737694 on Build GRCh38
rs954933988 Chr.5: 6737805-6737805 on Build GRCh38
rs28363345 Chr.5: 6737855-6737855 on Build GRCh38
rs2279657 Chr.5: 6737458-6737458 on Build GRCh38
rs11539826 Chr.5: 6737551-6737551 on Build GRCh38
rs1001065733 Chr.5: 6737476-6737476 on Build GRCh38
rs1022350819 Chr.5: 6737603-6737603 on Build GRCh38
rs755522952 Chr.5: 6737661-6737661 on Build GRCh38
rs905001557 Chr.5: 6737743-6737743 on Build GRCh38
rs374832320 Chr.5: 6737674-6737674 on Build GRCh38
rs558045770 Chr.5: 6737852-6737852 on Build GRCh38
rs1032597259 Chr.5: 6737599-6737599 on Build GRCh38
rs28363344 Chr.5: 6737783-6737783 on Build GRCh38
rs992642713 Chr.5: 6737867-6737867 on Build GRCh38
rs749130040 Chr.5: 6737577-6737577 on Build GRCh38
rs569570059 Chr.5: 6737483-6737483 on Build GRCh38
rs917063830 Chr.5: 6737869-6737869 on Build GRCh38
rs896531403 Chr.5: 6737710-6737710 on Build GRCh38

View Full Product Details