Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 35955506-35956050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UGT3A1
Entrez Gene ID
133688
Gene Name
UDP glycosyltransferase family 3 member A1
Gene Aliases
-
UniGene
Hs.254699
Gene Chromosome Location
Chr.5: 35953089-36001055 on Build GRCh38
Transcripts
NM_152404.3 XM_005248243.4 XM_011513957.2 XM_011513958.2 XM_011513959.2 XM_011513961.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5650584 c.1232G>A p.R411Q Chr.5: 35955708-35955708 on Build GRCh38
6470305 c.1104T>C p.T368T Chr.5: 35955836-35955836 on Build GRCh38
6439558 c.1272G>A p.M424I Chr.5: 35955668-35955668 on Build GRCh38
4739120 c.1199G>A p.R400Q Chr.5: 35955741-35955741 on Build GRCh38
298944 c.1090C>T p.R364C Chr.5: 35955850-35955850 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761985145 Chr.5: 35955746-35955746 on Build GRCh38
rs772440877 Chr.5: 35955817-35955817 on Build GRCh38
rs375000458 Chr.5: 35955708-35955708 on Build GRCh38
rs778521267 Chr.5: 35955690-35955690 on Build GRCh38
rs753110453 Chr.5: 35955947-35955947 on Build GRCh38
rs371955454 Chr.5: 35955749-35955749 on Build GRCh38
rs754349872 Chr.5: 35955847-35955847 on Build GRCh38
rs996394938 Chr.5: 35955972-35955972 on Build GRCh38
rs61734803 Chr.5: 35955742-35955742 on Build GRCh38
rs148008807 Chr.5: 35955727-35955727 on Build GRCh38
rs1003741277 Chr.5: 35955919-35955919 on Build GRCh38
rs537122130 Chr.5: 35955870-35955870 on Build GRCh38
rs143085376 Chr.5: 35955866-35955866 on Build GRCh38
rs375449121 Chr.5: 35955634-35955634 on Build GRCh38
rs763024246 Chr.5: 35955651-35955651 on Build GRCh38
rs376306813 Chr.5: 35955969-35955969 on Build GRCh38
rs906189804 Chr.5: 35955899-35955899 on Build GRCh38
rs749212852 Chr.5: 35955884-35955884 on Build GRCh38
rs748247008 Chr.5: 35955902-35955902 on Build GRCh38
rs191737460 Chr.5: 35955748-35955748 on Build GRCh38
rs751793802 Chr.5: 35955664-35955664 on Build GRCh38
rs762050381 Chr.5: 35955806-35955806 on Build GRCh38
rs745663203 Chr.5: 35955791-35955791 on Build GRCh38
rs776641547 Chr.5: 35955774-35955774 on Build GRCh38
rs75976806 Chr.5: 35955901-35955901 on Build GRCh38
rs768623 Chr.5: 35955794-35955794 on Build GRCh38
rs768621 Chr.5: 35955924-35955924 on Build GRCh38
rs947951706 Chr.5: 35955881-35955881 on Build GRCh38
rs201810712 Chr.5: 35955802-35955802 on Build GRCh38
rs769736050 Chr.5: 35955729-35955729 on Build GRCh38
rs900873143 Chr.5: 35955540-35955540 on Build GRCh38
rs756925782 Chr.5: 35955776-35955776 on Build GRCh38
rs201756189 Chr.5: 35955647-35955647 on Build GRCh38
rs763655316 Chr.5: 35955752-35955763 on Build GRCh38
rs113340829 Chr.5: 35955639-35955639 on Build GRCh38
rs780906291 Chr.5: 35955787-35955787 on Build GRCh38
rs375679556 Chr.5: 35955801-35955801 on Build GRCh38
rs771779351 Chr.5: 35955582-35955582 on Build GRCh38
rs753469068 Chr.5: 35955841-35955842 on Build GRCh38
rs376132419 Chr.5: 35955559-35955559 on Build GRCh38
rs754376736 Chr.5: 35956008-35956008 on Build GRCh38
rs368671400 Chr.5: 35955846-35955847 on Build GRCh38
rs369769596 Chr.5: 35955831-35955831 on Build GRCh38
rs866339794 Chr.5: 35955781-35955781 on Build GRCh38
rs61734786 Chr.5: 35955773-35955773 on Build GRCh38
rs138607277 Chr.5: 35956005-35956005 on Build GRCh38
rs780814300 Chr.5: 35955859-35955859 on Build GRCh38
rs554821154 Chr.5: 35955850-35955850 on Build GRCh38
rs773557334 Chr.5: 35955912-35955912 on Build GRCh38
rs756563677 Chr.5: 35955984-35955984 on Build GRCh38
rs778472570 Chr.5: 35955539-35955539 on Build GRCh38
rs756835981 Chr.5: 35955853-35955853 on Build GRCh38
rs756723625 Chr.5: 35955689-35955689 on Build GRCh38
rs761117203 Chr.5: 35955830-35955830 on Build GRCh38
rs768622 Chr.5: 35955580-35955580 on Build GRCh38
rs777478979 Chr.5: 35955972-35955973 on Build GRCh38
rs772345737 Chr.5: 35955908-35955908 on Build GRCh38
rs780025698 Chr.5: 35955795-35955795 on Build GRCh38
rs776526035 Chr.5: 35955625-35955625 on Build GRCh38
rs141143848 Chr.5: 35955709-35955709 on Build GRCh38
rs990177027 Chr.5: 35955983-35955983 on Build GRCh38
rs183549396 Chr.5: 35955793-35955793 on Build GRCh38
rs968961117 Chr.5: 35955966-35955966 on Build GRCh38
rs375876119 Chr.5: 35955923-35955923 on Build GRCh38
rs375763628 Chr.5: 35955598-35955598 on Build GRCh38
rs777479381 Chr.5: 35955710-35955710 on Build GRCh38
rs749903674 Chr.5: 35955903-35955904 on Build GRCh38
rs745552698 Chr.5: 35955574-35955574 on Build GRCh38
rs772826350 Chr.5: 35955592-35955592 on Build GRCh38
rs762872593 Chr.5: 35955796-35955796 on Build GRCh38
rs759805132 Chr.5: 35955894-35955894 on Build GRCh38
rs900937540 Chr.5: 35955841-35955841 on Build GRCh38
rs764377459 Chr.5: 35955652-35955652 on Build GRCh38
rs746827095 Chr.5: 35955604-35955604 on Build GRCh38
rs970819547 Chr.5: 35955950-35955950 on Build GRCh38
rs773645359 Chr.5: 35955818-35955818 on Build GRCh38
rs186401248 Chr.5: 35956010-35956010 on Build GRCh38
rs998049236 Chr.5: 35955558-35955558 on Build GRCh38
rs757914558 Chr.5: 35955764-35955764 on Build GRCh38
rs368644392 Chr.5: 35955630-35955630 on Build GRCh38
rs776440342 Chr.5: 35955723-35955723 on Build GRCh38
rs147780586 Chr.5: 35955688-35955688 on Build GRCh38
rs149057248 Chr.5: 35955849-35955849 on Build GRCh38
rs766910396 Chr.5: 35955835-35955835 on Build GRCh38
rs754372841 Chr.5: 35955763-35955763 on Build GRCh38
rs371222114 Chr.5: 35955661-35955661 on Build GRCh38
rs770726663 Chr.5: 35955720-35955720 on Build GRCh38
rs778462352 Chr.5: 35955876-35955879 on Build GRCh38
rs143483561 Chr.5: 35955717-35955717 on Build GRCh38
rs199844466 Chr.5: 35955668-35955668 on Build GRCh38
rs768047152 Chr.5: 35955669-35955669 on Build GRCh38
rs974758361 Chr.5: 35955780-35955780 on Build GRCh38
rs201292612 Chr.5: 35955618-35955618 on Build GRCh38
rs954730959 Chr.5: 35956002-35956002 on Build GRCh38
rs373428697 Chr.5: 35955951-35955951 on Build GRCh38
rs766998224 Chr.5: 35955757-35955757 on Build GRCh38
rs775477816 Chr.5: 35955740-35955740 on Build GRCh38

View Full Product Details