Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 36257221-36257719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP3L
Entrez Gene ID
202151
Gene Name
RAN binding protein 3 like
Gene Aliases
-
UniGene
Hs.199777
Gene Chromosome Location
Chr.5: 36246919-36301936 on Build GRCh38
Transcripts
NM_001161429.2 NM_001323273.1 NM_001323274.1 NM_001323275.1 NM_001323276.1 NM_001323277.1 NM_001323278.1 NM_001323279.1 NM_001323280.1 NM_145000.4 XM_006714452.3 XM_006714453.3 XM_006714455.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7402398 c.706G>C p.D236H Chr.5: 36257520-36257520 on Build GRCh38
7386832 c.676C>A p.Q226K Chr.5: 36257550-36257550 on Build GRCh38
4510092 c.751C>T p.P251S Chr.5: 36257475-36257475 on Build GRCh38
737921 c.772+1G>A p.? Chr.5: 36257453-36257453 on Build GRCh38
5778402 c.744G>A p.P248P Chr.5: 36257482-36257482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905698534 Chr.5: 36257678-36257678 on Build GRCh38
rs759509502 Chr.5: 36257462-36257462 on Build GRCh38
rs776523109 Chr.5: 36257539-36257539 on Build GRCh38
rs374850317 Chr.5: 36257564-36257564 on Build GRCh38
rs375269424 Chr.5: 36257604-36257604 on Build GRCh38
rs138358489 Chr.5: 36257482-36257482 on Build GRCh38
rs182979617 Chr.5: 36257413-36257413 on Build GRCh38
rs762858150 Chr.5: 36257554-36257554 on Build GRCh38
rs890511563 Chr.5: 36257345-36257345 on Build GRCh38
rs150395742 Chr.5: 36257369-36257369 on Build GRCh38
rs879721924 Chr.5: 36257288-36257288 on Build GRCh38
rs371320991 Chr.5: 36257418-36257418 on Build GRCh38
rs539547009 Chr.5: 36257261-36257261 on Build GRCh38
rs371570640 Chr.5: 36257467-36257467 on Build GRCh38
rs556987108 Chr.5: 36257355-36257355 on Build GRCh38
rs559459334 Chr.5: 36257675-36257676 on Build GRCh38
rs996113927 Chr.5: 36257342-36257342 on Build GRCh38
rs532073313 Chr.5: 36257337-36257337 on Build GRCh38
rs979555296 Chr.5: 36257499-36257499 on Build GRCh38
rs76783647 Chr.5: 36257677-36257677 on Build GRCh38
rs772884729 Chr.5: 36257444-36257444 on Build GRCh38
rs969265816 Chr.5: 36257615-36257615 on Build GRCh38
rs751770258 Chr.5: 36257483-36257483 on Build GRCh38
rs749796789 Chr.5: 36257520-36257520 on Build GRCh38
rs772913076 Chr.5: 36257532-36257532 on Build GRCh38
rs747715509 Chr.5: 36257420-36257420 on Build GRCh38
rs757485595 Chr.5: 36257490-36257490 on Build GRCh38
rs576835254 Chr.5: 36257361-36257361 on Build GRCh38
rs369254399 Chr.5: 36257370-36257374 on Build GRCh38
rs200661750 Chr.5: 36257457-36257457 on Build GRCh38
rs112018936 Chr.5: 36257430-36257430 on Build GRCh38
rs995749996 Chr.5: 36257674-36257674 on Build GRCh38
rs771527330 Chr.5: 36257441-36257441 on Build GRCh38
rs770741477 Chr.5: 36257534-36257534 on Build GRCh38
rs114465031 Chr.5: 36257565-36257565 on Build GRCh38
rs150084810 Chr.5: 36257480-36257480 on Build GRCh38
rs764177185 Chr.5: 36257562-36257562 on Build GRCh38
rs932247292 Chr.5: 36257501-36257501 on Build GRCh38
rs898341432 Chr.5: 36257351-36257351 on Build GRCh38
rs980422789 Chr.5: 36257422-36257422 on Build GRCh38
rs904818625 Chr.5: 36257320-36257320 on Build GRCh38
rs746314654 Chr.5: 36257409-36257409 on Build GRCh38
rs960662625 Chr.5: 36257676-36257676 on Build GRCh38
rs754354127 Chr.5: 36257607-36257607 on Build GRCh38
rs202231727 Chr.5: 36257514-36257514 on Build GRCh38
rs10214173 Chr.5: 36257302-36257302 on Build GRCh38
rs780587007 Chr.5: 36257519-36257519 on Build GRCh38
rs760468586 Chr.5: 36257456-36257456 on Build GRCh38
rs750692258 Chr.5: 36257597-36257597 on Build GRCh38
rs564212414 Chr.5: 36257599-36257599 on Build GRCh38
rs752630263 Chr.5: 36257476-36257476 on Build GRCh38
rs188050813 Chr.5: 36257461-36257461 on Build GRCh38
rs1028977944 Chr.5: 36257343-36257343 on Build GRCh38
rs1036846251 Chr.5: 36257548-36257548 on Build GRCh38
rs552943299 Chr.5: 36257373-36257373 on Build GRCh38
rs781467794 Chr.5: 36257508-36257508 on Build GRCh38
rs143481517 Chr.5: 36257542-36257542 on Build GRCh38
rs1002140730 Chr.5: 36257645-36257645 on Build GRCh38
rs542050330 Chr.5: 36257427-36257427 on Build GRCh38
rs763025815 Chr.5: 36257596-36257596 on Build GRCh38
rs771603279 Chr.5: 36257529-36257529 on Build GRCh38
rs962192127 Chr.5: 36257389-36257391 on Build GRCh38
rs773420375 Chr.5: 36257452-36257457 on Build GRCh38
rs149245234 Chr.5: 36257515-36257515 on Build GRCh38

View Full Product Details