Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 37379147-37379641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR70
Entrez Gene ID
55100
Gene Name
WD repeat domain 70
Gene Aliases
-
UniGene
Hs.213690
Gene Chromosome Location
Chr.5: 37379280-37752672 on Build GRCh38
Transcripts
NM_018034.2 XM_005248318.3 XM_017009604.1 XM_017009605.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3776682 c.75G>A p.M25I Chr.5: 37379538-37379538 on Build GRCh38
6358341 c.54C>T p.D18D Chr.5: 37379517-37379517 on Build GRCh38
4767008 c.73A>G p.M25V Chr.5: 37379536-37379536 on Build GRCh38
1437391 c.6G>A p.E2E Chr.5: 37379373-37379373 on Build GRCh38
7091686 c.69C>T p.V23V Chr.5: 37379532-37379532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763154718 Chr.5: 37379443-37379443 on Build GRCh38
rs552178739 Chr.5: 37379438-37379438 on Build GRCh38
rs541467872 Chr.5: 37379481-37379481 on Build GRCh38
rs1017225484 Chr.5: 37379364-37379364 on Build GRCh38
rs760410149 Chr.5: 37379377-37379377 on Build GRCh38
rs780128060 Chr.5: 37379597-37379597 on Build GRCh38
rs751909772 Chr.5: 37379402-37379402 on Build GRCh38
rs761981506 Chr.5: 37379519-37379519 on Build GRCh38
rs1042444423 Chr.5: 37379536-37379536 on Build GRCh38
rs374281450 Chr.5: 37379474-37379474 on Build GRCh38
rs757730785 Chr.5: 37379344-37379344 on Build GRCh38
rs745590955 Chr.5: 37379590-37379590 on Build GRCh38
rs747777440 Chr.5: 37379428-37379428 on Build GRCh38
rs747865663 Chr.5: 37379373-37379373 on Build GRCh38
rs758829953 Chr.5: 37379319-37379319 on Build GRCh38
rs781578603 Chr.5: 37379347-37379347 on Build GRCh38
rs200819438 Chr.5: 37379328-37379328 on Build GRCh38
rs199839331 Chr.5: 37379283-37379283 on Build GRCh38
rs918633950 Chr.5: 37379178-37379178 on Build GRCh38
rs553601530 Chr.5: 37379245-37379245 on Build GRCh38
rs770687137 Chr.5: 37379436-37379436 on Build GRCh38
rs757918863 Chr.5: 37379569-37379569 on Build GRCh38
rs374431593 Chr.5: 37379605-37379605 on Build GRCh38
rs776444993 Chr.5: 37379383-37379383 on Build GRCh38
rs377497841 Chr.5: 37379441-37379441 on Build GRCh38
rs754484291 Chr.5: 37379417-37379417 on Build GRCh38
rs748768629 Chr.5: 37379353-37379353 on Build GRCh38
rs775661431 Chr.5: 37379442-37379442 on Build GRCh38
rs773110090 Chr.5: 37379376-37379376 on Build GRCh38
rs942919319 Chr.5: 37379257-37379257 on Build GRCh38
rs757564657 Chr.5: 37379408-37379408 on Build GRCh38
rs767343467 Chr.5: 37379398-37379398 on Build GRCh38
rs367957947 Chr.5: 37379363-37379363 on Build GRCh38
rs754359498 Chr.5: 37379567-37379567 on Build GRCh38
rs764430024 Chr.5: 37379400-37379400 on Build GRCh38
rs777323862 Chr.5: 37379433-37379433 on Build GRCh38
rs755983886 Chr.5: 37379592-37379592 on Build GRCh38
rs541368729 Chr.5: 37379396-37379396 on Build GRCh38
rs746703449 Chr.5: 37379487-37379487 on Build GRCh38
rs201435039 Chr.5: 37379397-37379397 on Build GRCh38
rs565635169 Chr.5: 37379570-37379570 on Build GRCh38
rs543864820 Chr.5: 37379317-37379317 on Build GRCh38
rs959311493 Chr.5: 37379337-37379337 on Build GRCh38
rs893487736 Chr.5: 37379272-37379272 on Build GRCh38
rs574131159 Chr.5: 37379378-37379378 on Build GRCh38
rs747447493 Chr.5: 37379334-37379334 on Build GRCh38
rs549822170 Chr.5: 37379503-37379503 on Build GRCh38
rs539476871 Chr.5: 37379581-37379581 on Build GRCh38
rs539437986 Chr.5: 37379280-37379280 on Build GRCh38
rs752165618 Chr.5: 37379477-37379477 on Build GRCh38
rs753841669 Chr.5: 37379335-37379335 on Build GRCh38
rs377035284 Chr.5: 37379612-37379612 on Build GRCh38
rs369181286 Chr.5: 37379617-37379617 on Build GRCh38
rs1055090377 Chr.5: 37379271-37379271 on Build GRCh38
rs537151813 Chr.5: 37379616-37379616 on Build GRCh38
rs773982335 Chr.5: 37379362-37379362 on Build GRCh38
rs767900939 Chr.5: 37379520-37379520 on Build GRCh38
rs759673295 Chr.5: 37379392-37379392 on Build GRCh38
rs761062481 Chr.5: 37379528-37379528 on Build GRCh38
rs890067116 Chr.5: 37379215-37379215 on Build GRCh38
rs771823178 Chr.5: 37379375-37379375 on Build GRCh38
rs527327883 Chr.5: 37379434-37379434 on Build GRCh38
rs572116034 Chr.5: 37379273-37379273 on Build GRCh38
rs142856067 Chr.5: 37379496-37379496 on Build GRCh38
rs762225720 Chr.5: 37379459-37379459 on Build GRCh38
rs377642196 Chr.5: 37379483-37379483 on Build GRCh38
rs751198244 Chr.5: 37379577-37379577 on Build GRCh38
rs778438273 Chr.5: 37379419-37379419 on Build GRCh38
rs769814261 Chr.5: 37379505-37379505 on Build GRCh38
rs750898834 Chr.5: 37379467-37379467 on Build GRCh38
rs369130173 Chr.5: 37379451-37379451 on Build GRCh38
rs576106388 Chr.5: 37379299-37379299 on Build GRCh38
rs535404319 Chr.5: 37379199-37379199 on Build GRCh38
rs374806437 Chr.5: 37379357-37379357 on Build GRCh38
rs934182938 Chr.5: 37379180-37379180 on Build GRCh38
rs188085649 Chr.5: 37379171-37379171 on Build GRCh38
rs181030055 Chr.5: 37379465-37379465 on Build GRCh38
rs771665517 Chr.5: 37379429-37379429 on Build GRCh38
rs756666961 Chr.5: 37379469-37379469 on Build GRCh38
rs371538175 Chr.5: 37379583-37379583 on Build GRCh38
rs535598828 Chr.5: 37379513-37379513 on Build GRCh38
rs773633844 Chr.5: 37379350-37379351 on Build GRCh38
rs763516637 Chr.5: 37379561-37379561 on Build GRCh38
rs558310846 Chr.5: 37379314-37379314 on Build GRCh38
rs373406711 Chr.5: 37379414-37379414 on Build GRCh38
rs751745164 Chr.5: 37379455-37379455 on Build GRCh38
rs781684261 Chr.5: 37379413-37379413 on Build GRCh38
rs151067156 Chr.5: 37379508-37379508 on Build GRCh38
rs141035069 Chr.5: 37379550-37379550 on Build GRCh38
rs568209919 Chr.5: 37379188-37379188 on Build GRCh38
rs777491668 Chr.5: 37379212-37379212 on Build GRCh38

View Full Product Details