Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 38945581-38946115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RICTOR
Entrez Gene ID
253260
Gene Name
RPTOR independent companion of MTOR complex 2
Gene Aliases
AVO3, PIA, hAVO3
UniGene
Hs.407926
Gene Chromosome Location
Chr.5: 38937920-39074421 on Build GRCh38
Transcripts
NM_001285439.1 NM_001285440.1 NM_152756.4 XM_006714463.3 XM_011514005.2 XM_011514006.2 XM_017009311.1 XM_017009312.1 XM_017009313.1 XM_017009314.1 XM_017009315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6984398 c.4591A>G p.T1531A Chr.5: 38945605-38945605 on Build GRCh38
3855188 c.4472A>C p.E1491A Chr.5: 38945652-38945652 on Build GRCh38
2689232 c.4494A>C p.S1498S Chr.5: 38945630-38945630 on Build GRCh38
6943138 c.4441C>T p.H1481Y Chr.5: 38945683-38945683 on Build GRCh38
6949185 c.4604G>A p.G1535E Chr.5: 38945592-38945592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116364842 Chr.5: 38946018-38946018 on Build GRCh38
rs888919554 Chr.5: 38946071-38946071 on Build GRCh38
rs980585802 Chr.5: 38945830-38945830 on Build GRCh38
rs770102440 Chr.5: 38945664-38945664 on Build GRCh38
rs183359753 Chr.5: 38945888-38945888 on Build GRCh38
rs1034912282 Chr.5: 38945835-38945835 on Build GRCh38
rs933385376 Chr.5: 38945973-38945973 on Build GRCh38
rs771188617 Chr.5: 38945647-38945647 on Build GRCh38
rs1042620150 Chr.5: 38945648-38945648 on Build GRCh38
rs188644913 Chr.5: 38945995-38945995 on Build GRCh38
rs62352642 Chr.5: 38945817-38945817 on Build GRCh38
rs1003197029 Chr.5: 38945754-38945754 on Build GRCh38
rs777852200 Chr.5: 38945630-38945630 on Build GRCh38
rs199683262 Chr.5: 38945625-38945625 on Build GRCh38
rs936505475 Chr.5: 38946072-38946072 on Build GRCh38
rs765899771 Chr.5: 38945758-38945758 on Build GRCh38
rs567981982 Chr.5: 38946084-38946084 on Build GRCh38
rs373254827 Chr.5: 38945705-38945705 on Build GRCh38
rs570451799 Chr.5: 38945948-38945948 on Build GRCh38
rs747033304 Chr.5: 38945638-38945638 on Build GRCh38
rs551947585 Chr.5: 38945942-38945942 on Build GRCh38
rs757332185 Chr.5: 38945790-38945790 on Build GRCh38
rs369190681 Chr.5: 38945756-38945756 on Build GRCh38
rs529259343 Chr.5: 38945781-38945781 on Build GRCh38
rs763579152 Chr.5: 38945716-38945716 on Build GRCh38
rs909472129 Chr.5: 38945929-38945929 on Build GRCh38
rs376867631 Chr.5: 38945697-38945697 on Build GRCh38
rs1032272899 Chr.5: 38945655-38945655 on Build GRCh38
rs955260730 Chr.5: 38945974-38945974 on Build GRCh38
rs748125650 Chr.5: 38945611-38945611 on Build GRCh38
rs111564062 Chr.5: 38945824-38945824 on Build GRCh38
rs79020728 Chr.5: 38945654-38945654 on Build GRCh38
rs764640747 Chr.5: 38945727-38945727 on Build GRCh38
rs753998963 Chr.5: 38945951-38945951 on Build GRCh38
rs759787504 Chr.5: 38945659-38945659 on Build GRCh38
rs1053663242 Chr.5: 38946088-38946088 on Build GRCh38
rs987491254 Chr.5: 38945991-38945991 on Build GRCh38
rs958929753 Chr.5: 38945903-38945903 on Build GRCh38
rs149029231 Chr.5: 38945810-38945810 on Build GRCh38
rs527496176 Chr.5: 38945916-38945916 on Build GRCh38
rs749962017 Chr.5: 38945731-38945731 on Build GRCh38
rs541959049 Chr.5: 38945658-38945658 on Build GRCh38

View Full Product Details