Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 50760189-50760688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARP8
Entrez Gene ID
79668
Gene Name
poly(ADP-ribose) polymerase family member 8
Gene Aliases
ARTD16, pART16
UniGene
Hs.369581
Gene Chromosome Location
Chr.5: 50665899-50846522 on Build GRCh38
Transcripts
NM_001178055.1 NM_001178056.1 NM_024615.3 XM_005248596.4 XM_011543631.2 XM_011543632.2 XM_011543633.2 XM_011543634.2 XM_011543635.2 XM_011543636.2 XM_011543640.2 XM_011543641.2 XM_011543642.2 XM_011543643.2 XM_017009850.1 XM_017009851.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3616843 c.334G>A p.E112K Chr.5: 50760351-50760351 on Build GRCh38
5536488 c.343G>A p.E115K Chr.5: 50760360-50760360 on Build GRCh38
3616842 c.312C>T p.S104S Chr.5: 50760329-50760329 on Build GRCh38
6372764 c.300C>A p.N100K Chr.5: 50760317-50760317 on Build GRCh38
3616844 c.342G>A p.G114G Chr.5: 50760359-50760359 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026035643 Chr.5: 50760467-50760467 on Build GRCh38
rs746544306 Chr.5: 50760347-50760347 on Build GRCh38
rs922593603 Chr.5: 50760322-50760322 on Build GRCh38
rs764334878 Chr.5: 50760636-50760636 on Build GRCh38
rs767954941 Chr.5: 50760379-50760379 on Build GRCh38
rs575005459 Chr.5: 50760515-50760515 on Build GRCh38
rs377179868 Chr.5: 50760568-50760568 on Build GRCh38
rs746165254 Chr.5: 50760262-50760262 on Build GRCh38
rs749666309 Chr.5: 50760400-50760400 on Build GRCh38
rs543346737 Chr.5: 50760508-50760508 on Build GRCh38
rs760123129 Chr.5: 50760358-50760358 on Build GRCh38
rs906864064 Chr.5: 50760448-50760448 on Build GRCh38
rs765784524 Chr.5: 50760469-50760469 on Build GRCh38
rs777326067 Chr.5: 50760416-50760416 on Build GRCh38
rs763445428 Chr.5: 50760625-50760625 on Build GRCh38
rs772759774 Chr.5: 50760338-50760338 on Build GRCh38
rs1006030064 Chr.5: 50760561-50760561 on Build GRCh38
rs763210704 Chr.5: 50760371-50760371 on Build GRCh38
rs374961090 Chr.5: 50760399-50760399 on Build GRCh38
rs780342832 Chr.5: 50760312-50760312 on Build GRCh38
rs576867309 Chr.5: 50760599-50760599 on Build GRCh38
rs775186138 Chr.5: 50760366-50760366 on Build GRCh38
rs993281777 Chr.5: 50760456-50760456 on Build GRCh38
rs765268241 Chr.5: 50760376-50760377 on Build GRCh38
rs282548 Chr.5: 50760601-50760601 on Build GRCh38
rs144017619 Chr.5: 50760438-50760438 on Build GRCh38
rs111823389 Chr.5: 50760495-50760495 on Build GRCh38
rs113744716 Chr.5: 50760532-50760532 on Build GRCh38
rs758023234 Chr.5: 50760415-50760415 on Build GRCh38
rs554842223 Chr.5: 50760496-50760496 on Build GRCh38
rs903367844 Chr.5: 50760382-50760382 on Build GRCh38
rs562508205 Chr.5: 50760602-50760602 on Build GRCh38
rs750917236 Chr.5: 50760394-50760394 on Build GRCh38
rs756485100 Chr.5: 50760267-50760267 on Build GRCh38
rs747834399 Chr.5: 50760321-50760321 on Build GRCh38
rs756845409 Chr.5: 50760475-50760475 on Build GRCh38
rs894697102 Chr.5: 50760419-50760419 on Build GRCh38
rs757712678 Chr.5: 50760249-50760249 on Build GRCh38
rs910430930 Chr.5: 50760253-50760253 on Build GRCh38
rs770956022 Chr.5: 50760348-50760348 on Build GRCh38
rs781686661 Chr.5: 50760259-50760259 on Build GRCh38
rs756647634 Chr.5: 50760396-50760396 on Build GRCh38
rs751598703 Chr.5: 50760374-50760374 on Build GRCh38
rs776719074 Chr.5: 50760357-50760357 on Build GRCh38
rs776238227 Chr.5: 50760336-50760336 on Build GRCh38
rs539093526 Chr.5: 50760228-50760228 on Build GRCh38
rs149943672 Chr.5: 50760423-50760423 on Build GRCh38
rs764244795 Chr.5: 50760372-50760372 on Build GRCh38
rs139195302 Chr.5: 50760239-50760239 on Build GRCh38

View Full Product Details