Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 56920231-56920736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIER3
Entrez Gene ID
166968
Gene Name
MIER family member 3
Gene Aliases
-
UniGene
Hs.657594
Gene Chromosome Location
Chr.5: 56919602-56952130 on Build GRCh38
Transcripts
NM_001297598.1 NM_001297599.1 NM_152622.4 XM_011543216.1 XM_011543217.1 XM_017009149.1 NM_001171990.2 NR_136555.1

Gene Symbol
SETD9
Entrez Gene ID
133383
Gene Name
SET domain containing 9
Gene Aliases
C5orf35
UniGene
Hs.85950
Gene Chromosome Location
Chr.5: 56909260-56928900 on Build GRCh38
Transcripts
NM_001297598.1 NM_001297599.1 NM_152622.4 XM_011543216.1 XM_011543217.1 XM_017009149.1 NM_001171990.2 NR_136555.1

SNP Detail

SNP ID Chromosome Location
rs543952376 Chr.5: 56920342-56920342 on Build GRCh38
rs796118176 Chr.5: 56920624-56920625 on Build GRCh38
rs751644320 Chr.5: 56920367-56920367 on Build GRCh38
rs552271402 Chr.5: 56920394-56920394 on Build GRCh38
rs901719989 Chr.5: 56920417-56920417 on Build GRCh38
rs570789569 Chr.5: 56920448-56920448 on Build GRCh38
rs935331530 Chr.5: 56920644-56920644 on Build GRCh38
rs767458641 Chr.5: 56920457-56920457 on Build GRCh38
rs955464281 Chr.5: 56920492-56920492 on Build GRCh38
rs903806203 Chr.5: 56920427-56920427 on Build GRCh38
rs1000780506 Chr.5: 56920448-56920448 on Build GRCh38
rs78961030 Chr.5: 56920282-56920282 on Build GRCh38
rs574801229 Chr.5: 56920547-56920547 on Build GRCh38
rs1052788638 Chr.5: 56920401-56920401 on Build GRCh38
rs757910400 Chr.5: 56920486-56920488 on Build GRCh38
rs896333624 Chr.5: 56920348-56920348 on Build GRCh38
rs562225492 Chr.5: 56920449-56920450 on Build GRCh38
rs917683457 Chr.5: 56920604-56920604 on Build GRCh38
rs993146317 Chr.5: 56920512-56920512 on Build GRCh38
rs754667620 Chr.5: 56920605-56920605 on Build GRCh38
rs375564193 Chr.5: 56920535-56920535 on Build GRCh38
rs916748634 Chr.5: 56920312-56920312 on Build GRCh38
rs941657434 Chr.5: 56920518-56920520 on Build GRCh38
rs529673297 Chr.5: 56920479-56920479 on Build GRCh38
rs545849347 Chr.5: 56920635-56920635 on Build GRCh38
rs541437222 Chr.5: 56920519-56920519 on Build GRCh38
rs750736982 Chr.5: 56920274-56920274 on Build GRCh38
rs766136489 Chr.5: 56920528-56920528 on Build GRCh38
rs576961154 Chr.5: 56920308-56920308 on Build GRCh38
rs923254858 Chr.5: 56920698-56920698 on Build GRCh38
rs1023460311 Chr.5: 56920407-56920407 on Build GRCh38
rs755916198 Chr.5: 56920508-56920508 on Build GRCh38
rs534728486 Chr.5: 56920458-56920458 on Build GRCh38
rs979524343 Chr.5: 56920634-56920634 on Build GRCh38
rs925293927 Chr.5: 56920383-56920383 on Build GRCh38
rs142127120 Chr.5: 56920610-56920610 on Build GRCh38
rs759927745 Chr.5: 56920462-56920462 on Build GRCh38
rs553584491 Chr.5: 56920461-56920461 on Build GRCh38
rs948152197 Chr.5: 56920334-56920334 on Build GRCh38
rs560063777 Chr.5: 56920599-56920599 on Build GRCh38
rs191367711 Chr.5: 56920345-56920345 on Build GRCh38
rs761705224 Chr.5: 56920385-56920385 on Build GRCh38
rs1038036928 Chr.5: 56920273-56920273 on Build GRCh38
rs891811251 Chr.5: 56920679-56920679 on Build GRCh38
rs535650576 Chr.5: 56920589-56920589 on Build GRCh38
rs1044958562 Chr.5: 56920625-56920625 on Build GRCh38
rs753614540 Chr.5: 56920548-56920548 on Build GRCh38
rs1052482810 Chr.5: 56920685-56920685 on Build GRCh38

View Full Product Details