Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 58973328-58973825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE4D
Entrez Gene ID
5144
Gene Name
phosphodiesterase 4D
Gene Aliases
ACRDYS2, DPDE3, HSPDE4D, PDE43, PDE4DN2, STRK1
UniGene
Hs.117545
Gene Chromosome Location
Chr.5: 58969038-60488098 on Build GRCh38
Transcripts
NM_001104631.1 NM_001165899.1 NM_001197218.1 NM_001197219.1 NM_001197220.1 NM_001197221.1 NM_001197222.1 NM_001197223.1 NM_006203.4 XM_005248537.3 XM_005248538.4 XM_011543469.1 XM_011543470.2 XM_011543471.1 XM_011543473.1 XM_011543477.2 XM_017009565.1 XM_017009566.1 XM_017009567.1 XM_017009568.1 XM_017009569.1 XM_017009570.1 XM_017009571.1

SNP Detail

SNP ID Chromosome Location
rs755406685 Chr.5: 58973798-58973798 on Build GRCh38
rs570292182 Chr.5: 58973397-58973397 on Build GRCh38
rs553526225 Chr.5: 58973526-58973526 on Build GRCh38
rs1012317949 Chr.5: 58973682-58973682 on Build GRCh38
rs1036017424 Chr.5: 58973732-58973732 on Build GRCh38
rs971330026 Chr.5: 58973412-58973412 on Build GRCh38
rs749948288 Chr.5: 58973546-58973546 on Build GRCh38
rs573384171 Chr.5: 58973792-58973793 on Build GRCh38
rs897606388 Chr.5: 58973800-58973800 on Build GRCh38
rs1044567243 Chr.5: 58973773-58973774 on Build GRCh38
rs766480342 Chr.5: 58973458-58973459 on Build GRCh38
rs888034262 Chr.5: 58973661-58973661 on Build GRCh38
rs923446839 Chr.5: 58973630-58973630 on Build GRCh38
rs979452973 Chr.5: 58973566-58973566 on Build GRCh38
rs775984561 Chr.5: 58973386-58973386 on Build GRCh38
rs3839217 Chr.5: 58973789-58973790 on Build GRCh38
rs772386939 Chr.5: 58973365-58973365 on Build GRCh38
rs895229530 Chr.5: 58973674-58973675 on Build GRCh38
rs935389957 Chr.5: 58973606-58973606 on Build GRCh38
rs556877242 Chr.5: 58973369-58973369 on Build GRCh38
rs985619216 Chr.5: 58973600-58973601 on Build GRCh38
rs530742885 Chr.5: 58973684-58973684 on Build GRCh38
rs780488224 Chr.5: 58973438-58973439 on Build GRCh38
rs183295023 Chr.5: 58973437-58973437 on Build GRCh38
rs886060709 Chr.5: 58973396-58973396 on Build GRCh38
rs541866920 Chr.5: 58973560-58973560 on Build GRCh38
rs968453316 Chr.5: 58973564-58973564 on Build GRCh38
rs199967190 Chr.5: 58973788-58973788 on Build GRCh38
rs577495801 Chr.5: 58973464-58973464 on Build GRCh38
rs17719258 Chr.5: 58973413-58973413 on Build GRCh38
rs559220819 Chr.5: 58973463-58973463 on Build GRCh38
rs1032019041 Chr.5: 58973755-58973755 on Build GRCh38
rs914459105 Chr.5: 58973545-58973545 on Build GRCh38
rs140301103 Chr.5: 58973788-58973789 on Build GRCh38
rs1022738915 Chr.5: 58973688-58973689 on Build GRCh38
rs372901892 Chr.5: 58973776-58973777 on Build GRCh38
rs1002769389 Chr.5: 58973724-58973724 on Build GRCh38
rs907052777 Chr.5: 58973707-58973707 on Build GRCh38
rs887449542 Chr.5: 58973396-58973396 on Build GRCh38
rs886060710 Chr.5: 58973630-58973630 on Build GRCh38
rs202165590 Chr.5: 58973795-58973795 on Build GRCh38
rs932135974 Chr.5: 58973646-58973646 on Build GRCh38
rs1051123488 Chr.5: 58973655-58973655 on Build GRCh38
rs961762846 Chr.5: 58973462-58973462 on Build GRCh38
rs541604095 Chr.5: 58973487-58973487 on Build GRCh38
rs1031316461 Chr.5: 58973498-58973498 on Build GRCh38
rs574827993 Chr.5: 58973535-58973535 on Build GRCh38
rs145750635 Chr.5: 58973629-58973629 on Build GRCh38
rs912765104 Chr.5: 58973604-58973604 on Build GRCh38
rs1021335909 Chr.5: 58973784-58973785 on Build GRCh38
rs765434017 Chr.5: 58973441-58973464 on Build GRCh38
rs989816935 Chr.5: 58973782-58973783 on Build GRCh38
rs550896063 Chr.5: 58973581-58973581 on Build GRCh38
rs386403886 Chr.5: 58973790-58973791 on Build GRCh38
rs995582527 Chr.5: 58973360-58973361 on Build GRCh38
rs764716521 Chr.5: 58973424-58973424 on Build GRCh38
rs569185580 Chr.5: 58973703-58973703 on Build GRCh38
rs989869291 Chr.5: 58973529-58973529 on Build GRCh38
rs991825244 Chr.5: 58973534-58973534 on Build GRCh38
rs886060711 Chr.5: 58973641-58973642 on Build GRCh38
rs931316042 Chr.5: 58973638-58973638 on Build GRCh38

View Full Product Details