Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60903298-60903821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC8
Entrez Gene ID
1161
Gene Name
ERCC excision repair 8, CSA ubiquitin ligase complex subunit
Gene Aliases
CKN1, CSA, UVSS2
UniGene
Hs.435237
Gene Chromosome Location
Chr.5: 60873832-60945078 on Build GRCh38
Transcripts
NM_000082.3 NM_001007233.2 NM_001007234.2 NM_001290285.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1671667 c.524C>T p.S175F Chr.5: 60903674-60903674 on Build GRCh38
1069216 c.532T>C p.C178R Chr.5: 60903666-60903666 on Build GRCh38
449946 c.521A>G p.K174R Chr.5: 60903677-60903677 on Build GRCh38
5868672 c.494G>A p.G165E Chr.5: 60903704-60903704 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757206580 Chr.5: 60903752-60903752 on Build GRCh38
rs752236421 Chr.5: 60903599-60903599 on Build GRCh38
rs369365521 Chr.5: 60903606-60903606 on Build GRCh38
rs951705515 Chr.5: 60903360-60903360 on Build GRCh38
rs756333058 Chr.5: 60903764-60903764 on Build GRCh38
rs1008673098 Chr.5: 60903630-60903631 on Build GRCh38
rs201833381 Chr.5: 60903626-60903626 on Build GRCh38
rs927024717 Chr.5: 60903472-60903472 on Build GRCh38
rs557578028 Chr.5: 60903379-60903379 on Build GRCh38
rs377155839 Chr.5: 60903746-60903746 on Build GRCh38
rs1038144 Chr.5: 60903374-60903374 on Build GRCh38
rs368308377 Chr.5: 60903578-60903578 on Build GRCh38
rs772429684 Chr.5: 60903680-60903680 on Build GRCh38
rs770494554 Chr.5: 60903613-60903624 on Build GRCh38
rs577778078 Chr.5: 60903401-60903401 on Build GRCh38
rs142139247 Chr.5: 60903521-60903521 on Build GRCh38
rs138212997 Chr.5: 60903505-60903505 on Build GRCh38
rs962991558 Chr.5: 60903628-60903628 on Build GRCh38
rs201126529 Chr.5: 60903733-60903733 on Build GRCh38
rs781538504 Chr.5: 60903532-60903532 on Build GRCh38
rs191031496 Chr.5: 60903566-60903566 on Build GRCh38
rs916892488 Chr.5: 60903525-60903525 on Build GRCh38
rs545565172 Chr.5: 60903390-60903390 on Build GRCh38
rs4647102 Chr.5: 60903536-60903536 on Build GRCh38
rs886042104 Chr.5: 60903688-60903688 on Build GRCh38
rs978185345 Chr.5: 60903780-60903780 on Build GRCh38
rs778380701 Chr.5: 60903750-60903756 on Build GRCh38
rs372370196 Chr.5: 60903587-60903587 on Build GRCh38
rs201840328 Chr.5: 60903665-60903665 on Build GRCh38
rs767821376 Chr.5: 60903584-60903584 on Build GRCh38
rs977090932 Chr.5: 60903734-60903734 on Build GRCh38
rs959156200 Chr.5: 60903691-60903691 on Build GRCh38
rs759120854 Chr.5: 60903588-60903588 on Build GRCh38
rs576560045 Chr.5: 60903336-60903336 on Build GRCh38
rs756058348 Chr.5: 60903446-60903446 on Build GRCh38
rs961490880 Chr.5: 60903670-60903670 on Build GRCh38
rs182401243 Chr.5: 60903575-60903575 on Build GRCh38
rs762499617 Chr.5: 60903732-60903732 on Build GRCh38
rs764057678 Chr.5: 60903603-60903603 on Build GRCh38
rs1030174627 Chr.5: 60903605-60903605 on Build GRCh38
rs925872848 Chr.5: 60903776-60903776 on Build GRCh38
rs764937712 Chr.5: 60903707-60903707 on Build GRCh38
rs980969833 Chr.5: 60903456-60903456 on Build GRCh38
rs992568227 Chr.5: 60903696-60903696 on Build GRCh38
rs758331383 Chr.5: 60903600-60903600 on Build GRCh38
rs961438517 Chr.5: 60903368-60903368 on Build GRCh38
rs750242117 Chr.5: 60903763-60903763 on Build GRCh38
rs770002373 Chr.5: 60903553-60903553 on Build GRCh38
rs992888617 Chr.5: 60903515-60903515 on Build GRCh38
rs748614275 Chr.5: 60903676-60903676 on Build GRCh38
rs1017200468 Chr.5: 60903382-60903382 on Build GRCh38
rs774899829 Chr.5: 60903708-60903708 on Build GRCh38
rs565247324 Chr.5: 60903639-60903639 on Build GRCh38
rs937128607 Chr.5: 60903501-60903501 on Build GRCh38
rs746110609 Chr.5: 60903627-60903627 on Build GRCh38
rs763429701 Chr.5: 60903557-60903557 on Build GRCh38
rs745699331 Chr.5: 60903413-60903413 on Build GRCh38
rs35366433 Chr.5: 60903694-60903694 on Build GRCh38
rs527752850 Chr.5: 60903647-60903647 on Build GRCh38
rs962836522 Chr.5: 60903395-60903395 on Build GRCh38
rs919753311 Chr.5: 60903398-60903398 on Build GRCh38
rs140185830 Chr.5: 60903652-60903652 on Build GRCh38
rs542550501 Chr.5: 60903556-60903556 on Build GRCh38
rs748762631 Chr.5: 60903562-60903562 on Build GRCh38
rs145610983 Chr.5: 60903689-60903689 on Build GRCh38
rs916988854 Chr.5: 60903717-60903717 on Build GRCh38
rs774944497 Chr.5: 60903666-60903666 on Build GRCh38

View Full Product Details