Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69306725-69307221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC125
Entrez Gene ID
202243
Gene Name
coiled-coil domain containing 125
Gene Aliases
KENAE
UniGene
Hs.654850
Gene Chromosome Location
Chr.5: 69273087-69333069 on Build GRCh38
Transcripts
NM_001297696.1 NM_001297697.1 NM_176816.4 XM_005248458.4 XM_005248459.4 XM_005248461.3 XM_005248463.3 XM_005248464.3 XM_006714569.3 XM_006714570.3 XM_011543255.2 XM_011543256.2 XM_011543257.2 XM_011543258.2 XM_011543259.2 XM_011543260.2 XM_017009206.1 XM_017009207.1 XM_017009208.1 XM_017009209.1 XM_017009210.1 XM_017009211.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3429597 c.591G>T p.E197D Chr.5: 69306843-69306843 on Build GRCh38
1069720 c.559G>T p.E187* Chr.5: 69306875-69306875 on Build GRCh38
7166841 c.609A>G p.K203K Chr.5: 69306825-69306825 on Build GRCh38
738323 c.589G>C p.E197Q Chr.5: 69306845-69306845 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372311055 Chr.5: 69306846-69306846 on Build GRCh38
rs766265303 Chr.5: 69306849-69306849 on Build GRCh38
rs34904038 Chr.5: 69306995-69306996 on Build GRCh38
rs201630962 Chr.5: 69306803-69306803 on Build GRCh38
rs374875429 Chr.5: 69306941-69306941 on Build GRCh38
rs539017688 Chr.5: 69307189-69307189 on Build GRCh38
rs6881767 Chr.5: 69306919-69306919 on Build GRCh38
rs745899356 Chr.5: 69306811-69306811 on Build GRCh38
rs781765630 Chr.5: 69306810-69306810 on Build GRCh38
rs759026633 Chr.5: 69306873-69306873 on Build GRCh38
rs752341021 Chr.5: 69306912-69306912 on Build GRCh38
rs903921264 Chr.5: 69307004-69307004 on Build GRCh38
rs763122284 Chr.5: 69306822-69306822 on Build GRCh38
rs1043829029 Chr.5: 69306980-69306980 on Build GRCh38
rs896776983 Chr.5: 69307125-69307125 on Build GRCh38
rs778064356 Chr.5: 69306782-69306782 on Build GRCh38
rs923209673 Chr.5: 69306866-69306866 on Build GRCh38
rs771889626 Chr.5: 69306843-69306843 on Build GRCh38
rs200132145 Chr.5: 69306812-69306812 on Build GRCh38
rs985337001 Chr.5: 69307190-69307190 on Build GRCh38
rs1036019340 Chr.5: 69306830-69306830 on Build GRCh38
rs754514202 Chr.5: 69306769-69306769 on Build GRCh38
rs747536262 Chr.5: 69306802-69306802 on Build GRCh38
rs756229786 Chr.5: 69306948-69306948 on Build GRCh38
rs1024267292 Chr.5: 69307169-69307169 on Build GRCh38
rs565339465 Chr.5: 69307185-69307185 on Build GRCh38
rs753283292 Chr.5: 69306767-69306767 on Build GRCh38
rs547048628 Chr.5: 69307147-69307147 on Build GRCh38
rs778489126 Chr.5: 69306776-69306776 on Build GRCh38
rs999503718 Chr.5: 69307006-69307006 on Build GRCh38
rs772282101 Chr.5: 69306874-69306875 on Build GRCh38
rs888963240 Chr.5: 69306944-69306944 on Build GRCh38
rs57580570 Chr.5: 69306962-69306962 on Build GRCh38
rs112304723 Chr.5: 69307146-69307146 on Build GRCh38
rs35682673 Chr.5: 69306757-69306758 on Build GRCh38
rs558236427 Chr.5: 69306859-69306859 on Build GRCh38
rs764770070 Chr.5: 69306876-69306876 on Build GRCh38
rs1001168390 Chr.5: 69307130-69307130 on Build GRCh38
rs758760062 Chr.5: 69306781-69306781 on Build GRCh38
rs111665928 Chr.5: 69306900-69306900 on Build GRCh38
rs529021981 Chr.5: 69307043-69307043 on Build GRCh38
rs892323562 Chr.5: 69307158-69307158 on Build GRCh38
rs1052345804 Chr.5: 69307155-69307155 on Build GRCh38
rs760527388 Chr.5: 69306848-69306848 on Build GRCh38
rs781744529 Chr.5: 69306938-69306938 on Build GRCh38
rs371146327 Chr.5: 69306920-69306920 on Build GRCh38
rs1013798649 Chr.5: 69307151-69307151 on Build GRCh38
rs746084234 Chr.5: 69306870-69306871 on Build GRCh38
rs758042975 Chr.5: 69306916-69306916 on Build GRCh38
rs564347627 Chr.5: 69306820-69306820 on Build GRCh38
rs768205744 Chr.5: 69307095-69307095 on Build GRCh38
rs752396674 Chr.5: 69306777-69306777 on Build GRCh38

View Full Product Details