Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 72358458-72358961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTCD2
Entrez Gene ID
79810
Gene Name
pentatricopeptide repeat domain 2
Gene Aliases
-
UniGene
Hs.126906
Gene Chromosome Location
Chr.5: 72320367-72368395 on Build GRCh38
Transcripts
NM_001284403.1 NM_001284404.1 NM_001284405.1 NM_024754.4 XM_005248601.2 XM_005248603.2 XM_017009866.1 XM_017009867.1 XM_017009868.1

SNP Detail

SNP ID Chromosome Location
rs557677381 Chr.5: 72358755-72358755 on Build GRCh38
rs550439769 Chr.5: 72358516-72358516 on Build GRCh38
rs1034476824 Chr.5: 72358842-72358842 on Build GRCh38
rs147073609 Chr.5: 72358823-72358823 on Build GRCh38
rs576017866 Chr.5: 72358811-72358811 on Build GRCh38
rs974955116 Chr.5: 72358606-72358606 on Build GRCh38
rs759657769 Chr.5: 72358500-72358500 on Build GRCh38
rs770590858 Chr.5: 72358786-72358786 on Build GRCh38
rs73761771 Chr.5: 72358695-72358695 on Build GRCh38
rs989196574 Chr.5: 72358656-72358656 on Build GRCh38
rs1021942366 Chr.5: 72358895-72358895 on Build GRCh38
rs553464139 Chr.5: 72358677-72358677 on Build GRCh38
rs986703229 Chr.5: 72358917-72358917 on Build GRCh38
rs561314452 Chr.5: 72358482-72358483 on Build GRCh38
rs74714328 Chr.5: 72358770-72358770 on Build GRCh38
rs1001931164 Chr.5: 72358831-72358831 on Build GRCh38
rs756010848 Chr.5: 72358645-72358645 on Build GRCh38
rs80066877 Chr.5: 72358772-72358772 on Build GRCh38
rs912248318 Chr.5: 72358714-72358714 on Build GRCh38
rs1025447929 Chr.5: 72358908-72358908 on Build GRCh38
rs745453080 Chr.5: 72358861-72358861 on Build GRCh38
rs916593662 Chr.5: 72358763-72358763 on Build GRCh38
rs903594241 Chr.5: 72358780-72358780 on Build GRCh38
rs367804692 Chr.5: 72358872-72358872 on Build GRCh38
rs1027707530 Chr.5: 72358581-72358581 on Build GRCh38
rs951197649 Chr.5: 72358912-72358912 on Build GRCh38
rs895145658 Chr.5: 72358521-72358521 on Build GRCh38
rs902004756 Chr.5: 72358821-72358821 on Build GRCh38
rs1004958146 Chr.5: 72358806-72358806 on Build GRCh38
rs942353107 Chr.5: 72358721-72358721 on Build GRCh38
rs953726128 Chr.5: 72358849-72358849 on Build GRCh38
rs983482735 Chr.5: 72358796-72358796 on Build GRCh38
rs200176631 Chr.5: 72358592-72358592 on Build GRCh38
rs1055040360 Chr.5: 72358803-72358803 on Build GRCh38
rs567706710 Chr.5: 72358602-72358602 on Build GRCh38
rs1010504113 Chr.5: 72358853-72358853 on Build GRCh38
rs755094333 Chr.5: 72358596-72358596 on Build GRCh38
rs545731866 Chr.5: 72358482-72358482 on Build GRCh38
rs568862434 Chr.5: 72358580-72358580 on Build GRCh38
rs1055009991 Chr.5: 72358757-72358757 on Build GRCh38
rs978353732 Chr.5: 72358697-72358697 on Build GRCh38
rs7734699 Chr.5: 72358931-72358931 on Build GRCh38
rs939264128 Chr.5: 72358505-72358505 on Build GRCh38
rs1024325273 Chr.5: 72358584-72358585 on Build GRCh38
rs939289192 Chr.5: 72358734-72358734 on Build GRCh38
rs771670813 Chr.5: 72358876-72358876 on Build GRCh38
rs748146102 Chr.5: 72358616-72358616 on Build GRCh38
rs143095749 Chr.5: 72358670-72358670 on Build GRCh38
rs112472809 Chr.5: 72358824-72358824 on Build GRCh38
rs775547138 Chr.5: 72358733-72358733 on Build GRCh38
rs73102462 Chr.5: 72358899-72358899 on Build GRCh38
rs760806857 Chr.5: 72358789-72358789 on Build GRCh38
rs539079104 Chr.5: 72358706-72358706 on Build GRCh38
rs138050489 Chr.5: 72358590-72358590 on Build GRCh38
rs1040975434 Chr.5: 72358814-72358814 on Build GRCh38
rs974309191 Chr.5: 72358623-72358623 on Build GRCh38
rs11555200 Chr.5: 72358869-72358869 on Build GRCh38
rs994926585 Chr.5: 72358527-72358527 on Build GRCh38
rs930603571 Chr.5: 72358771-72358771 on Build GRCh38
rs956278249 Chr.5: 72358522-72358522 on Build GRCh38
rs1000759692 Chr.5: 72358504-72358504 on Build GRCh38

View Full Product Details