Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 72989362-72989909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCHO2
Entrez Gene ID
115548
Gene Name
FCH domain only 2
Gene Aliases
-
UniGene
Hs.165762
Gene Chromosome Location
Chr.5: 72955981-73090522 on Build GRCh38
Transcripts
NM_001146032.1 NM_138782.2 XM_017009016.1 XM_017009017.1 XM_017009018.1 XM_017009019.1 XM_017009020.1 XM_017009021.1 XM_017009022.1 XM_017009023.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3726456 c.134T>C p.I45T Chr.5: 72989435-72989435 on Build GRCh38
3726458 c.134T>C p.I45T Chr.5: 72989435-72989435 on Build GRCh38
3856218 c.160A>C p.T54P Chr.5: 72989461-72989461 on Build GRCh38
3856217 c.160A>C p.T54P Chr.5: 72989461-72989461 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1023971219 Chr.5: 72989770-72989770 on Build GRCh38
rs974458283 Chr.5: 72989616-72989616 on Build GRCh38
rs761987775 Chr.5: 72989531-72989534 on Build GRCh38
rs769968574 Chr.5: 72989516-72989517 on Build GRCh38
rs942520069 Chr.5: 72989735-72989735 on Build GRCh38
rs758962752 Chr.5: 72989540-72989540 on Build GRCh38
rs774413260 Chr.5: 72989745-72989745 on Build GRCh38
rs893851927 Chr.5: 72989558-72989558 on Build GRCh38
rs768412203 Chr.5: 72989675-72989675 on Build GRCh38
rs772580984 Chr.5: 72989455-72989455 on Build GRCh38
rs114392573 Chr.5: 72989734-72989734 on Build GRCh38
rs371860690 Chr.5: 72989511-72989511 on Build GRCh38
rs905382268 Chr.5: 72989882-72989883 on Build GRCh38
rs113928792 Chr.5: 72989705-72989705 on Build GRCh38
rs912691401 Chr.5: 72989685-72989685 on Build GRCh38
rs181048129 Chr.5: 72989666-72989666 on Build GRCh38
rs368553021 Chr.5: 72989461-72989461 on Build GRCh38
rs942657951 Chr.5: 72989395-72989395 on Build GRCh38
rs369327724 Chr.5: 72989534-72989534 on Build GRCh38
rs761669818 Chr.5: 72989387-72989387 on Build GRCh38
rs578172606 Chr.5: 72989496-72989496 on Build GRCh38
rs536410503 Chr.5: 72989840-72989840 on Build GRCh38
rs145570949 Chr.5: 72989637-72989637 on Build GRCh38
rs1045162574 Chr.5: 72989845-72989845 on Build GRCh38
rs765911995 Chr.5: 72989466-72989466 on Build GRCh38
rs866775339 Chr.5: 72989576-72989576 on Build GRCh38
rs542320104 Chr.5: 72989603-72989603 on Build GRCh38
rs183638705 Chr.5: 72989786-72989786 on Build GRCh38
rs911137661 Chr.5: 72989392-72989394 on Build GRCh38
rs963228262 Chr.5: 72989869-72989869 on Build GRCh38
rs747962037 Chr.5: 72989458-72989458 on Build GRCh38
rs185151065 Chr.5: 72989527-72989527 on Build GRCh38
rs1054807629 Chr.5: 72989746-72989746 on Build GRCh38
rs781016745 Chr.5: 72989405-72989405 on Build GRCh38
rs535107551 Chr.5: 72989762-72989762 on Build GRCh38
rs976716491 Chr.5: 72989736-72989736 on Build GRCh38
rs751562610 Chr.5: 72989551-72989551 on Build GRCh38
rs972780845 Chr.5: 72989658-72989658 on Build GRCh38
rs560638348 Chr.5: 72989605-72989605 on Build GRCh38
rs1052546478 Chr.5: 72989554-72989554 on Build GRCh38
rs963008862 Chr.5: 72989594-72989594 on Build GRCh38
rs779777475 Chr.5: 72989532-72989532 on Build GRCh38
rs1034544408 Chr.5: 72989827-72989827 on Build GRCh38
rs1038898850 Chr.5: 72989710-72989710 on Build GRCh38
rs555399777 Chr.5: 72989781-72989781 on Build GRCh38
rs772325442 Chr.5: 72989766-72989766 on Build GRCh38
rs866273726 Chr.5: 72989715-72989715 on Build GRCh38
rs980243817 Chr.5: 72989486-72989486 on Build GRCh38
rs749121661 Chr.5: 72989571-72989571 on Build GRCh38
rs549342427 Chr.5: 72989591-72989591 on Build GRCh38
rs1035661781 Chr.5: 72989635-72989635 on Build GRCh38
rs199818912 Chr.5: 72989546-72989546 on Build GRCh38
rs189995922 Chr.5: 72989553-72989553 on Build GRCh38
rs974338357 Chr.5: 72989878-72989878 on Build GRCh38
rs553676077 Chr.5: 72989407-72989407 on Build GRCh38
rs549539699 Chr.5: 72989880-72989880 on Build GRCh38
rs532319006 Chr.5: 72989709-72989709 on Build GRCh38

View Full Product Details