Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80039990-80040492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THBS4
Entrez Gene ID
7060
Gene Name
thrombospondin 4
Gene Aliases
TSP-4, TSP4
UniGene
Hs.211426
Gene Chromosome Location
Chr.5: 79991296-80083287 on Build GRCh38
Transcripts
NM_001306212.1 NM_001306213.1 NM_001306214.1 NM_003248.5 XM_017009798.1 XM_017009799.1 XM_017009800.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7264388 c.276G>T p.M92I Chr.5: 80040264-80040264 on Build GRCh38
7234703 c.128C>T p.P43L Chr.5: 80040116-80040116 on Build GRCh38
4457314 c.103C>T p.P35S Chr.5: 80040091-80040091 on Build GRCh38
1070601 c.160G>A p.A54T Chr.5: 80040148-80040148 on Build GRCh38
739093 c.117G>T p.Q39H Chr.5: 80040105-80040105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962837846 Chr.5: 80040463-80040463 on Build GRCh38
rs776280262 Chr.5: 80040217-80040217 on Build GRCh38
rs971884981 Chr.5: 80040462-80040462 on Build GRCh38
rs372405988 Chr.5: 80040234-80040234 on Build GRCh38
rs898398226 Chr.5: 80040352-80040352 on Build GRCh38
rs140883597 Chr.5: 80040374-80040374 on Build GRCh38
rs369576000 Chr.5: 80040210-80040210 on Build GRCh38
rs760078973 Chr.5: 80040300-80040300 on Build GRCh38
rs754194222 Chr.5: 80040088-80040088 on Build GRCh38
rs763835245 Chr.5: 80040055-80040055 on Build GRCh38
rs536778118 Chr.5: 80040014-80040014 on Build GRCh38
rs149738326 Chr.5: 80040419-80040419 on Build GRCh38
rs139299549 Chr.5: 80040146-80040146 on Build GRCh38
rs200373343 Chr.5: 80040268-80040268 on Build GRCh38
rs958714361 Chr.5: 80040461-80040461 on Build GRCh38
rs758421463 Chr.5: 80040230-80040230 on Build GRCh38
rs200185391 Chr.5: 80040455-80040455 on Build GRCh38
rs775293278 Chr.5: 80040043-80040043 on Build GRCh38
rs890349882 Chr.5: 80040143-80040143 on Build GRCh38
rs971605303 Chr.5: 80040432-80040432 on Build GRCh38
rs983068843 Chr.5: 80040429-80040429 on Build GRCh38
rs762262683 Chr.5: 80040159-80040159 on Build GRCh38
rs769604290 Chr.5: 80040144-80040144 on Build GRCh38
rs201953068 Chr.5: 80040041-80040041 on Build GRCh38
rs541422494 Chr.5: 80040213-80040213 on Build GRCh38
rs752848388 Chr.5: 80040221-80040221 on Build GRCh38
rs11741724 Chr.5: 80040454-80040454 on Build GRCh38
rs761304448 Chr.5: 80040195-80040195 on Build GRCh38
rs759659411 Chr.5: 80040227-80040233 on Build GRCh38
rs373743893 Chr.5: 80040111-80040111 on Build GRCh38
rs768535270 Chr.5: 80040263-80040263 on Build GRCh38
rs150862893 Chr.5: 80040121-80040121 on Build GRCh38
rs372241107 Chr.5: 80040205-80040205 on Build GRCh38
rs60660392 Chr.5: 80040448-80040448 on Build GRCh38
rs111349419 Chr.5: 80040118-80040118 on Build GRCh38
rs397811966 Chr.5: 80040454-80040454 on Build GRCh38
rs998958386 Chr.5: 80040465-80040465 on Build GRCh38
rs971652389 Chr.5: 80040421-80040421 on Build GRCh38
rs766990461 Chr.5: 80040189-80040189 on Build GRCh38
rs761263904 Chr.5: 80040167-80040167 on Build GRCh38
rs150043767 Chr.5: 80040214-80040214 on Build GRCh38
rs749062616 Chr.5: 80040148-80040148 on Build GRCh38
rs780117789 Chr.5: 80040235-80040235 on Build GRCh38
rs768472544 Chr.5: 80040149-80040149 on Build GRCh38
rs745758223 Chr.5: 80040142-80040142 on Build GRCh38
rs367779999 Chr.5: 80040120-80040120 on Build GRCh38
rs774045443 Chr.5: 80040057-80040057 on Build GRCh38
rs772816812 Chr.5: 80040297-80040297 on Build GRCh38
rs750907207 Chr.5: 80040077-80040077 on Build GRCh38
rs766710790 Chr.5: 80040084-80040084 on Build GRCh38
rs756441727 Chr.5: 80040079-80040079 on Build GRCh38
rs775493782 Chr.5: 80040147-80040147 on Build GRCh38
rs1002963039 Chr.5: 80040456-80040456 on Build GRCh38
rs556334583 Chr.5: 80040016-80040016 on Build GRCh38
rs17881847 Chr.5: 80040152-80040152 on Build GRCh38
rs908702340 Chr.5: 80040450-80040450 on Build GRCh38
rs1009258837 Chr.5: 80040151-80040151 on Build GRCh38
rs536099381 Chr.5: 80040101-80040101 on Build GRCh38
rs1034825675 Chr.5: 80040460-80040460 on Build GRCh38
rs1038497230 Chr.5: 80040085-80040085 on Build GRCh38
rs572372132 Chr.5: 80040193-80040193 on Build GRCh38
rs745681783 Chr.5: 80040039-80040039 on Build GRCh38
rs62363963 Chr.5: 80040116-80040116 on Build GRCh38
rs543977261 Chr.5: 80040325-80040325 on Build GRCh38
rs369984137 Chr.5: 80040250-80040250 on Build GRCh38
rs1032690390 Chr.5: 80040211-80040211 on Build GRCh38
rs780996876 Chr.5: 80040130-80040130 on Build GRCh38
rs1038324194 Chr.5: 80040344-80040344 on Build GRCh38
rs771446876 Chr.5: 80040302-80040302 on Build GRCh38
rs767935939 Chr.5: 80040074-80040074 on Build GRCh38
rs774338599 Chr.5: 80040026-80040027 on Build GRCh38
rs755230190 Chr.5: 80040093-80040093 on Build GRCh38
rs774098673 Chr.5: 80040162-80040162 on Build GRCh38
rs767991849 Chr.5: 80040286-80040286 on Build GRCh38
rs771613271 Chr.5: 80040269-80040269 on Build GRCh38
rs762753944 Chr.5: 80040045-80040045 on Build GRCh38
rs761385018 Chr.5: 80040064-80040064 on Build GRCh38

View Full Product Details