Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 93740793-93741294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM172A
Entrez Gene ID
83989
Gene Name
family with sequence similarity 172 member A
Gene Aliases
C5orf21
UniGene
Hs.600086
Gene Chromosome Location
Chr.5: 93615130-94111699 on Build GRCh38
Transcripts
NM_001163417.1 NM_001163418.1 NM_032042.5 NR_028080.1 XM_005272105.3 XM_005272106.3 XM_005272108.3 XM_005272110.3 XM_006714717.3 XM_006714718.3 XM_006714719.3 XM_011543671.2 XM_011543672.2 XM_011543673.2 XM_017009952.1 XM_017009953.1 XM_017009955.1 XM_017009956.1 XM_017009957.1 XM_017009958.1 XM_017009960.1 NM_153216.1

Gene Symbol
POU5F2
Entrez Gene ID
134187
Gene Name
POU domain class 5, transcription factor 2
Gene Aliases
SPRM-1
UniGene
Hs.678995
Gene Chromosome Location
Chr.5: 93740309-93741603 on Build GRCh38
Transcripts
NM_001163417.1 NM_001163418.1 NM_032042.5 NR_028080.1 XM_005272105.3 XM_005272106.3 XM_005272108.3 XM_005272110.3 XM_006714717.3 XM_006714718.3 XM_006714719.3 XM_011543671.2 XM_011543672.2 XM_011543673.2 XM_017009952.1 XM_017009953.1 XM_017009955.1 XM_017009956.1 XM_017009957.1 XM_017009958.1 XM_017009960.1 NM_153216.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5942002 c.331C>T p.P111S Chr.5: 93741233-93741233 on Build GRCh38
3856908 c.342G>A p.P114P Chr.5: 93741222-93741222 on Build GRCh38
5975711 c.414G>A p.L138L Chr.5: 93741150-93741150 on Build GRCh38
3828603 c.636G>C p.W212C Chr.5: 93740928-93740928 on Build GRCh38
5808165 c.345G>T p.P115P Chr.5: 93741219-93741219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559769557 Chr.5: 93741178-93741178 on Build GRCh38
rs374189353 Chr.5: 93741276-93741276 on Build GRCh38
rs546181953 Chr.5: 93741141-93741141 on Build GRCh38
rs748177777 Chr.5: 93740993-93740993 on Build GRCh38
rs777533339 Chr.5: 93740876-93740876 on Build GRCh38
rs768452555 Chr.5: 93741249-93741249 on Build GRCh38
rs543468590 Chr.5: 93740905-93740905 on Build GRCh38
rs760297101 Chr.5: 93741050-93741050 on Build GRCh38
rs757009479 Chr.5: 93740886-93740886 on Build GRCh38
rs369080417 Chr.5: 93740823-93740823 on Build GRCh38
rs745818915 Chr.5: 93740900-93740900 on Build GRCh38
rs961225576 Chr.5: 93741223-93741223 on Build GRCh38
rs552948117 Chr.5: 93740874-93740874 on Build GRCh38
rs771230898 Chr.5: 93740908-93740908 on Build GRCh38
rs1017135332 Chr.5: 93740931-93740931 on Build GRCh38
rs376579858 Chr.5: 93741175-93741175 on Build GRCh38
rs772689274 Chr.5: 93741252-93741252 on Build GRCh38
rs199511635 Chr.5: 93740901-93740901 on Build GRCh38
rs371786597 Chr.5: 93740872-93740872 on Build GRCh38
rs746749908 Chr.5: 93740882-93740882 on Build GRCh38
rs142976192 Chr.5: 93741087-93741087 on Build GRCh38
rs758502965 Chr.5: 93741111-93741111 on Build GRCh38
rs535265639 Chr.5: 93741042-93741042 on Build GRCh38
rs755947826 Chr.5: 93741080-93741080 on Build GRCh38
rs748374007 Chr.5: 93740904-93740904 on Build GRCh38
rs200489758 Chr.5: 93741026-93741026 on Build GRCh38
rs754470635 Chr.5: 93740845-93740845 on Build GRCh38
rs772280728 Chr.5: 93741007-93741007 on Build GRCh38
rs371062975 Chr.5: 93741260-93741260 on Build GRCh38
rs1042110357 Chr.5: 93740854-93740854 on Build GRCh38
rs376341807 Chr.5: 93741219-93741219 on Build GRCh38
rs747250845 Chr.5: 93741120-93741120 on Build GRCh38
rs765019645 Chr.5: 93741271-93741271 on Build GRCh38
rs763672762 Chr.5: 93740967-93740967 on Build GRCh38
rs760075591 Chr.5: 93741037-93741037 on Build GRCh38
rs530516757 Chr.5: 93741258-93741258 on Build GRCh38
rs749847930 Chr.5: 93741146-93741146 on Build GRCh38
rs373405434 Chr.5: 93741143-93741143 on Build GRCh38
rs926613223 Chr.5: 93740880-93740880 on Build GRCh38
rs866082084 Chr.5: 93741224-93741224 on Build GRCh38
rs867992633 Chr.5: 93741266-93741266 on Build GRCh38
rs760352735 Chr.5: 93741001-93741001 on Build GRCh38
rs768126763 Chr.5: 93741155-93741155 on Build GRCh38
rs763619426 Chr.5: 93741056-93741056 on Build GRCh38
rs757032596 Chr.5: 93740970-93740970 on Build GRCh38
rs781148855 Chr.5: 93740895-93740895 on Build GRCh38
rs779169489 Chr.5: 93740902-93740902 on Build GRCh38
rs373841980 Chr.5: 93741209-93741209 on Build GRCh38
rs554717601 Chr.5: 93740949-93740949 on Build GRCh38
rs750340501 Chr.5: 93740983-93740983 on Build GRCh38
rs755005919 Chr.5: 93741199-93741199 on Build GRCh38
rs190094264 Chr.5: 93741270-93741270 on Build GRCh38
rs752712796 Chr.5: 93741273-93741273 on Build GRCh38
rs775982611 Chr.5: 93741045-93741045 on Build GRCh38
rs767017668 Chr.5: 93741164-93741164 on Build GRCh38
rs1026902992 Chr.5: 93741251-93741251 on Build GRCh38
rs777147043 Chr.5: 93740912-93740912 on Build GRCh38
rs752263779 Chr.5: 93740944-93740944 on Build GRCh38
rs974618542 Chr.5: 93741139-93741139 on Build GRCh38
rs555552303 Chr.5: 93741074-93741074 on Build GRCh38
rs775400558 Chr.5: 93740986-93740986 on Build GRCh38
rs1056965537 Chr.5: 93740917-93740917 on Build GRCh38
rs773797061 Chr.5: 93741162-93741162 on Build GRCh38
rs778100007 Chr.5: 93741022-93741022 on Build GRCh38
rs543983400 Chr.5: 93741079-93741079 on Build GRCh38
rs939090026 Chr.5: 93741277-93741277 on Build GRCh38
rs867831676 Chr.5: 93741136-93741136 on Build GRCh38
rs780901290 Chr.5: 93740971-93740971 on Build GRCh38
rs762465212 Chr.5: 93740948-93740948 on Build GRCh38
rs766297322 Chr.5: 93741185-93741185 on Build GRCh38
rs911220828 Chr.5: 93741101-93741101 on Build GRCh38
rs760002979 Chr.5: 93740921-93740921 on Build GRCh38
rs771303188 Chr.5: 93741028-93741028 on Build GRCh38
rs754303500 Chr.5: 93740829-93740829 on Build GRCh38
rs528799392 Chr.5: 93741206-93741206 on Build GRCh38
rs746153932 Chr.5: 93741138-93741138 on Build GRCh38
rs769204489 Chr.5: 93741235-93741235 on Build GRCh38
rs373989380 Chr.5: 93741034-93741034 on Build GRCh38
rs749787710 Chr.5: 93741232-93741232 on Build GRCh38
rs867258564 Chr.5: 93741205-93741205 on Build GRCh38
rs577269513 Chr.5: 93741123-93741123 on Build GRCh38
rs753804275 Chr.5: 93741187-93741187 on Build GRCh38
rs756069005 Chr.5: 93740984-93740984 on Build GRCh38
rs764675526 Chr.5: 93740869-93740869 on Build GRCh38
rs746127873 Chr.5: 93741220-93741220 on Build GRCh38
rs377225323 Chr.5: 93740940-93740940 on Build GRCh38
rs147041924 Chr.5: 93741204-93741204 on Build GRCh38
rs774933866 Chr.5: 93741236-93741236 on Build GRCh38
rs938075584 Chr.5: 93740910-93740910 on Build GRCh38
rs550301121 Chr.5: 93741262-93741262 on Build GRCh38
rs994681346 Chr.5: 93741006-93741007 on Build GRCh38
rs747541526 Chr.5: 93741242-93741242 on Build GRCh38
rs185367860 Chr.5: 93741078-93741078 on Build GRCh38
rs757478491 Chr.5: 93741121-93741121 on Build GRCh38
rs751195358 Chr.5: 93741066-93741066 on Build GRCh38
rs753855484 Chr.5: 93741108-93741108 on Build GRCh38
rs751246695 Chr.5: 93740969-93740969 on Build GRCh38
rs761032270 Chr.5: 93740906-93740906 on Build GRCh38
rs756450444 Chr.5: 93741222-93741222 on Build GRCh38
rs369187302 Chr.5: 93741119-93741119 on Build GRCh38

View Full Product Details