Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95554993-95555503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSK
Entrez Gene ID
153642
Gene Name
arylsulfatase family member K
Gene Aliases
TSULF
UniGene
Hs.585051
Gene Chromosome Location
Chr.5: 95555108-95605102 on Build GRCh38
Transcripts
NM_198150.2 XM_005271904.3 NM_014639.3

Gene Symbol
TTC37
Entrez Gene ID
9652
Gene Name
tetratricopeptide repeat domain 37
Gene Aliases
KIAA0372, Ski3, THES
UniGene
Hs.482868
Gene Chromosome Location
Chr.5: 95463895-95555005 on Build GRCh38
Transcripts
NM_198150.2 XM_005271904.3 NM_014639.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6248688 c.9G>T p.L3L Chr.5: 95555287-95555287 on Build GRCh38
3776929 c.72G>A p.Q24Q Chr.5: 95555350-95555350 on Build GRCh38
6285080 c.93G>A p.A31A Chr.5: 95555371-95555371 on Build GRCh38
6285557 c.61G>A p.A21T Chr.5: 95555339-95555339 on Build GRCh38
6307899 c.53C>A p.A18D Chr.5: 95555331-95555331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751399694 Chr.5: 95555277-95555277 on Build GRCh38
rs757446679 Chr.5: 95555395-95555395 on Build GRCh38
rs888487902 Chr.5: 95555074-95555074 on Build GRCh38
rs768915188 Chr.5: 95555041-95555041 on Build GRCh38
rs910034041 Chr.5: 95555478-95555478 on Build GRCh38
rs988507007 Chr.5: 95555193-95555193 on Build GRCh38
rs750369903 Chr.5: 95555290-95555290 on Build GRCh38
rs559641162 Chr.5: 95555050-95555050 on Build GRCh38
rs886804529 Chr.5: 95555207-95555207 on Build GRCh38
rs992749231 Chr.5: 95555462-95555462 on Build GRCh38
rs780548129 Chr.5: 95555423-95555423 on Build GRCh38
rs897105632 Chr.5: 95555099-95555099 on Build GRCh38
rs965256903 Chr.5: 95555062-95555062 on Build GRCh38
rs770283155 Chr.5: 95555323-95555323 on Build GRCh38
rs368304272 Chr.5: 95555243-95555243 on Build GRCh38
rs948916973 Chr.5: 95555137-95555137 on Build GRCh38
rs1007096415 Chr.5: 95555124-95555124 on Build GRCh38
rs747827747 Chr.5: 95555453-95555453 on Build GRCh38
rs770518307 Chr.5: 95555461-95555461 on Build GRCh38
rs992885386 Chr.5: 95555121-95555121 on Build GRCh38
rs755977894 Chr.5: 95555292-95555292 on Build GRCh38
rs866509994 Chr.5: 95555091-95555091 on Build GRCh38
rs372655782 Chr.5: 95555444-95555444 on Build GRCh38
rs554154899 Chr.5: 95555479-95555479 on Build GRCh38
rs374986243 Chr.5: 95555384-95555384 on Build GRCh38
rs928171503 Chr.5: 95555061-95555061 on Build GRCh38
rs534526684 Chr.5: 95555151-95555151 on Build GRCh38
rs781431376 Chr.5: 95555396-95555396 on Build GRCh38
rs774524845 Chr.5: 95555364-95555364 on Build GRCh38
rs528260358 Chr.5: 95555070-95555070 on Build GRCh38
rs956057283 Chr.5: 95555457-95555457 on Build GRCh38
rs768433423 Chr.5: 95555341-95555341 on Build GRCh38
rs112683194 Chr.5: 95555251-95555251 on Build GRCh38
rs773980651 Chr.5: 95555450-95555450 on Build GRCh38
rs182161531 Chr.5: 95555083-95555083 on Build GRCh38
rs765256660 Chr.5: 95555298-95555298 on Build GRCh38
rs971058403 Chr.5: 95555434-95555434 on Build GRCh38
rs912885665 Chr.5: 95555194-95555194 on Build GRCh38
rs1047988472 Chr.5: 95555201-95555201 on Build GRCh38
rs982397051 Chr.5: 95555238-95555238 on Build GRCh38
rs767419361 Chr.5: 95555288-95555288 on Build GRCh38
rs747820910 Chr.5: 95555187-95555187 on Build GRCh38
rs760563103 Chr.5: 95555370-95555370 on Build GRCh38
rs757684496 Chr.5: 95555316-95555316 on Build GRCh38
rs17552017 Chr.5: 95555115-95555115 on Build GRCh38
rs112432496 Chr.5: 95555429-95555429 on Build GRCh38
rs998039342 Chr.5: 95555113-95555113 on Build GRCh38
rs111947730 Chr.5: 95555096-95555096 on Build GRCh38
rs752732890 Chr.5: 95555376-95555376 on Build GRCh38
rs957133720 Chr.5: 95555409-95555409 on Build GRCh38
rs941435365 Chr.5: 95555318-95555319 on Build GRCh38
rs1031519893 Chr.5: 95555452-95555452 on Build GRCh38
rs917298206 Chr.5: 95555126-95555126 on Build GRCh38
rs557080694 Chr.5: 95555464-95555464 on Build GRCh38
rs939640557 Chr.5: 95555226-95555226 on Build GRCh38
rs370861379 Chr.5: 95555248-95555248 on Build GRCh38
rs748760776 Chr.5: 95555335-95555335 on Build GRCh38
rs113539792 Chr.5: 95555114-95555114 on Build GRCh38
rs1055221514 Chr.5: 95555072-95555072 on Build GRCh38
rs763797717 Chr.5: 95555274-95555274 on Build GRCh38
rs537100451 Chr.5: 95555381-95555381 on Build GRCh38
rs777914445 Chr.5: 95555306-95555306 on Build GRCh38
rs980640323 Chr.5: 95555147-95555147 on Build GRCh38
rs978642540 Chr.5: 95555106-95555106 on Build GRCh38
rs957104089 Chr.5: 95555183-95555183 on Build GRCh38
rs920520458 Chr.5: 95555031-95555031 on Build GRCh38
rs749741870 Chr.5: 95555443-95555443 on Build GRCh38
rs778143666 Chr.5: 95555242-95555242 on Build GRCh38
rs762532926 Chr.5: 95555268-95555268 on Build GRCh38
rs138271997 Chr.5: 95555295-95555295 on Build GRCh38
rs61733082 Chr.5: 95555355-95555355 on Build GRCh38
rs550701683 Chr.5: 95555157-95555157 on Build GRCh38
rs751819599 Chr.5: 95555389-95555389 on Build GRCh38
rs149648302 Chr.5: 95555334-95555334 on Build GRCh38
rs1000093461 Chr.5: 95555405-95555405 on Build GRCh38
rs533002261 Chr.5: 95555181-95555181 on Build GRCh38
rs568536380 Chr.5: 95555080-95555080 on Build GRCh38
rs545690642 Chr.5: 95555019-95555019 on Build GRCh38
rs956026534 Chr.5: 95555116-95555116 on Build GRCh38
rs774891593 Chr.5: 95555327-95555327 on Build GRCh38
rs769225750 Chr.5: 95555252-95555252 on Build GRCh38
rs771442716 Chr.5: 95555247-95555247 on Build GRCh38
rs570412518 Chr.5: 95555178-95555178 on Build GRCh38
rs746201423 Chr.5: 95555421-95555421 on Build GRCh38
rs769555617 Chr.5: 95555291-95555291 on Build GRCh38
rs150467708 Chr.5: 95555284-95555284 on Build GRCh38
rs761530133 Chr.5: 95555354-95555354 on Build GRCh38
rs781502727 Chr.5: 95555317-95555317 on Build GRCh38
rs111345636 Chr.5: 95555472-95555472 on Build GRCh38
rs552893251 Chr.5: 95555240-95555240 on Build GRCh38
rs368737426 Chr.5: 95555422-95555422 on Build GRCh38
rs775042221 Chr.5: 95555259-95555259 on Build GRCh38
rs368063444 Chr.5: 95555309-95555309 on Build GRCh38
rs371671370 Chr.5: 95555342-95555342 on Build GRCh38
rs904354882 Chr.5: 95555401-95555401 on Build GRCh38
rs201158981 Chr.5: 95555331-95555331 on Build GRCh38
rs758554347 Chr.5: 95555304-95555304 on Build GRCh38
rs773014351 Chr.5: 95555363-95555363 on Build GRCh38
rs746409212 Chr.5: 95555319-95555319 on Build GRCh38

View Full Product Details