Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 96390230-96390779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101929710
Entrez Gene ID
101929710
Gene Name
uncharacterized LOC101929710
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.5: 95962001-96631085 on Build GRCh38
Transcripts
NR_130776.1 NM_000439.4 NM_001177875.1

Gene Symbol
PCSK1
Entrez Gene ID
5122
Gene Name
proprotein convertase subtilisin/kexin type 1
Gene Aliases
BMIQ12, NEC1, PC1, PC3, SPC3
UniGene
Hs.78977
Gene Chromosome Location
Chr.5: 96390336-96433281 on Build GRCh38
Transcripts
NR_130776.1 NM_000439.4 NM_001177875.1

SNP Detail

SNP ID Chromosome Location
rs959133473 Chr.5: 96390678-96390678 on Build GRCh38
rs963896306 Chr.5: 96390514-96390514 on Build GRCh38
rs967513865 Chr.5: 96390353-96390355 on Build GRCh38
rs754474461 Chr.5: 96390411-96390411 on Build GRCh38
rs984935863 Chr.5: 96390713-96390713 on Build GRCh38
rs940688879 Chr.5: 96390295-96390295 on Build GRCh38
rs1011379880 Chr.5: 96390335-96390335 on Build GRCh38
rs1050484297 Chr.5: 96390367-96390367 on Build GRCh38
rs575092021 Chr.5: 96390349-96390349 on Build GRCh38
rs746472388 Chr.5: 96390390-96390390 on Build GRCh38
rs942309623 Chr.5: 96390609-96390609 on Build GRCh38
rs1036402046 Chr.5: 96390312-96390312 on Build GRCh38
rs187137855 Chr.5: 96390431-96390431 on Build GRCh38
rs893015328 Chr.5: 96390446-96390446 on Build GRCh38
rs1039656759 Chr.5: 96390688-96390688 on Build GRCh38
rs1006696656 Chr.5: 96390415-96390415 on Build GRCh38
rs556231424 Chr.5: 96390611-96390611 on Build GRCh38
rs367854792 Chr.5: 96390608-96390608 on Build GRCh38
rs1019936726 Chr.5: 96390576-96390576 on Build GRCh38
rs1010568177 Chr.5: 96390478-96390478 on Build GRCh38
rs527453582 Chr.5: 96390263-96390263 on Build GRCh38
rs537724412 Chr.5: 96390594-96390594 on Build GRCh38
rs909429923 Chr.5: 96390280-96390280 on Build GRCh38
rs879943012 Chr.5: 96390596-96390596 on Build GRCh38
rs919406892 Chr.5: 96390570-96390570 on Build GRCh38
rs923260455 Chr.5: 96390336-96390336 on Build GRCh38
rs1034031341 Chr.5: 96390653-96390653 on Build GRCh38
rs570346851 Chr.5: 96390435-96390435 on Build GRCh38
rs892968992 Chr.5: 96390321-96390321 on Build GRCh38
rs909436389 Chr.5: 96390467-96390467 on Build GRCh38
rs886060874 Chr.5: 96390713-96390713 on Build GRCh38
rs972280403 Chr.5: 96390516-96390516 on Build GRCh38

View Full Product Details