Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 133956508-133957018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf15
Entrez Gene ID
56951
Gene Name
chromosome 5 open reading frame 15
Gene Aliases
HTGN29, KCT2
UniGene
Hs.744034
Gene Chromosome Location
Chr.5: 133955507-133968715 on Build GRCh38
Transcripts
NM_020199.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5729992 c.733C>T p.R245C Chr.5: 133956924-133956924 on Build GRCh38
1060834 c.704G>A p.G235D Chr.5: 133956953-133956953 on Build GRCh38
4667799 c.773A>T p.K258M Chr.5: 133956884-133956884 on Build GRCh38
3850117 c.732T>C p.H244H Chr.5: 133956925-133956925 on Build GRCh38
4667800 c.727T>C p.Y243H Chr.5: 133956930-133956930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187246788 Chr.5: 133956953-133956953 on Build GRCh38
rs530633222 Chr.5: 133956651-133956651 on Build GRCh38
rs546904041 Chr.5: 133956711-133956711 on Build GRCh38
rs1037969830 Chr.5: 133956653-133956653 on Build GRCh38
rs11540541 Chr.5: 133956712-133956712 on Build GRCh38
rs192785651 Chr.5: 133956707-133956707 on Build GRCh38
rs746656501 Chr.5: 133956809-133956809 on Build GRCh38
rs541713224 Chr.5: 133956810-133956810 on Build GRCh38
rs753942566 Chr.5: 133956898-133956898 on Build GRCh38
rs750589127 Chr.5: 133956878-133956878 on Build GRCh38
rs564326181 Chr.5: 133956709-133956709 on Build GRCh38
rs762294013 Chr.5: 133956959-133956959 on Build GRCh38
rs568340511 Chr.5: 133956774-133956774 on Build GRCh38
rs775052686 Chr.5: 133956946-133956946 on Build GRCh38
rs766514661 Chr.5: 133956897-133956897 on Build GRCh38
rs139570849 Chr.5: 133956931-133956931 on Build GRCh38
rs146335440 Chr.5: 133956875-133956875 on Build GRCh38
rs752638440 Chr.5: 133956956-133956956 on Build GRCh38
rs61730680 Chr.5: 133956955-133956955 on Build GRCh38
rs767784325 Chr.5: 133956867-133956867 on Build GRCh38
rs374778255 Chr.5: 133956815-133956815 on Build GRCh38
rs184172978 Chr.5: 133956710-133956710 on Build GRCh38
rs372276960 Chr.5: 133956851-133956851 on Build GRCh38
rs755153221 Chr.5: 133956911-133956911 on Build GRCh38
rs760835895 Chr.5: 133956890-133956890 on Build GRCh38
rs923503504 Chr.5: 133956924-133956924 on Build GRCh38
rs776150380 Chr.5: 133956819-133956819 on Build GRCh38
rs964797466 Chr.5: 133956846-133956846 on Build GRCh38
rs774904834 Chr.5: 133956840-133956840 on Build GRCh38
rs933621892 Chr.5: 133956551-133956551 on Build GRCh38
rs1002815746 Chr.5: 133956758-133956758 on Build GRCh38
rs898254203 Chr.5: 133956662-133956662 on Build GRCh38
rs566618405 Chr.5: 133956532-133956532 on Build GRCh38
rs368939447 Chr.5: 133956831-133956831 on Build GRCh38
rs923398704 Chr.5: 133956533-133956533 on Build GRCh38
rs933464035 Chr.5: 133956948-133956948 on Build GRCh38
rs778993410 Chr.5: 133956923-133956923 on Build GRCh38
rs1048445092 Chr.5: 133956591-133956591 on Build GRCh38
rs1013049048 Chr.5: 133956683-133956683 on Build GRCh38
rs563416255 Chr.5: 133956610-133956610 on Build GRCh38
rs777436502 Chr.5: 133956989-133956989 on Build GRCh38
rs886796956 Chr.5: 133956595-133956595 on Build GRCh38

View Full Product Details