Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140568058-140568553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35A4
Entrez Gene ID
113829
Gene Name
solute carrier family 35 member A4
Gene Aliases
-
UniGene
Hs.406840
Gene Chromosome Location
Chr.5: 140564565-140569104 on Build GRCh38
Transcripts
NM_080670.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1061720 c.962A>G p.Y321C Chr.5: 140568131-140568131 on Build GRCh38
4141446 c.898C>A p.L300M Chr.5: 140568067-140568067 on Build GRCh38
1061719 c.925G>A p.A309T Chr.5: 140568094-140568094 on Build GRCh38
5476557 c.896G>A p.R299Q Chr.5: 140568065-140568065 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754639645 Chr.5: 140568110-140568110 on Build GRCh38
rs930374512 Chr.5: 140568409-140568409 on Build GRCh38
rs779614603 Chr.5: 140568105-140568105 on Build GRCh38
rs1010676466 Chr.5: 140568477-140568477 on Build GRCh38
rs904382952 Chr.5: 140568324-140568324 on Build GRCh38
rs755889495 Chr.5: 140568190-140568190 on Build GRCh38
rs747162805 Chr.5: 140568343-140568343 on Build GRCh38
rs1027145334 Chr.5: 140568143-140568143 on Build GRCh38
rs750964666 Chr.5: 140568174-140568174 on Build GRCh38
rs11551314 Chr.5: 140568417-140568417 on Build GRCh38
rs775600465 Chr.5: 140568246-140568246 on Build GRCh38
rs181678496 Chr.5: 140568450-140568450 on Build GRCh38
rs1036527221 Chr.5: 140568120-140568120 on Build GRCh38
rs142973260 Chr.5: 140568081-140568081 on Build GRCh38
rs142141597 Chr.5: 140568447-140568447 on Build GRCh38
rs74950706 Chr.5: 140568099-140568099 on Build GRCh38
rs906034812 Chr.5: 140568435-140568435 on Build GRCh38
rs76819422 Chr.5: 140568373-140568373 on Build GRCh38
rs900931538 Chr.5: 140568230-140568230 on Build GRCh38
rs777680140 Chr.5: 140568203-140568203 on Build GRCh38
rs921759027 Chr.5: 140568317-140568317 on Build GRCh38
rs779180626 Chr.5: 140568187-140568187 on Build GRCh38
rs769647639 Chr.5: 140568119-140568119 on Build GRCh38
rs536561312 Chr.5: 140568445-140568445 on Build GRCh38
rs772233745 Chr.5: 140568202-140568202 on Build GRCh38
rs771413609 Chr.5: 140568210-140568211 on Build GRCh38
rs367666945 Chr.5: 140568114-140568114 on Build GRCh38
rs759483807 Chr.5: 140568135-140568135 on Build GRCh38
rs749083873 Chr.5: 140568206-140568206 on Build GRCh38
rs774466380 Chr.5: 140568134-140568134 on Build GRCh38
rs201059756 Chr.5: 140568131-140568131 on Build GRCh38
rs980178500 Chr.5: 140568480-140568480 on Build GRCh38
rs896046197 Chr.5: 140568452-140568452 on Build GRCh38
rs542018886 Chr.5: 140568169-140568169 on Build GRCh38
rs1027682033 Chr.5: 140568496-140568496 on Build GRCh38
rs754285017 Chr.5: 140568157-140568157 on Build GRCh38
rs911777616 Chr.5: 140568355-140568355 on Build GRCh38
rs201868697 Chr.5: 140568162-140568162 on Build GRCh38
rs78905299 Chr.5: 140568361-140568361 on Build GRCh38
rs757767027 Chr.5: 140568102-140568102 on Build GRCh38
rs747231745 Chr.5: 140568256-140568256 on Build GRCh38
rs760984896 Chr.5: 140568083-140568083 on Build GRCh38
rs762354256 Chr.5: 140568170-140568170 on Build GRCh38
rs745819372 Chr.5: 140568228-140568228 on Build GRCh38
rs747835752 Chr.5: 140568118-140568118 on Build GRCh38
rs759771359 Chr.5: 140568077-140568077 on Build GRCh38
rs993212240 Chr.5: 140568097-140568097 on Build GRCh38
rs775619051 Chr.5: 140568140-140568140 on Build GRCh38
rs760896865 Chr.5: 140568141-140568141 on Build GRCh38
rs539842799 Chr.5: 140568122-140568122 on Build GRCh38
rs1048634435 Chr.5: 140568421-140568421 on Build GRCh38
rs746101565 Chr.5: 140568189-140568189 on Build GRCh38
rs199583070 Chr.5: 140568139-140568139 on Build GRCh38
rs530999822 Chr.5: 140568225-140568225 on Build GRCh38
rs369732831 Chr.5: 140568382-140568382 on Build GRCh38
rs931939480 Chr.5: 140568332-140568332 on Build GRCh38
rs777725051 Chr.5: 140568121-140568121 on Build GRCh38
rs765716086 Chr.5: 140568171-140568171 on Build GRCh38
rs546308141 Chr.5: 140568301-140568301 on Build GRCh38
rs776961612 Chr.5: 140568274-140568274 on Build GRCh38
rs147904672 Chr.5: 140568337-140568337 on Build GRCh38
rs767141450 Chr.5: 140568188-140568188 on Build GRCh38
rs771984896 Chr.5: 140568229-140568229 on Build GRCh38
rs956096039 Chr.5: 140568085-140568085 on Build GRCh38
rs764425197 Chr.5: 140568150-140568150 on Build GRCh38
rs754281959 Chr.5: 140568101-140568101 on Build GRCh38
rs757096461 Chr.5: 140568222-140568222 on Build GRCh38
rs1049504456 Chr.5: 140568315-140568315 on Build GRCh38
rs866665504 Chr.5: 140568115-140568115 on Build GRCh38
rs780138579 Chr.5: 140568266-140568266 on Build GRCh38
rs762036614 Chr.5: 140568285-140568285 on Build GRCh38
rs557551479 Chr.5: 140568082-140568082 on Build GRCh38
rs751073961 Chr.5: 140568109-140568109 on Build GRCh38
rs968533809 Chr.5: 140568080-140568080 on Build GRCh38
rs368323800 Chr.5: 140568184-140568184 on Build GRCh38

View Full Product Details