Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140647720-140648155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IK
Entrez Gene ID
3550
Gene Name
IK cytokine, down-regulator of HLA II
Gene Aliases
CSA2, RED, RER
UniGene
Hs.421245
Gene Chromosome Location
Chr.5: 140647799-140662480 on Build GRCh38
Transcripts
NM_006083.3 NR_037428.1 NM_001185012.1 NM_002488.4 NR_033697.1

Gene Symbol
MIR3655
Entrez Gene ID
100500820
Gene Name
microRNA 3655
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.5: 140647844-140647926 on Build GRCh38
Transcripts
NM_006083.3 NR_037428.1 NM_001185012.1 NM_002488.4 NR_033697.1

Gene Symbol
NDUFA2
Entrez Gene ID
4695
Gene Name
NADH:ubiquinone oxidoreductase subunit A2
Gene Aliases
B8, CD14, CIB8
UniGene
Hs.75914
Gene Chromosome Location
Chr.5: 140645363-140647785 on Build GRCh38
Transcripts
NM_006083.3 NR_037428.1 NM_001185012.1 NM_002488.4 NR_033697.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5000544 c.5C>T p.P2L Chr.5: 140647913-140647913 on Build GRCh38
6233399 c.10C>A p.R4R Chr.5: 140647918-140647918 on Build GRCh38
3826925 c.16+9C>T p.? Chr.5: 140647933-140647933 on Build GRCh38
3239031 c.8A>T p.E3V Chr.5: 140647916-140647916 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1056409237 Chr.5: 140648068-140648068 on Build GRCh38
rs1009713574 Chr.5: 140647993-140647993 on Build GRCh38
rs202067959 Chr.5: 140648046-140648051 on Build GRCh38
rs2245861 Chr.5: 140648050-140648050 on Build GRCh38
rs374534339 Chr.5: 140647899-140647899 on Build GRCh38
rs940767479 Chr.5: 140647886-140647886 on Build GRCh38
rs761616160 Chr.5: 140647778-140647778 on Build GRCh38
rs779534000 Chr.5: 140647835-140647835 on Build GRCh38
rs879204870 Chr.5: 140648043-140648050 on Build GRCh38
rs926679030 Chr.5: 140648052-140648052 on Build GRCh38
rs1027331724 Chr.5: 140648130-140648130 on Build GRCh38
rs775426500 Chr.5: 140647957-140647957 on Build GRCh38
rs759580754 Chr.5: 140647801-140647801 on Build GRCh38
rs748188529 Chr.5: 140647930-140647930 on Build GRCh38
rs979841264 Chr.5: 140648085-140648085 on Build GRCh38
rs975069545 Chr.5: 140648007-140648007 on Build GRCh38
rs758857731 Chr.5: 140648113-140648113 on Build GRCh38
rs747265872 Chr.5: 140647855-140647855 on Build GRCh38
rs60377719 Chr.5: 140648044-140648053 on Build GRCh38
rs747859371 Chr.5: 140647822-140647822 on Build GRCh38
rs535080968 Chr.5: 140647875-140647875 on Build GRCh38
rs967660470 Chr.5: 140648003-140648003 on Build GRCh38
rs756466349 Chr.5: 140647741-140647741 on Build GRCh38
rs545685884 Chr.5: 140648067-140648067 on Build GRCh38
rs746448356 Chr.5: 140647836-140647836 on Build GRCh38
rs752956448 Chr.5: 140647740-140647740 on Build GRCh38
rs370832670 Chr.5: 140647947-140647947 on Build GRCh38
rs1022031364 Chr.5: 140647995-140647995 on Build GRCh38
rs750777592 Chr.5: 140648112-140648112 on Build GRCh38
rs778058720 Chr.5: 140647828-140647828 on Build GRCh38
rs760643148 Chr.5: 140647961-140647961 on Build GRCh38
rs368962559 Chr.5: 140647838-140647838 on Build GRCh38
rs776731402 Chr.5: 140648008-140648019 on Build GRCh38
rs939263716 Chr.5: 140648064-140648064 on Build GRCh38
rs201961300 Chr.5: 140647891-140647891 on Build GRCh38
rs755697876 Chr.5: 140647914-140647914 on Build GRCh38
rs779484520 Chr.5: 140648042-140648049 on Build GRCh38
rs892339213 Chr.5: 140647950-140647950 on Build GRCh38
rs564268917 Chr.5: 140648107-140648107 on Build GRCh38
rs371585072 Chr.5: 140647887-140647887 on Build GRCh38
rs762057029 Chr.5: 140648019-140648020 on Build GRCh38
rs575180800 Chr.5: 140648044-140648044 on Build GRCh38
rs777258491 Chr.5: 140647920-140647920 on Build GRCh38
rs769500961 Chr.5: 140647791-140647791 on Build GRCh38
rs775477746 Chr.5: 140647874-140647874 on Build GRCh38
rs553111700 Chr.5: 140647890-140647890 on Build GRCh38
rs753149634 Chr.5: 140648043-140648054 on Build GRCh38
rs1013954112 Chr.5: 140648029-140648050 on Build GRCh38
rs780739269 Chr.5: 140647905-140647905 on Build GRCh38
rs766560115 Chr.5: 140648041-140648050 on Build GRCh38
rs377685148 Chr.5: 140647937-140647937 on Build GRCh38
rs987957327 Chr.5: 140648040-140648040 on Build GRCh38
rs752899359 Chr.5: 140647814-140647814 on Build GRCh38
rs556880603 Chr.5: 140647998-140647998 on Build GRCh38
rs967712893 Chr.5: 140647808-140647808 on Build GRCh38
rs746497677 Chr.5: 140647756-140647756 on Build GRCh38
rs367602188 Chr.5: 140648045-140648050 on Build GRCh38
rs769477072 Chr.5: 140648039-140648050 on Build GRCh38
rs768325530 Chr.5: 140647770-140647770 on Build GRCh38
rs776450501 Chr.5: 140648007-140648008 on Build GRCh38
rs987777919 Chr.5: 140648027-140648027 on Build GRCh38
rs953748648 Chr.5: 140648015-140648015 on Build GRCh38
rs766177915 Chr.5: 140647796-140647796 on Build GRCh38
rs769446600 Chr.5: 140648047-140648054 on Build GRCh38
rs766971884 Chr.5: 140648117-140648117 on Build GRCh38
rs572868422 Chr.5: 140648122-140648122 on Build GRCh38
rs1000065251 Chr.5: 140648011-140648011 on Build GRCh38
rs905111941 Chr.5: 140647924-140647924 on Build GRCh38
rs777399295 Chr.5: 140648125-140648125 on Build GRCh38
rs377007416 Chr.5: 140647871-140647871 on Build GRCh38
rs375301523 Chr.5: 140647978-140647978 on Build GRCh38
rs70988770 Chr.5: 140648045-140648046 on Build GRCh38
rs780978888 Chr.5: 140648041-140648042 on Build GRCh38
rs1043807689 Chr.5: 140648005-140648006 on Build GRCh38
rs754133084 Chr.5: 140647878-140647878 on Build GRCh38
rs747563860 Chr.5: 140647910-140647910 on Build GRCh38
rs185280379 Chr.5: 140647831-140647831 on Build GRCh38
rs896728339 Chr.5: 140648019-140648050 on Build GRCh38
rs139482464 Chr.5: 140647863-140647863 on Build GRCh38
rs867446887 Chr.5: 140647781-140647781 on Build GRCh38
rs754227476 Chr.5: 140648045-140648054 on Build GRCh38
rs767649892 Chr.5: 140647805-140647805 on Build GRCh38
rs755574127 Chr.5: 140648123-140648123 on Build GRCh38
rs146400503 Chr.5: 140647850-140647850 on Build GRCh38
rs771273606 Chr.5: 140647753-140647753 on Build GRCh38
rs932034802 Chr.5: 140648002-140648002 on Build GRCh38
rs992864035 Chr.5: 140647832-140647832 on Build GRCh38
rs111226907 Chr.5: 140647839-140647839 on Build GRCh38
rs969805512 Chr.5: 140648055-140648055 on Build GRCh38
rs940896389 Chr.5: 140648032-140648032 on Build GRCh38
rs770798087 Chr.5: 140647861-140647861 on Build GRCh38
rs774071371 Chr.5: 140647931-140647931 on Build GRCh38
rs751545213 Chr.5: 140647800-140647800 on Build GRCh38
rs764303843 Chr.5: 140647971-140647971 on Build GRCh38
rs778054089 Chr.5: 140647752-140647752 on Build GRCh38
rs879137324 Chr.5: 140648042-140648043 on Build GRCh38
rs776708427 Chr.5: 140648105-140648105 on Build GRCh38
rs749013145 Chr.5: 140647857-140647857 on Build GRCh38
rs912028572 Chr.5: 140648030-140648030 on Build GRCh38
rs1045097817 Chr.5: 140648042-140648042 on Build GRCh38
rs2569161 Chr.5: 140648046-140648046 on Build GRCh38
rs900623800 Chr.5: 140647959-140647959 on Build GRCh38
rs1029164158 Chr.5: 140648009-140648009 on Build GRCh38
rs759527321 Chr.5: 140647866-140647866 on Build GRCh38
rs539275827 Chr.5: 140647898-140647898 on Build GRCh38
rs987943600 Chr.5: 140647758-140647758 on Build GRCh38
rs530439525 Chr.5: 140647772-140647772 on Build GRCh38
rs747666614 Chr.5: 140647843-140647843 on Build GRCh38
rs746278030 Chr.5: 140647795-140647795 on Build GRCh38
rs113236159 Chr.5: 140647751-140647751 on Build GRCh38
rs866714168 Chr.5: 140648038-140648038 on Build GRCh38
rs772818096 Chr.5: 140647793-140647793 on Build GRCh38
rs1023910818 Chr.5: 140648033-140648050 on Build GRCh38
rs768635352 Chr.5: 140647959-140647967 on Build GRCh38
rs756228978 Chr.5: 140647820-140647820 on Build GRCh38
rs999798095 Chr.5: 140647754-140647754 on Build GRCh38
rs535439456 Chr.5: 140647984-140647984 on Build GRCh38
rs972113079 Chr.5: 140648036-140648036 on Build GRCh38
rs749036471 Chr.5: 140647926-140647926 on Build GRCh38
rs748761559 Chr.5: 140648128-140648128 on Build GRCh38

View Full Product Details