Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140795589-140796094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHA1
Entrez Gene ID
56147
Gene Name
protocadherin alpha 1
Gene Aliases
PCDH-ALPHA1
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140786136-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_031495.1 NM_031496.1

Gene Symbol
PCDHA2
Entrez Gene ID
56146
Gene Name
protocadherin alpha 2
Gene Aliases
PCDH-ALPHA2
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140794859-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_031495.1 NM_031496.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5008076 c.860C>G p.S287C Chr.5: 140795824-140795824 on Build GRCh38
3609522 c.1129C>T p.R377C Chr.5: 140796093-140796093 on Build GRCh38
4796784 c.693G>A p.K231K Chr.5: 140795657-140795657 on Build GRCh38
3239458 c.877T>C p.F293L Chr.5: 140795841-140795841 on Build GRCh38
1061829 c.951G>T p.E317D Chr.5: 140795915-140795915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782787763 Chr.5: 140795881-140795881 on Build GRCh38
rs782205660 Chr.5: 140795916-140795916 on Build GRCh38
rs782399257 Chr.5: 140795657-140795657 on Build GRCh38
rs368109410 Chr.5: 140795872-140795872 on Build GRCh38
rs782408299 Chr.5: 140795704-140795704 on Build GRCh38
rs192661342 Chr.5: 140795893-140795893 on Build GRCh38
rs782144250 Chr.5: 140795760-140795760 on Build GRCh38
rs386352344 Chr.5: 140795974-140795974 on Build GRCh38
rs782660322 Chr.5: 140795720-140795720 on Build GRCh38
rs945777469 Chr.5: 140796057-140796057 on Build GRCh38
rs533295201 Chr.5: 140795986-140795986 on Build GRCh38
rs17844246 Chr.5: 140795913-140795913 on Build GRCh38
rs138543529 Chr.5: 140795912-140795912 on Build GRCh38
rs927272442 Chr.5: 140795987-140795987 on Build GRCh38
rs782245686 Chr.5: 140795843-140795843 on Build GRCh38
rs562047607 Chr.5: 140795862-140795862 on Build GRCh38
rs782718867 Chr.5: 140795898-140795898 on Build GRCh38
rs782795009 Chr.5: 140795798-140795798 on Build GRCh38
rs895598108 Chr.5: 140795682-140795682 on Build GRCh38
rs782391222 Chr.5: 140796070-140796070 on Build GRCh38
rs782170874 Chr.5: 140795715-140795715 on Build GRCh38
rs782727470 Chr.5: 140795963-140795963 on Build GRCh38
rs150248027 Chr.5: 140795759-140795759 on Build GRCh38
rs781812595 Chr.5: 140795668-140795668 on Build GRCh38
rs781846968 Chr.5: 140795889-140795889 on Build GRCh38
rs372029609 Chr.5: 140795927-140795927 on Build GRCh38
rs782311905 Chr.5: 140795751-140795751 on Build GRCh38
rs782729838 Chr.5: 140795789-140795789 on Build GRCh38
rs372524197 Chr.5: 140795840-140795840 on Build GRCh38
rs145876467 Chr.5: 140795910-140795910 on Build GRCh38
rs781883265 Chr.5: 140795804-140795804 on Build GRCh38
rs782198727 Chr.5: 140796062-140796062 on Build GRCh38
rs782748843 Chr.5: 140795666-140795666 on Build GRCh38
rs782522858 Chr.5: 140795693-140795693 on Build GRCh38
rs782551806 Chr.5: 140796027-140796027 on Build GRCh38
rs781944142 Chr.5: 140795977-140795977 on Build GRCh38
rs369994142 Chr.5: 140795690-140795690 on Build GRCh38
rs73791786 Chr.5: 140795876-140795876 on Build GRCh38
rs782443801 Chr.5: 140795795-140795795 on Build GRCh38
rs782292458 Chr.5: 140795834-140795834 on Build GRCh38
rs368563521 Chr.5: 140795725-140795725 on Build GRCh38
rs782070624 Chr.5: 140795748-140795748 on Build GRCh38
rs937213419 Chr.5: 140795650-140795650 on Build GRCh38
rs529364047 Chr.5: 140795864-140795864 on Build GRCh38
rs781957318 Chr.5: 140795733-140795733 on Build GRCh38
rs782606233 Chr.5: 140795615-140795615 on Build GRCh38
rs782218525 Chr.5: 140795812-140795812 on Build GRCh38
rs781857494 Chr.5: 140795779-140795779 on Build GRCh38
rs782575162 Chr.5: 140795826-140795826 on Build GRCh38
rs375544937 Chr.5: 140795717-140795717 on Build GRCh38
rs199883861 Chr.5: 140795794-140795794 on Build GRCh38
rs201828641 Chr.5: 140795841-140795841 on Build GRCh38
rs782580934 Chr.5: 140795817-140795818 on Build GRCh38
rs782460542 Chr.5: 140795825-140795825 on Build GRCh38
rs781990083 Chr.5: 140795661-140795661 on Build GRCh38
rs781888206 Chr.5: 140795692-140795692 on Build GRCh38
rs782461058 Chr.5: 140795710-140795710 on Build GRCh38
rs1057279342 Chr.5: 140795653-140795653 on Build GRCh38
rs782038098 Chr.5: 140796064-140796064 on Build GRCh38
rs782097002 Chr.5: 140795783-140795783 on Build GRCh38
rs140965227 Chr.5: 140795680-140795680 on Build GRCh38
rs782366591 Chr.5: 140795729-140795729 on Build GRCh38
rs782417894 Chr.5: 140795837-140795837 on Build GRCh38
rs200785856 Chr.5: 140796038-140796038 on Build GRCh38
rs782378293 Chr.5: 140795614-140795614 on Build GRCh38
rs536063756 Chr.5: 140795664-140795664 on Build GRCh38
rs781955291 Chr.5: 140795709-140795709 on Build GRCh38
rs952716239 Chr.5: 140795954-140795954 on Build GRCh38
rs36007004 Chr.5: 140796057-140796058 on Build GRCh38
rs781838822 Chr.5: 140796005-140796005 on Build GRCh38
rs782456180 Chr.5: 140795711-140795711 on Build GRCh38
rs782547899 Chr.5: 140795881-140795882 on Build GRCh38
rs368808250 Chr.5: 140795957-140795957 on Build GRCh38
rs781789373 Chr.5: 140796023-140796023 on Build GRCh38
rs376704812 Chr.5: 140795937-140795937 on Build GRCh38
rs150530045 Chr.5: 140795648-140795648 on Build GRCh38
rs889935841 Chr.5: 140795848-140795848 on Build GRCh38
rs782626454 Chr.5: 140795808-140795808 on Build GRCh38
rs781892845 Chr.5: 140795999-140795999 on Build GRCh38
rs782060780 Chr.5: 140795982-140795982 on Build GRCh38
rs992640978 Chr.5: 140796019-140796019 on Build GRCh38
rs200424378 Chr.5: 140795944-140795944 on Build GRCh38
rs141546686 Chr.5: 140795945-140795945 on Build GRCh38
rs781891525 Chr.5: 140795856-140795856 on Build GRCh38
rs182370314 Chr.5: 140795754-140795754 on Build GRCh38
rs782806378 Chr.5: 140795691-140795691 on Build GRCh38
rs782136038 Chr.5: 140795880-140795880 on Build GRCh38
rs782621383 Chr.5: 140796051-140796051 on Build GRCh38
rs782329141 Chr.5: 140795918-140795918 on Build GRCh38
rs73278777 Chr.5: 140795714-140795714 on Build GRCh38
rs965421102 Chr.5: 140795917-140795917 on Build GRCh38
rs782631973 Chr.5: 140795695-140795695 on Build GRCh38
rs374222606 Chr.5: 140795849-140795849 on Build GRCh38
rs138887896 Chr.5: 140795765-140795765 on Build GRCh38
rs375678815 Chr.5: 140795639-140795639 on Build GRCh38
rs782659941 Chr.5: 140796048-140796048 on Build GRCh38
rs782135378 Chr.5: 140796003-140796003 on Build GRCh38
rs781797873 Chr.5: 140795899-140795899 on Build GRCh38
rs1032626475 Chr.5: 140795835-140795835 on Build GRCh38
rs781982977 Chr.5: 140796073-140796073 on Build GRCh38
rs782572566 Chr.5: 140796018-140796018 on Build GRCh38
rs782767651 Chr.5: 140796004-140796004 on Build GRCh38
rs782208465 Chr.5: 140795635-140795635 on Build GRCh38
rs782315024 Chr.5: 140796063-140796063 on Build GRCh38
rs781839041 Chr.5: 140795705-140795705 on Build GRCh38
rs782477722 Chr.5: 140796008-140796008 on Build GRCh38

View Full Product Details