Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140828302-140828799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHA1
Entrez Gene ID
56147
Gene Name
protocadherin alpha 1
Gene Aliases
PCDH-ALPHA1
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140786136-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031848.2 NM_031849.2

Gene Symbol
PCDHA2
Entrez Gene ID
56146
Gene Name
protocadherin alpha 2
Gene Aliases
PCDH-ALPHA2
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140794859-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031848.2 NM_031849.2

Gene Symbol
PCDHA3
Entrez Gene ID
56145
Gene Name
protocadherin alpha 3
Gene Aliases
PCDH-ALPHA3
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140801198-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031848.2 NM_031849.2

Gene Symbol
PCDHA4
Entrez Gene ID
56144
Gene Name
protocadherin alpha 4
Gene Aliases
CNR1, CNRN1, CRNR1, PCDH-ALPHA4
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140807074-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031848.2 NM_031849.2

Gene Symbol
PCDHA5
Entrez Gene ID
56143
Gene Name
protocadherin alpha 5
Gene Aliases
CNR6, CNRN6, CNRS6, CRNR6, PCDH-ALPHA5
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140821776-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031848.2 NM_031849.2

Gene Symbol
PCDHA6
Entrez Gene ID
56142
Gene Name
protocadherin alpha 6
Gene Aliases
CNR2, CNRN2, CNRS2, CRNR2, PCDH-ALPHA6
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140827922-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031848.2 NM_031849.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3776261 c.557G>A p.S186N Chr.5: 140828648-140828648 on Build GRCh38
1433841 c.297C>T p.S99S Chr.5: 140828388-140828388 on Build GRCh38
313965 c.225G>T p.L75L Chr.5: 140828316-140828316 on Build GRCh38
4913083 c.704T>C p.V235A Chr.5: 140828795-140828795 on Build GRCh38
6050793 c.470C>T p.A157V Chr.5: 140828561-140828561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782783283 Chr.5: 140828665-140828665 on Build GRCh38
rs782081687 Chr.5: 140828616-140828616 on Build GRCh38
rs782780518 Chr.5: 140828635-140828635 on Build GRCh38
rs148766603 Chr.5: 140828566-140828566 on Build GRCh38
rs782571134 Chr.5: 140828390-140828390 on Build GRCh38
rs782374670 Chr.5: 140828334-140828334 on Build GRCh38
rs782176995 Chr.5: 140828426-140828426 on Build GRCh38
rs782010399 Chr.5: 140828429-140828429 on Build GRCh38
rs782343743 Chr.5: 140828423-140828423 on Build GRCh38
rs184176193 Chr.5: 140828686-140828686 on Build GRCh38
rs374677079 Chr.5: 140828733-140828733 on Build GRCh38
rs577643736 Chr.5: 140828476-140828476 on Build GRCh38
rs782008680 Chr.5: 140828449-140828449 on Build GRCh38
rs140846988 Chr.5: 140828361-140828361 on Build GRCh38
rs371049380 Chr.5: 140828336-140828336 on Build GRCh38
rs781991870 Chr.5: 140828635-140828636 on Build GRCh38
rs782092268 Chr.5: 140828452-140828452 on Build GRCh38
rs782016889 Chr.5: 140828724-140828724 on Build GRCh38
rs377600907 Chr.5: 140828467-140828467 on Build GRCh38
rs782282808 Chr.5: 140828598-140828598 on Build GRCh38
rs782486776 Chr.5: 140828668-140828668 on Build GRCh38
rs782527541 Chr.5: 140828485-140828485 on Build GRCh38
rs781937389 Chr.5: 140828324-140828324 on Build GRCh38
rs373076921 Chr.5: 140828559-140828559 on Build GRCh38
rs782612074 Chr.5: 140828712-140828712 on Build GRCh38
rs200187130 Chr.5: 140828776-140828776 on Build GRCh38
rs782198345 Chr.5: 140828412-140828412 on Build GRCh38
rs142386076 Chr.5: 140828716-140828716 on Build GRCh38
rs201515728 Chr.5: 140828741-140828741 on Build GRCh38
rs782580018 Chr.5: 140828677-140828677 on Build GRCh38
rs544725649 Chr.5: 140828477-140828477 on Build GRCh38
rs782260049 Chr.5: 140828700-140828700 on Build GRCh38
rs782489134 Chr.5: 140828384-140828384 on Build GRCh38
rs782234690 Chr.5: 140828580-140828580 on Build GRCh38
rs782076570 Chr.5: 140828337-140828337 on Build GRCh38
rs540932674 Chr.5: 140828509-140828509 on Build GRCh38
rs781898636 Chr.5: 140828636-140828636 on Build GRCh38
rs150162226 Chr.5: 140828380-140828380 on Build GRCh38
rs782308685 Chr.5: 140828583-140828583 on Build GRCh38
rs782552925 Chr.5: 140828693-140828693 on Build GRCh38
rs144353163 Chr.5: 140828544-140828544 on Build GRCh38
rs61730778 Chr.5: 140828523-140828523 on Build GRCh38
rs782122229 Chr.5: 140828658-140828658 on Build GRCh38
rs781966680 Chr.5: 140828517-140828517 on Build GRCh38
rs782698898 Chr.5: 140828557-140828557 on Build GRCh38
rs371704232 Chr.5: 140828657-140828657 on Build GRCh38
rs781859688 Chr.5: 140828561-140828561 on Build GRCh38
rs782423460 Chr.5: 140828604-140828604 on Build GRCh38
rs782255340 Chr.5: 140828421-140828421 on Build GRCh38
rs782277050 Chr.5: 140828505-140828505 on Build GRCh38
rs782086693 Chr.5: 140828749-140828749 on Build GRCh38
rs782678285 Chr.5: 140828560-140828560 on Build GRCh38
rs781990697 Chr.5: 140828508-140828508 on Build GRCh38
rs782432594 Chr.5: 140828493-140828493 on Build GRCh38
rs147962839 Chr.5: 140828760-140828760 on Build GRCh38
rs267600391 Chr.5: 140828442-140828442 on Build GRCh38
rs148032873 Chr.5: 140828348-140828348 on Build GRCh38
rs552780058 Chr.5: 140828685-140828685 on Build GRCh38
rs367748457 Chr.5: 140828620-140828620 on Build GRCh38
rs782056319 Chr.5: 140828362-140828362 on Build GRCh38
rs111411595 Chr.5: 140828666-140828666 on Build GRCh38
rs782792755 Chr.5: 140828766-140828766 on Build GRCh38
rs782717263 Chr.5: 140828520-140828520 on Build GRCh38
rs782541724 Chr.5: 140828413-140828413 on Build GRCh38
rs370641869 Chr.5: 140828481-140828481 on Build GRCh38
rs782768043 Chr.5: 140828534-140828534 on Build GRCh38
rs782348722 Chr.5: 140828511-140828511 on Build GRCh38
rs782159518 Chr.5: 140828328-140828328 on Build GRCh38
rs782072726 Chr.5: 140828762-140828762 on Build GRCh38
rs782337385 Chr.5: 140828708-140828708 on Build GRCh38
rs782536026 Chr.5: 140828565-140828565 on Build GRCh38
rs782356866 Chr.5: 140828617-140828617 on Build GRCh38
rs141978908 Chr.5: 140828437-140828437 on Build GRCh38
rs782811756 Chr.5: 140828367-140828367 on Build GRCh38
rs375061677 Chr.5: 140828518-140828518 on Build GRCh38
rs782588798 Chr.5: 140828589-140828589 on Build GRCh38
rs782510418 Chr.5: 140828379-140828379 on Build GRCh38
rs138737999 Chr.5: 140828424-140828424 on Build GRCh38
rs782579824 Chr.5: 140828499-140828499 on Build GRCh38
rs782114453 Chr.5: 140828525-140828525 on Build GRCh38
rs781908580 Chr.5: 140828371-140828371 on Build GRCh38
rs782747374 Chr.5: 140828462-140828462 on Build GRCh38
rs781786009 Chr.5: 140828356-140828356 on Build GRCh38
rs782059484 Chr.5: 140828621-140828621 on Build GRCh38
rs782639326 Chr.5: 140828417-140828417 on Build GRCh38
rs781996310 Chr.5: 140828613-140828613 on Build GRCh38
rs146288766 Chr.5: 140828454-140828454 on Build GRCh38
rs369224613 Chr.5: 140828538-140828538 on Build GRCh38
rs782689416 Chr.5: 140828699-140828699 on Build GRCh38
rs782007465 Chr.5: 140828738-140828738 on Build GRCh38
rs150883390 Chr.5: 140828671-140828671 on Build GRCh38
rs782321949 Chr.5: 140828717-140828717 on Build GRCh38
rs139629080 Chr.5: 140828492-140828492 on Build GRCh38

View Full Product Details