Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140877468-140877969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHA1
Entrez Gene ID
56147
Gene Name
protocadherin alpha 1
Gene Aliases
PCDH-ALPHA1
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140786136-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_031864.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA10
Entrez Gene ID
56139
Gene Name
protocadherin alpha 10
Gene Aliases
CNR8, CNRN8, CNRS8, CRNR8, PCDH-ALPHA10
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140855883-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_031864.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA11
Entrez Gene ID
56138
Gene Name
protocadherin alpha 11
Gene Aliases
CNR7, CNRN7, CNRS7, CRNR7, PCDH-ALPHA11
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140867513-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_031864.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA12
Entrez Gene ID
56137
Gene Name
protocadherin alpha 12
Gene Aliases
PCDH-ALPHA12
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140875302-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_031864.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA2
Entrez Gene ID
56146
Gene Name
protocadherin alpha 2
Gene Aliases
PCDH-ALPHA2
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140794859-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_031864.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA3
Entrez Gene ID
56145
Gene Name
protocadherin alpha 3
Gene Aliases
PCDH-ALPHA3
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140801198-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_031864.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA4
Entrez Gene ID
56144
Gene Name
protocadherin alpha 4
Gene Aliases
CNR1, CNRN1, CRNR1, PCDH-ALPHA4
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140807074-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_031864.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA5
Entrez Gene ID
56143
Gene Name
protocadherin alpha 5
Gene Aliases
CNR6, CNRN6, CNRS6, CRNR6, PCDH-ALPHA5
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140821776-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_031864.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA6
Entrez Gene ID
56142
Gene Name
protocadherin alpha 6
Gene Aliases
CNR2, CNRN2, CNRS2, CRNR2, PCDH-ALPHA6
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140827922-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_031864.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA7
Entrez Gene ID
56141
Gene Name
protocadherin alpha 7
Gene Aliases
CNR4, CNRN4, CNRS4, CRNR4, PCDH-ALPHA7
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140834384-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_031864.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA8
Entrez Gene ID
56140
Gene Name
protocadherin alpha 8
Gene Aliases
PCDH-ALPHA8
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140841322-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_031864.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA9
Entrez Gene ID
9752
Gene Name
protocadherin alpha 9
Gene Aliases
PCDH-ALPHA9
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140847772-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_031864.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3725359 c.2177C>T p.P726L Chr.5: 140877649-140877649 on Build GRCh38
3851236 c.2184C>T p.T728T Chr.5: 140877656-140877656 on Build GRCh38
7091690 c.2178G>A p.P726P Chr.5: 140877650-140877650 on Build GRCh38
6298617 c.2220G>T p.V740V Chr.5: 140877692-140877692 on Build GRCh38
5633176 c.2224T>A p.S742T Chr.5: 140877696-140877696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044322759 Chr.5: 140877540-140877540 on Build GRCh38
rs782765998 Chr.5: 140877627-140877627 on Build GRCh38
rs782346022 Chr.5: 140877702-140877702 on Build GRCh38
rs377126743 Chr.5: 140877765-140877765 on Build GRCh38
rs782747532 Chr.5: 140877869-140877869 on Build GRCh38
rs376081263 Chr.5: 140877604-140877604 on Build GRCh38
rs979836328 Chr.5: 140877736-140877736 on Build GRCh38
rs781985023 Chr.5: 140877676-140877676 on Build GRCh38
rs782090888 Chr.5: 140877583-140877583 on Build GRCh38
rs781933436 Chr.5: 140877553-140877553 on Build GRCh38
rs781796819 Chr.5: 140877706-140877707 on Build GRCh38
rs904032241 Chr.5: 140877889-140877889 on Build GRCh38
rs781878840 Chr.5: 140877500-140877500 on Build GRCh38
rs996962116 Chr.5: 140877590-140877590 on Build GRCh38
rs904001764 Chr.5: 140877581-140877581 on Build GRCh38
rs369703340 Chr.5: 140877649-140877649 on Build GRCh38
rs200651425 Chr.5: 140877744-140877744 on Build GRCh38
rs781824762 Chr.5: 140877509-140877509 on Build GRCh38
rs782259804 Chr.5: 140877792-140877792 on Build GRCh38
rs782271327 Chr.5: 140877517-140877517 on Build GRCh38
rs782112072 Chr.5: 140877719-140877719 on Build GRCh38
rs570815670 Chr.5: 140877725-140877725 on Build GRCh38
rs782179941 Chr.5: 140877558-140877558 on Build GRCh38
rs782370638 Chr.5: 140877810-140877810 on Build GRCh38
rs892674985 Chr.5: 140877508-140877508 on Build GRCh38
rs539793128 Chr.5: 140877738-140877738 on Build GRCh38
rs782021677 Chr.5: 140877568-140877568 on Build GRCh38
rs781814794 Chr.5: 140877726-140877726 on Build GRCh38
rs782249537 Chr.5: 140877751-140877751 on Build GRCh38
rs116613760 Chr.5: 140877704-140877704 on Build GRCh38
rs782804856 Chr.5: 140877850-140877850 on Build GRCh38
rs945643108 Chr.5: 140877844-140877844 on Build GRCh38
rs538675333 Chr.5: 140877944-140877944 on Build GRCh38
rs138162923 Chr.5: 140877671-140877671 on Build GRCh38
rs781916987 Chr.5: 140877828-140877828 on Build GRCh38
rs782433528 Chr.5: 140877668-140877668 on Build GRCh38
rs781827236 Chr.5: 140877873-140877873 on Build GRCh38
rs782472866 Chr.5: 140877646-140877646 on Build GRCh38
rs974017564 Chr.5: 140877705-140877705 on Build GRCh38
rs189370080 Chr.5: 140877740-140877740 on Build GRCh38
rs782480713 Chr.5: 140877758-140877758 on Build GRCh38
rs782719581 Chr.5: 140877594-140877594 on Build GRCh38
rs782156769 Chr.5: 140877832-140877832 on Build GRCh38
rs781785108 Chr.5: 140877488-140877488 on Build GRCh38
rs782342116 Chr.5: 140877547-140877547 on Build GRCh38
rs267600400 Chr.5: 140877555-140877555 on Build GRCh38
rs372461540 Chr.5: 140877772-140877772 on Build GRCh38
rs142399025 Chr.5: 140877706-140877706 on Build GRCh38
rs782686661 Chr.5: 140877774-140877774 on Build GRCh38
rs782533203 Chr.5: 140877607-140877607 on Build GRCh38
rs782617127 Chr.5: 140877506-140877506 on Build GRCh38
rs781878627 Chr.5: 140877852-140877852 on Build GRCh38
rs782076422 Chr.5: 140877815-140877815 on Build GRCh38
rs782654875 Chr.5: 140877750-140877750 on Build GRCh38
rs548426920 Chr.5: 140877669-140877669 on Build GRCh38
rs782471585 Chr.5: 140877874-140877874 on Build GRCh38
rs782663782 Chr.5: 140877799-140877799 on Build GRCh38
rs559831598 Chr.5: 140877580-140877580 on Build GRCh38
rs548787462 Chr.5: 140877641-140877641 on Build GRCh38
rs377753884 Chr.5: 140877685-140877685 on Build GRCh38
rs782391639 Chr.5: 140877542-140877542 on Build GRCh38
rs549678873 Chr.5: 140877710-140877711 on Build GRCh38
rs563591404 Chr.5: 140877496-140877496 on Build GRCh38
rs782120587 Chr.5: 140877867-140877867 on Build GRCh38
rs781999747 Chr.5: 140877714-140877714 on Build GRCh38
rs782565949 Chr.5: 140877881-140877881 on Build GRCh38
rs782624202 Chr.5: 140877544-140877544 on Build GRCh38
rs782165291 Chr.5: 140877489-140877489 on Build GRCh38
rs781966059 Chr.5: 140877813-140877813 on Build GRCh38
rs782129855 Chr.5: 140877569-140877569 on Build GRCh38
rs782465464 Chr.5: 140877730-140877730 on Build GRCh38
rs146522449 Chr.5: 140877816-140877816 on Build GRCh38
rs781946062 Chr.5: 140877703-140877703 on Build GRCh38
rs782395925 Chr.5: 140877672-140877672 on Build GRCh38
rs782415331 Chr.5: 140877564-140877564 on Build GRCh38
rs532509235 Chr.5: 140877504-140877504 on Build GRCh38
rs61274888 Chr.5: 140877941-140877941 on Build GRCh38
rs1056644080 Chr.5: 140877503-140877503 on Build GRCh38
rs782777600 Chr.5: 140877578-140877578 on Build GRCh38
rs782609531 Chr.5: 140877760-140877760 on Build GRCh38

View Full Product Details