Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141052912-141053421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101926905
Entrez Gene ID
101926905
Gene Name
uncharacterized LOC101926905
Gene Aliases
-
UniGene
Hs.570900
Gene Chromosome Location
Chr.5: 141046205-141078031 on Build GRCh38
Transcripts
NR_105056.2 NM_013340.3

Gene Symbol
PCDHB1
Entrez Gene ID
29930
Gene Name
protocadherin beta 1
Gene Aliases
PCDH-BETA1
UniGene
Hs.278950
Gene Chromosome Location
Chr.5: 141051376-141053962 on Build GRCh38
Transcripts
NR_105056.2 NM_013340.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3610490 c.1705G>A p.G569S Chr.5: 141053175-141053175 on Build GRCh38
1434142 c.1482delA p.N496fs*12 Chr.5: 141052952-141052952 on Build GRCh38
6012003 c.1601C>A p.A534E Chr.5: 141053071-141053071 on Build GRCh38
1062523 c.1660G>T p.D554Y Chr.5: 141053130-141053130 on Build GRCh38
3610488 c.1663G>A p.D555N Chr.5: 141053133-141053133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782030135 Chr.5: 141053204-141053204 on Build GRCh38
rs181406572 Chr.5: 141053061-141053061 on Build GRCh38
rs782598854 Chr.5: 141053150-141053150 on Build GRCh38
rs781946694 Chr.5: 141053361-141053361 on Build GRCh38
rs782294928 Chr.5: 141053331-141053331 on Build GRCh38
rs782702515 Chr.5: 141052988-141052988 on Build GRCh38
rs781895139 Chr.5: 141052978-141052978 on Build GRCh38
rs2233593 Chr.5: 141053257-141053257 on Build GRCh38
rs928695414 Chr.5: 141052990-141052990 on Build GRCh38
rs782542950 Chr.5: 141053331-141053332 on Build GRCh38
rs782348063 Chr.5: 141052959-141052959 on Build GRCh38
rs782612281 Chr.5: 141053289-141053289 on Build GRCh38
rs782610594 Chr.5: 141053025-141053025 on Build GRCh38
rs782466025 Chr.5: 141053129-141053129 on Build GRCh38
rs374507673 Chr.5: 141053397-141053397 on Build GRCh38
rs782677143 Chr.5: 141053348-141053348 on Build GRCh38
rs782278329 Chr.5: 141053029-141053029 on Build GRCh38
rs782569661 Chr.5: 141053391-141053391 on Build GRCh38
rs782455855 Chr.5: 141053284-141053284 on Build GRCh38
rs201674417 Chr.5: 141053375-141053375 on Build GRCh38
rs782637010 Chr.5: 141053322-141053322 on Build GRCh38
rs781947713 Chr.5: 141052970-141052970 on Build GRCh38
rs370099926 Chr.5: 141053044-141053044 on Build GRCh38
rs782141261 Chr.5: 141053209-141053209 on Build GRCh38
rs782520320 Chr.5: 141053297-141053297 on Build GRCh38
rs10476822 Chr.5: 141053302-141053302 on Build GRCh38
rs782541730 Chr.5: 141053016-141053016 on Build GRCh38
rs116101801 Chr.5: 141053017-141053017 on Build GRCh38
rs377174015 Chr.5: 141052999-141052999 on Build GRCh38
rs782279373 Chr.5: 141053042-141053042 on Build GRCh38
rs782703917 Chr.5: 141053218-141053218 on Build GRCh38
rs781845845 Chr.5: 141053003-141053003 on Build GRCh38
rs782693218 Chr.5: 141053110-141053110 on Build GRCh38
rs782781242 Chr.5: 141052951-141052952 on Build GRCh38
rs201337487 Chr.5: 141052976-141052976 on Build GRCh38
rs781931412 Chr.5: 141052958-141052958 on Build GRCh38
rs137984585 Chr.5: 141053174-141053174 on Build GRCh38
rs782535704 Chr.5: 141053146-141053146 on Build GRCh38
rs1043343231 Chr.5: 141053035-141053035 on Build GRCh38
rs186168843 Chr.5: 141053116-141053116 on Build GRCh38
rs782352744 Chr.5: 141053355-141053355 on Build GRCh38
rs782393645 Chr.5: 141052951-141052951 on Build GRCh38
rs193920928 Chr.5: 141053125-141053125 on Build GRCh38
rs782689641 Chr.5: 141053337-141053337 on Build GRCh38
rs139444803 Chr.5: 141052953-141052953 on Build GRCh38
rs781881695 Chr.5: 141053084-141053084 on Build GRCh38
rs782264329 Chr.5: 141053195-141053195 on Build GRCh38
rs782434716 Chr.5: 141053023-141053023 on Build GRCh38
rs781814624 Chr.5: 141053325-141053326 on Build GRCh38
rs200740778 Chr.5: 141053336-141053336 on Build GRCh38
rs782201253 Chr.5: 141053027-141053027 on Build GRCh38
rs782073325 Chr.5: 141052983-141052983 on Build GRCh38
rs200436440 Chr.5: 141053133-141053133 on Build GRCh38
rs782591373 Chr.5: 141053345-141053345 on Build GRCh38
rs782642176 Chr.5: 141053011-141053011 on Build GRCh38
rs370278362 Chr.5: 141053137-141053137 on Build GRCh38
rs782195352 Chr.5: 141053021-141053021 on Build GRCh38
rs202201120 Chr.5: 141053059-141053059 on Build GRCh38
rs782038099 Chr.5: 141053028-141053028 on Build GRCh38
rs782331553 Chr.5: 141053040-141053040 on Build GRCh38
rs782721792 Chr.5: 141053251-141053251 on Build GRCh38
rs782463985 Chr.5: 141053109-141053109 on Build GRCh38
rs782209457 Chr.5: 141052943-141052943 on Build GRCh38
rs782644370 Chr.5: 141053132-141053132 on Build GRCh38
rs140938729 Chr.5: 141053275-141053275 on Build GRCh38
rs17208383 Chr.5: 141053041-141053041 on Build GRCh38
rs782119603 Chr.5: 141052971-141052971 on Build GRCh38
rs781953615 Chr.5: 141053183-141053183 on Build GRCh38
rs782096648 Chr.5: 141053247-141053247 on Build GRCh38
rs890202647 Chr.5: 141053120-141053120 on Build GRCh38
rs782375909 Chr.5: 141053201-141053201 on Build GRCh38
rs144159085 Chr.5: 141053007-141053007 on Build GRCh38
rs782818866 Chr.5: 141053081-141053081 on Build GRCh38
rs781865599 Chr.5: 141053255-141053255 on Build GRCh38
rs549296755 Chr.5: 141053015-141053015 on Build GRCh38
rs781826035 Chr.5: 141053293-141053293 on Build GRCh38
rs782706930 Chr.5: 141053268-141053268 on Build GRCh38
rs782553505 Chr.5: 141053162-141053162 on Build GRCh38
rs782109640 Chr.5: 141053070-141053070 on Build GRCh38
rs928749339 Chr.5: 141053399-141053399 on Build GRCh38
rs947338315 Chr.5: 141053020-141053020 on Build GRCh38
rs781912955 Chr.5: 141053229-141053229 on Build GRCh38

View Full Product Details