Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141422823-141423315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA10
Entrez Gene ID
56106
Gene Name
protocadherin gamma subfamily A, 10
Gene Aliases
PCDH-GAMMA-A10
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141413176-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA11
Entrez Gene ID
56105
Gene Name
protocadherin gamma subfamily A, 11
Gene Aliases
PCDH-GAMMA-A11
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141420970-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA3
Entrez Gene ID
56112
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
PCDH-GAMMA-A3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141344034-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141355025-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA5
Entrez Gene ID
56110
Gene Name
protocadherin gamma subfamily A, 5
Gene Aliases
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141364331-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA6
Entrez Gene ID
56109
Gene Name
protocadherin gamma subfamily A, 6
Gene Aliases
PCDH-GAMMA-A6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141374084-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA7
Entrez Gene ID
56108
Gene Name
protocadherin gamma subfamily A, 7
Gene Aliases
PCDH-GAMMA-A7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141382739-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA8
Entrez Gene ID
9708
Gene Name
protocadherin gamma subfamily A, 8
Gene Aliases
PCDH-GAMMA-A8
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141391916-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA9
Entrez Gene ID
56107
Gene Name
protocadherin gamma subfamily A, 9
Gene Aliases
PCDH-GAMMA-A9
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141402953-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141350261-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141360136-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB3
Entrez Gene ID
56102
Gene Name
protocadherin gamma subfamily B, 3
Gene Aliases
PCDH-GAMMA-B3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141370264-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB4
Entrez Gene ID
8641
Gene Name
protocadherin gamma subfamily B, 4
Gene Aliases
CDH20, FIB2, PCDH-GAMMA-B4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141387885-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB5
Entrez Gene ID
56101
Gene Name
protocadherin gamma subfamily B, 5
Gene Aliases
PCDH-GAMMA-B5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141398128-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB6
Entrez Gene ID
56100
Gene Name
protocadherin gamma subfamily B, 6
Gene Aliases
PCDH-GAMMA-B6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141408203-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB7
Entrez Gene ID
56099
Gene Name
protocadherin gamma subfamily B, 7
Gene Aliases
ME6, PCDH-GAMMA-B7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141417647-141512981 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3768200 c.1843C>T p.P615S Chr.5: 141423070-141423070 on Build GRCh38
7269065 c.2048C>T p.A683V Chr.5: 141423275-141423275 on Build GRCh38
3409945 c.1891C>T p.R631W Chr.5: 141423118-141423118 on Build GRCh38
1261426 c.1987G>A p.V663M Chr.5: 141423214-141423214 on Build GRCh38
6740610 c.2002A>G p.S668G Chr.5: 141423229-141423229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756589496 Chr.5: 141422910-141422910 on Build GRCh38
rs530404769 Chr.5: 141423267-141423267 on Build GRCh38
rs188787674 Chr.5: 141422923-141422923 on Build GRCh38
rs1029118054 Chr.5: 141423190-141423190 on Build GRCh38
rs62621243 Chr.5: 141423110-141423110 on Build GRCh38
rs761825236 Chr.5: 141423237-141423237 on Build GRCh38
rs778561065 Chr.5: 141423102-141423102 on Build GRCh38
rs374000303 Chr.5: 141423116-141423116 on Build GRCh38
rs201821221 Chr.5: 141423089-141423089 on Build GRCh38
rs757110751 Chr.5: 141423067-141423067 on Build GRCh38
rs367805855 Chr.5: 141423166-141423166 on Build GRCh38
rs200154593 Chr.5: 141423053-141423053 on Build GRCh38
rs561451935 Chr.5: 141423179-141423179 on Build GRCh38
rs748586131 Chr.5: 141423124-141423124 on Build GRCh38
rs781145334 Chr.5: 141422928-141422928 on Build GRCh38
rs1049470142 Chr.5: 141423029-141423029 on Build GRCh38
rs954945082 Chr.5: 141423210-141423210 on Build GRCh38
rs940921497 Chr.5: 141423043-141423043 on Build GRCh38
rs759972749 Chr.5: 141422899-141422899 on Build GRCh38
rs62378455 Chr.5: 141422889-141422889 on Build GRCh38
rs554024395 Chr.5: 141423287-141423289 on Build GRCh38
rs558006468 Chr.5: 141422976-141422976 on Build GRCh38
rs376530221 Chr.5: 141423216-141423216 on Build GRCh38
rs773469060 Chr.5: 141422872-141422872 on Build GRCh38
rs527921011 Chr.5: 141423157-141423157 on Build GRCh38
rs372620011 Chr.5: 141422963-141422963 on Build GRCh38
rs777535652 Chr.5: 141422941-141422941 on Build GRCh38
rs375287728 Chr.5: 141423132-141423132 on Build GRCh38
rs116031289 Chr.5: 141422867-141422867 on Build GRCh38
rs752144906 Chr.5: 141423078-141423078 on Build GRCh38
rs760086052 Chr.5: 141423183-141423184 on Build GRCh38
rs769630623 Chr.5: 141422940-141422940 on Build GRCh38
rs114907564 Chr.5: 141422997-141422997 on Build GRCh38
rs752667215 Chr.5: 141423268-141423268 on Build GRCh38
rs750278899 Chr.5: 141423253-141423253 on Build GRCh38
rs750186629 Chr.5: 141423159-141423159 on Build GRCh38
rs776903094 Chr.5: 141423157-141423159 on Build GRCh38
rs13173820 Chr.5: 141423002-141423002 on Build GRCh38
rs375881737 Chr.5: 141422964-141422964 on Build GRCh38
rs996177363 Chr.5: 141423185-141423185 on Build GRCh38
rs775170753 Chr.5: 141423219-141423219 on Build GRCh38
rs779233337 Chr.5: 141423003-141423003 on Build GRCh38
rs755141371 Chr.5: 141423170-141423170 on Build GRCh38
rs771403541 Chr.5: 141423052-141423054 on Build GRCh38
rs765494785 Chr.5: 141423048-141423048 on Build GRCh38
rs202035589 Chr.5: 141422884-141422884 on Build GRCh38
rs747938944 Chr.5: 141423184-141423184 on Build GRCh38
rs746989617 Chr.5: 141422853-141422853 on Build GRCh38
rs761268652 Chr.5: 141423031-141423031 on Build GRCh38
rs545052756 Chr.5: 141423147-141423147 on Build GRCh38
rs62378456 Chr.5: 141423156-141423156 on Build GRCh38
rs368969800 Chr.5: 141423255-141423255 on Build GRCh38
rs748689237 Chr.5: 141423177-141423177 on Build GRCh38
rs762200805 Chr.5: 141423045-141423045 on Build GRCh38
rs13173779 Chr.5: 141422920-141422920 on Build GRCh38
rs970045921 Chr.5: 141422880-141422880 on Build GRCh38
rs747261683 Chr.5: 141422898-141422899 on Build GRCh38
rs776884698 Chr.5: 141423039-141423039 on Build GRCh38
rs768086403 Chr.5: 141422901-141422901 on Build GRCh38
rs13188208 Chr.5: 141422931-141422931 on Build GRCh38
rs376452870 Chr.5: 141423273-141423273 on Build GRCh38
rs752323344 Chr.5: 141422929-141422929 on Build GRCh38
rs368205535 Chr.5: 141423121-141423121 on Build GRCh38
rs771340929 Chr.5: 141423296-141423296 on Build GRCh38
rs749771302 Chr.5: 141423277-141423277 on Build GRCh38
rs759618535 Chr.5: 141422952-141422952 on Build GRCh38
rs763219526 Chr.5: 141422874-141422874 on Build GRCh38
rs781125798 Chr.5: 141423176-141423176 on Build GRCh38
rs765040062 Chr.5: 141423238-141423238 on Build GRCh38
rs772288489 Chr.5: 141422890-141422898 on Build GRCh38
rs755975773 Chr.5: 141422986-141422986 on Build GRCh38
rs13188214 Chr.5: 141422943-141422943 on Build GRCh38
rs372798900 Chr.5: 141423272-141423272 on Build GRCh38
rs568894245 Chr.5: 141422925-141422925 on Build GRCh38
rs769232324 Chr.5: 141423034-141423034 on Build GRCh38
rs758065876 Chr.5: 141422916-141422916 on Build GRCh38
rs370773437 Chr.5: 141423131-141423131 on Build GRCh38
rs771252840 Chr.5: 141423202-141423202 on Build GRCh38
rs769543752 Chr.5: 141422871-141422871 on Build GRCh38
rs763729316 Chr.5: 141423225-141423225 on Build GRCh38
rs766010601 Chr.5: 141422968-141422968 on Build GRCh38
rs771088007 Chr.5: 141423000-141423000 on Build GRCh38
rs760308436 Chr.5: 141423222-141423222 on Build GRCh38
rs769320490 Chr.5: 141423119-141423119 on Build GRCh38
rs745486981 Chr.5: 141422846-141422846 on Build GRCh38
rs370127569 Chr.5: 141422859-141422859 on Build GRCh38
rs751894091 Chr.5: 141423256-141423256 on Build GRCh38
rs776189143 Chr.5: 141422957-141422957 on Build GRCh38
rs749613139 Chr.5: 141423201-141423201 on Build GRCh38
rs13188215 Chr.5: 141422945-141422945 on Build GRCh38
rs777558098 Chr.5: 141422989-141422989 on Build GRCh38
rs772337723 Chr.5: 141423007-141423007 on Build GRCh38
rs778670588 Chr.5: 141422843-141422843 on Build GRCh38
rs973635802 Chr.5: 141423265-141423265 on Build GRCh38
rs375046528 Chr.5: 141422911-141422911 on Build GRCh38
rs753872725 Chr.5: 141423153-141423153 on Build GRCh38
rs184432819 Chr.5: 141422850-141422850 on Build GRCh38
rs766431596 Chr.5: 141422883-141422883 on Build GRCh38
rs760216287 Chr.5: 141423140-141423140 on Build GRCh38
rs200022455 Chr.5: 141423051-141423051 on Build GRCh38
rs199976232 Chr.5: 141422942-141422942 on Build GRCh38
rs778069795 Chr.5: 141422897-141422898 on Build GRCh38
rs562864937 Chr.5: 141423106-141423106 on Build GRCh38
rs763488632 Chr.5: 141423149-141423149 on Build GRCh38
rs770258338 Chr.5: 141423130-141423130 on Build GRCh38
rs770264100 Chr.5: 141423252-141423260 on Build GRCh38
rs535152193 Chr.5: 141422906-141422906 on Build GRCh38
rs751065595 Chr.5: 141422975-141422975 on Build GRCh38
rs757127033 Chr.5: 141423101-141423101 on Build GRCh38
rs200931939 Chr.5: 141423096-141423096 on Build GRCh38
rs780664832 Chr.5: 141422982-141422982 on Build GRCh38
rs758720418 Chr.5: 141423163-141423163 on Build GRCh38
rs762530904 Chr.5: 141422966-141422966 on Build GRCh38
rs747206648 Chr.5: 141423169-141423169 on Build GRCh38
rs775936938 Chr.5: 141423226-141423226 on Build GRCh38
rs767107885 Chr.5: 141423138-141423138 on Build GRCh38
rs746057587 Chr.5: 141423006-141423006 on Build GRCh38
rs776079289 Chr.5: 141423265-141423272 on Build GRCh38
rs371209178 Chr.5: 141423009-141423009 on Build GRCh38
rs752694873 Chr.5: 141422984-141422984 on Build GRCh38
rs756067751 Chr.5: 141423087-141423087 on Build GRCh38

View Full Product Details