Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141637943-141638446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RELL2
Entrez Gene ID
285613
Gene Name
RELT like 2
Gene Aliases
C5orf16
UniGene
Hs.120094
Gene Chromosome Location
Chr.5: 141636946-141641077 on Build GRCh38
Transcripts
NM_001130029.1 NM_173828.4 XM_005268414.3 XM_011537624.1 XM_011537625.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7156657 c.147G>A p.S49S Chr.5: 141638372-141638372 on Build GRCh38
3852235 c.29C>T p.P10L Chr.5: 141638254-141638254 on Build GRCh38
1671992 c.80T>C p.L27P Chr.5: 141638305-141638305 on Build GRCh38
5429709 c.147G>C p.S49S Chr.5: 141638372-141638372 on Build GRCh38
1741469 c.52C>T p.L18F Chr.5: 141638277-141638277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151008143 Chr.5: 141638153-141638153 on Build GRCh38
rs775375686 Chr.5: 141638420-141638420 on Build GRCh38
rs1045694010 Chr.5: 141637970-141637970 on Build GRCh38
rs1052656176 Chr.5: 141638035-141638035 on Build GRCh38
rs1009970377 Chr.5: 141638047-141638047 on Build GRCh38
rs199871078 Chr.5: 141638386-141638386 on Build GRCh38
rs904174615 Chr.5: 141638057-141638057 on Build GRCh38
rs976551 Chr.5: 141638019-141638019 on Build GRCh38
rs757296062 Chr.5: 141638244-141638244 on Build GRCh38
rs1010643920 Chr.5: 141638160-141638160 on Build GRCh38
rs571435495 Chr.5: 141638303-141638303 on Build GRCh38
rs756362183 Chr.5: 141638217-141638217 on Build GRCh38
rs991374358 Chr.5: 141638218-141638218 on Build GRCh38
rs758884850 Chr.5: 141638255-141638255 on Build GRCh38
rs551321177 Chr.5: 141638287-141638287 on Build GRCh38
rs959800075 Chr.5: 141638034-141638034 on Build GRCh38
rs149595207 Chr.5: 141638352-141638352 on Build GRCh38
rs1029292882 Chr.5: 141638082-141638082 on Build GRCh38
rs890385733 Chr.5: 141638092-141638092 on Build GRCh38
rs747885904 Chr.5: 141638399-141638399 on Build GRCh38
rs372310472 Chr.5: 141638369-141638369 on Build GRCh38
rs753172373 Chr.5: 141638211-141638212 on Build GRCh38
rs752016690 Chr.5: 141638384-141638384 on Build GRCh38
rs1038885008 Chr.5: 141638140-141638140 on Build GRCh38
rs1882771 Chr.5: 141638053-141638053 on Build GRCh38
rs752430991 Chr.5: 141638213-141638213 on Build GRCh38
rs771952662 Chr.5: 141638419-141638419 on Build GRCh38
rs918572126 Chr.5: 141638060-141638060 on Build GRCh38
rs756771287 Chr.5: 141638296-141638298 on Build GRCh38
rs764341429 Chr.5: 141638342-141638342 on Build GRCh38
rs987709797 Chr.5: 141638090-141638090 on Build GRCh38
rs757419727 Chr.5: 141638348-141638348 on Build GRCh38
rs941474977 Chr.5: 141638051-141638051 on Build GRCh38
rs943157250 Chr.5: 141638098-141638098 on Build GRCh38
rs774120279 Chr.5: 141638211-141638211 on Build GRCh38
rs933943713 Chr.5: 141638334-141638334 on Build GRCh38
rs374328671 Chr.5: 141638279-141638279 on Build GRCh38
rs201225885 Chr.5: 141638207-141638207 on Build GRCh38
rs747335366 Chr.5: 141638265-141638265 on Build GRCh38
rs954654866 Chr.5: 141638083-141638083 on Build GRCh38
rs982311445 Chr.5: 141637967-141637967 on Build GRCh38
rs769588762 Chr.5: 141638266-141638266 on Build GRCh38
rs746351665 Chr.5: 141638251-141638251 on Build GRCh38
rs561202507 Chr.5: 141638007-141638007 on Build GRCh38
rs1014787332 Chr.5: 141638103-141638103 on Build GRCh38
rs750453294 Chr.5: 141638354-141638354 on Build GRCh38
rs772877848 Chr.5: 141638202-141638202 on Build GRCh38
rs976549 Chr.5: 141638149-141638149 on Build GRCh38
rs200324612 Chr.5: 141638246-141638246 on Build GRCh38
rs775287908 Chr.5: 141638330-141638330 on Build GRCh38
rs541032911 Chr.5: 141638022-141638022 on Build GRCh38
rs748935819 Chr.5: 141638392-141638392 on Build GRCh38
rs781022213 Chr.5: 141638385-141638385 on Build GRCh38
rs141153106 Chr.5: 141638212-141638212 on Build GRCh38
rs767419911 Chr.5: 141638312-141638312 on Build GRCh38
rs754055293 Chr.5: 141638346-141638346 on Build GRCh38
rs2545029 Chr.5: 141638372-141638372 on Build GRCh38
rs146541687 Chr.5: 141638408-141638408 on Build GRCh38
rs1035203566 Chr.5: 141638208-141638208 on Build GRCh38
rs370699427 Chr.5: 141638260-141638260 on Build GRCh38
rs753968754 Chr.5: 141638228-141638228 on Build GRCh38
rs370207888 Chr.5: 141638196-141638196 on Build GRCh38
rs995813881 Chr.5: 141638062-141638062 on Build GRCh38
rs374162574 Chr.5: 141638377-141638377 on Build GRCh38
rs769506455 Chr.5: 141638195-141638195 on Build GRCh38
rs973460367 Chr.5: 141638175-141638175 on Build GRCh38
rs774403356 Chr.5: 141638300-141638300 on Build GRCh38
rs891540110 Chr.5: 141638046-141638046 on Build GRCh38
rs764253568 Chr.5: 141638224-141638224 on Build GRCh38
rs1035274450 Chr.5: 141638023-141638023 on Build GRCh38
rs976550 Chr.5: 141638054-141638054 on Build GRCh38
rs909919670 Chr.5: 141638050-141638050 on Build GRCh38
rs889167453 Chr.5: 141637999-141637999 on Build GRCh38
rs368745319 Chr.5: 141638336-141638336 on Build GRCh38
rs1021228844 Chr.5: 141638048-141638048 on Build GRCh38
rs370536809 Chr.5: 141638194-141638194 on Build GRCh38
rs966378065 Chr.5: 141637966-141637966 on Build GRCh38
rs755288107 Chr.5: 141638242-141638242 on Build GRCh38
rs879814995 Chr.5: 141638053-141638054 on Build GRCh38
rs906791561 Chr.5: 141637989-141637989 on Build GRCh38

View Full Product Details