Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141929660-141930153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0141
Entrez Gene ID
9812
Gene Name
KIAA0141
Gene Aliases
DELE
UniGene
Hs.210532
Gene Chromosome Location
Chr.5: 141923785-141942047 on Build GRCh38
Transcripts
NM_001142603.2 NM_014773.4 XM_005268547.3 XM_005268548.3 XM_005268552.2 XM_011537715.1 XM_011537716.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1211839 c.515G>A p.R172Q Chr.5: 141929684-141929684 on Build GRCh38
6692974 c.564T>A p.S188S Chr.5: 141929733-141929733 on Build GRCh38
4407079 c.531C>T p.N177N Chr.5: 141929700-141929700 on Build GRCh38
4797840 c.541G>A p.G181R Chr.5: 141929710-141929710 on Build GRCh38
1619768 c.533C>G p.S178C Chr.5: 141929702-141929702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766099524 Chr.5: 141930085-141930085 on Build GRCh38
rs189222083 Chr.5: 141929983-141929983 on Build GRCh38
rs751190255 Chr.5: 141929990-141929990 on Build GRCh38
rs189652814 Chr.5: 141929846-141929846 on Build GRCh38
rs759352073 Chr.5: 141929683-141929683 on Build GRCh38
rs908042027 Chr.5: 141929819-141929819 on Build GRCh38
rs756799409 Chr.5: 141929702-141929702 on Build GRCh38
rs762408279 Chr.5: 141929986-141929986 on Build GRCh38
rs369567807 Chr.5: 141930029-141930029 on Build GRCh38
rs773052391 Chr.5: 141930062-141930062 on Build GRCh38
rs987619273 Chr.5: 141929808-141929808 on Build GRCh38
rs372468560 Chr.5: 141929792-141929792 on Build GRCh38
rs10078802 Chr.5: 141929700-141929700 on Build GRCh38
rs138993288 Chr.5: 141929688-141929688 on Build GRCh38
rs771164064 Chr.5: 141929942-141929942 on Build GRCh38
rs781743807 Chr.5: 141929780-141929780 on Build GRCh38
rs374773989 Chr.5: 141929724-141929724 on Build GRCh38
rs138552747 Chr.5: 141930035-141930035 on Build GRCh38
rs959019259 Chr.5: 141929860-141929860 on Build GRCh38
rs369742160 Chr.5: 141929973-141929973 on Build GRCh38
rs764061758 Chr.5: 141929754-141929754 on Build GRCh38
rs373158872 Chr.5: 141929994-141929994 on Build GRCh38
rs141459129 Chr.5: 141929682-141929682 on Build GRCh38
rs1037568526 Chr.5: 141929761-141929761 on Build GRCh38
rs144583494 Chr.5: 141929704-141929704 on Build GRCh38
rs768461946 Chr.5: 141929966-141929966 on Build GRCh38
rs774478255 Chr.5: 141929953-141929953 on Build GRCh38
rs1052556911 Chr.5: 141929922-141929922 on Build GRCh38
rs564159851 Chr.5: 141929775-141929775 on Build GRCh38
rs753924025 Chr.5: 141930031-141930031 on Build GRCh38
rs955898498 Chr.5: 141929756-141929756 on Build GRCh38
rs772346514 Chr.5: 141930052-141930052 on Build GRCh38
rs747392410 Chr.5: 141929713-141929713 on Build GRCh38
rs200808757 Chr.5: 141929768-141929768 on Build GRCh38
rs571217703 Chr.5: 141929739-141929739 on Build GRCh38
rs780464113 Chr.5: 141929711-141929711 on Build GRCh38
rs759260447 Chr.5: 141930106-141930106 on Build GRCh38
rs1004032545 Chr.5: 141929900-141929900 on Build GRCh38
rs780951057 Chr.5: 141930054-141930054 on Build GRCh38
rs762472760 Chr.5: 141930063-141930063 on Build GRCh38
rs766012193 Chr.5: 141929989-141929989 on Build GRCh38
rs74819654 Chr.5: 141930083-141930083 on Build GRCh38
rs533546209 Chr.5: 141929766-141929766 on Build GRCh38
rs910139906 Chr.5: 141929883-141929883 on Build GRCh38
rs371151428 Chr.5: 141930103-141930103 on Build GRCh38
rs746082068 Chr.5: 141930036-141930036 on Build GRCh38
rs184514932 Chr.5: 141929972-141929972 on Build GRCh38
rs192000208 Chr.5: 141929878-141929878 on Build GRCh38
rs377418159 Chr.5: 141930056-141930056 on Build GRCh38
rs748202306 Chr.5: 141929717-141929717 on Build GRCh38
rs1037897609 Chr.5: 141929839-141929839 on Build GRCh38
rs761254815 Chr.5: 141929765-141929765 on Build GRCh38
rs758625390 Chr.5: 141929708-141929708 on Build GRCh38
rs371713234 Chr.5: 141929706-141929706 on Build GRCh38
rs933074705 Chr.5: 141929804-141929804 on Build GRCh38
rs116469790 Chr.5: 141930118-141930118 on Build GRCh38
rs756216202 Chr.5: 141929803-141929803 on Build GRCh38
rs753091992 Chr.5: 141930112-141930112 on Build GRCh38
rs770012506 Chr.5: 141929719-141929719 on Build GRCh38
rs779197079 Chr.5: 141930094-141930094 on Build GRCh38
rs1045480417 Chr.5: 141929856-141929856 on Build GRCh38
rs746779925 Chr.5: 141929965-141929965 on Build GRCh38
rs1037216410 Chr.5: 141929929-141929929 on Build GRCh38
rs781775516 Chr.5: 141929716-141929716 on Build GRCh38
rs948051666 Chr.5: 141929850-141929850 on Build GRCh38
rs775339536 Chr.5: 141929728-141929728 on Build GRCh38
rs34689728 Chr.5: 141930048-141930048 on Build GRCh38
rs769779271 Chr.5: 141929981-141929981 on Build GRCh38
rs748682491 Chr.5: 141929781-141929781 on Build GRCh38
rs6888914 Chr.5: 141929789-141929789 on Build GRCh38
rs369039562 Chr.5: 141929769-141929769 on Build GRCh38
rs751918075 Chr.5: 141929772-141929772 on Build GRCh38
rs941585223 Chr.5: 141929928-141929928 on Build GRCh38
rs755491125 Chr.5: 141929714-141929714 on Build GRCh38
rs773506426 Chr.5: 141929720-141929720 on Build GRCh38
rs757900208 Chr.5: 141930017-141930018 on Build GRCh38
rs766866131 Chr.5: 141929730-141929731 on Build GRCh38
rs145156674 Chr.5: 141929684-141929684 on Build GRCh38
rs201111574 Chr.5: 141929699-141929699 on Build GRCh38
rs769685717 Chr.5: 141930057-141930057 on Build GRCh38
rs768002372 Chr.5: 141929993-141929993 on Build GRCh38
rs6862824 Chr.5: 141929820-141929820 on Build GRCh38
rs570183351 Chr.5: 141929689-141929689 on Build GRCh38
rs372768684 Chr.5: 141929783-141929783 on Build GRCh38
rs200197275 Chr.5: 141929686-141929686 on Build GRCh38
rs891621669 Chr.5: 141929951-141929951 on Build GRCh38
rs778853673 Chr.5: 141930122-141930122 on Build GRCh38
rs138477141 Chr.5: 141929991-141929991 on Build GRCh38
rs776581221 Chr.5: 141929764-141929764 on Build GRCh38
rs761164092 Chr.5: 141930114-141930114 on Build GRCh38
rs200320676 Chr.5: 141930019-141930019 on Build GRCh38
rs770356193 Chr.5: 141930064-141930064 on Build GRCh38
rs749526729 Chr.5: 141929722-141929722 on Build GRCh38
rs757304984 Chr.5: 141930120-141930120 on Build GRCh38
rs777999050 Chr.5: 141929788-141929788 on Build GRCh38
rs80255092 Chr.5: 141930082-141930082 on Build GRCh38
rs757312975 Chr.5: 141930032-141930032 on Build GRCh38
rs995824294 Chr.5: 141930013-141930013 on Build GRCh38
rs1007473228 Chr.5: 141929779-141929779 on Build GRCh38
rs767339583 Chr.5: 141930107-141930107 on Build GRCh38
rs558109011 Chr.5: 141929775-141929775 on Build GRCh38

View Full Product Details