Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149857930-149858474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE6A
Entrez Gene ID
5145
Gene Name
phosphodiesterase 6A
Gene Aliases
CGPR-A, PDEA, RP43
UniGene
Hs.567314
Gene Chromosome Location
Chr.5: 149857956-149944793 on Build GRCh38
Transcripts
NM_000440.2

SNP Detail

SNP ID Chromosome Location
rs746054170 Chr.5: 149858389-149858389 on Build GRCh38
rs573718233 Chr.5: 149858295-149858295 on Build GRCh38
rs551329710 Chr.5: 149858434-149858434 on Build GRCh38
rs551002606 Chr.5: 149858363-149858363 on Build GRCh38
rs778443310 Chr.5: 149858164-149858164 on Build GRCh38
rs578087908 Chr.5: 149858178-149858178 on Build GRCh38
rs879779624 Chr.5: 149858290-149858291 on Build GRCh38
rs567865002 Chr.5: 149858417-149858417 on Build GRCh38
rs548763561 Chr.5: 149858403-149858403 on Build GRCh38
rs898054450 Chr.5: 149858276-149858277 on Build GRCh38
rs1014870825 Chr.5: 149858067-149858067 on Build GRCh38
rs1002872058 Chr.5: 149857958-149857958 on Build GRCh38
rs750630343 Chr.5: 149858278-149858278 on Build GRCh38
rs921100849 Chr.5: 149858102-149858102 on Build GRCh38
rs17110632 Chr.5: 149858124-149858124 on Build GRCh38
rs569137634 Chr.5: 149858254-149858258 on Build GRCh38
rs952382210 Chr.5: 149858312-149858312 on Build GRCh38
rs757612877 Chr.5: 149858006-149858006 on Build GRCh38
rs945906894 Chr.5: 149858429-149858429 on Build GRCh38
rs150127383 Chr.5: 149858233-149858233 on Build GRCh38
rs530632768 Chr.5: 149858391-149858391 on Build GRCh38
rs30884 Chr.5: 149858353-149858353 on Build GRCh38
rs905355607 Chr.5: 149858236-149858236 on Build GRCh38
rs758455690 Chr.5: 149858272-149858272 on Build GRCh38
rs925886821 Chr.5: 149858143-149858143 on Build GRCh38
rs777837397 Chr.5: 149858279-149858279 on Build GRCh38
rs965859377 Chr.5: 149858069-149858069 on Build GRCh38
rs188603235 Chr.5: 149858275-149858275 on Build GRCh38
rs542706002 Chr.5: 149858274-149858274 on Build GRCh38
rs1043042673 Chr.5: 149858269-149858269 on Build GRCh38
rs978671593 Chr.5: 149858433-149858433 on Build GRCh38
rs945957451 Chr.5: 149858266-149858266 on Build GRCh38
rs534880825 Chr.5: 149858446-149858446 on Build GRCh38
rs78827133 Chr.5: 149858126-149858126 on Build GRCh38
rs998676408 Chr.5: 149858298-149858298 on Build GRCh38
rs73796297 Chr.5: 149858042-149858042 on Build GRCh38
rs183299988 Chr.5: 149858271-149858271 on Build GRCh38
rs982770260 Chr.5: 149858012-149858012 on Build GRCh38
rs1015163573 Chr.5: 149858079-149858080 on Build GRCh38
rs752529909 Chr.5: 149858396-149858396 on Build GRCh38
rs911003892 Chr.5: 149858183-149858183 on Build GRCh38
rs202239422 Chr.5: 149858268-149858269 on Build GRCh38
rs559249270 Chr.5: 149858334-149858334 on Build GRCh38
rs953998578 Chr.5: 149858315-149858315 on Build GRCh38
rs367679417 Chr.5: 149858269-149858269 on Build GRCh38
rs540946744 Chr.5: 149858300-149858300 on Build GRCh38
rs549601359 Chr.5: 149857996-149857996 on Build GRCh38
rs965453260 Chr.5: 149858280-149858280 on Build GRCh38
rs978612782 Chr.5: 149858320-149858320 on Build GRCh38
rs919607113 Chr.5: 149858129-149858129 on Build GRCh38
rs965518387 Chr.5: 149858343-149858343 on Build GRCh38
rs557379826 Chr.5: 149858243-149858243 on Build GRCh38
rs765952634 Chr.5: 149858092-149858092 on Build GRCh38
rs949847155 Chr.5: 149858193-149858193 on Build GRCh38
rs111440604 Chr.5: 149858370-149858370 on Build GRCh38
rs925834721 Chr.5: 149858321-149858321 on Build GRCh38
rs934294409 Chr.5: 149858154-149858154 on Build GRCh38

View Full Product Details