Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 152404302-152404807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NMUR2
Entrez Gene ID
56923
Gene Name
neuromedin U receptor 2
Gene Aliases
FM-4, FM4, NMU-R2, NMU2R, TGR-1, TGR1
UniGene
Hs.731911
Gene Chromosome Location
Chr.5: 152391541-152405279 on Build GRCh38
Transcripts
NM_020167.4 XM_011537670.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
362412 c.439G>C p.A147P Chr.5: 152404675-152404675 on Build GRCh38
94840 c.356G>T p.C119F Chr.5: 152404758-152404758 on Build GRCh38
6102746 c.369G>C p.T123T Chr.5: 152404745-152404745 on Build GRCh38
3715026 c.502G>T p.G168C Chr.5: 152404612-152404612 on Build GRCh38
449150 c.624G>T p.K208N Chr.5: 152404490-152404490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs921481773 Chr.5: 152404618-152404618 on Build GRCh38
rs142503678 Chr.5: 152404651-152404651 on Build GRCh38
rs371163777 Chr.5: 152404396-152404396 on Build GRCh38
rs973341206 Chr.5: 152404330-152404330 on Build GRCh38
rs143837854 Chr.5: 152404496-152404496 on Build GRCh38
rs777856453 Chr.5: 152404376-152404377 on Build GRCh38
rs867147425 Chr.5: 152404466-152404466 on Build GRCh38
rs768746134 Chr.5: 152404673-152404673 on Build GRCh38
rs755475199 Chr.5: 152404560-152404560 on Build GRCh38
rs201308697 Chr.5: 152404728-152404728 on Build GRCh38
rs774557750 Chr.5: 152404684-152404684 on Build GRCh38
rs765944267 Chr.5: 152404638-152404638 on Build GRCh38
rs770598805 Chr.5: 152404772-152404772 on Build GRCh38
rs780153464 Chr.5: 152404737-152404737 on Build GRCh38
rs973246796 Chr.5: 152404542-152404542 on Build GRCh38
rs756827531 Chr.5: 152404432-152404432 on Build GRCh38
rs776110616 Chr.5: 152404746-152404746 on Build GRCh38
rs370216531 Chr.5: 152404575-152404575 on Build GRCh38
rs767520067 Chr.5: 152404501-152404501 on Build GRCh38
rs773129806 Chr.5: 152404536-152404536 on Build GRCh38
rs753594169 Chr.5: 152404430-152404430 on Build GRCh38
rs368042517 Chr.5: 152404641-152404641 on Build GRCh38
rs1004624182 Chr.5: 152404655-152404655 on Build GRCh38
rs151084376 Chr.5: 152404497-152404497 on Build GRCh38
rs963142407 Chr.5: 152404657-152404657 on Build GRCh38
rs530506474 Chr.5: 152404678-152404678 on Build GRCh38
rs748103637 Chr.5: 152404745-152404745 on Build GRCh38
rs200288600 Chr.5: 152404417-152404417 on Build GRCh38
rs138969419 Chr.5: 152404419-152404419 on Build GRCh38
rs760667012 Chr.5: 152404511-152404511 on Build GRCh38
rs748466234 Chr.5: 152404489-152404489 on Build GRCh38
rs773059218 Chr.5: 152404683-152404683 on Build GRCh38
rs755485294 Chr.5: 152404643-152404643 on Build GRCh38
rs773156966 Chr.5: 152404383-152404383 on Build GRCh38
rs764966543 Chr.5: 152404632-152404632 on Build GRCh38
rs762004799 Chr.5: 152404355-152404355 on Build GRCh38
rs985446202 Chr.5: 152404591-152404591 on Build GRCh38
rs762638405 Chr.5: 152404636-152404636 on Build GRCh38
rs745877198 Chr.5: 152404664-152404664 on Build GRCh38
rs768901564 Chr.5: 152404350-152404350 on Build GRCh38
rs771274623 Chr.5: 152404662-152404662 on Build GRCh38
rs760760955 Chr.5: 152404615-152404615 on Build GRCh38
rs749981074 Chr.5: 152404545-152404545 on Build GRCh38
rs772204712 Chr.5: 152404499-152404499 on Build GRCh38
rs891172802 Chr.5: 152404429-152404429 on Build GRCh38
rs141825293 Chr.5: 152404697-152404697 on Build GRCh38
rs778545109 Chr.5: 152404440-152404440 on Build GRCh38
rs773941371 Chr.5: 152404783-152404783 on Build GRCh38
rs745311917 Chr.5: 152404451-152404451 on Build GRCh38
rs148445666 Chr.5: 152404730-152404730 on Build GRCh38
rs753190788 Chr.5: 152404713-152404713 on Build GRCh38
rs759004038 Chr.5: 152404701-152404701 on Build GRCh38
rs760241640 Chr.5: 152404656-152404660 on Build GRCh38
rs367573145 Chr.5: 152404524-152404524 on Build GRCh38
rs3749787 Chr.5: 152404622-152404622 on Build GRCh38
rs144599013 Chr.5: 152404663-152404663 on Build GRCh38
rs912589908 Chr.5: 152404599-152404599 on Build GRCh38
rs747309360 Chr.5: 152404345-152404345 on Build GRCh38
rs777815977 Chr.5: 152404606-152404606 on Build GRCh38
rs1050210237 Chr.5: 152404378-152404378 on Build GRCh38
rs757799993 Chr.5: 152404548-152404548 on Build GRCh38
rs765402294 Chr.5: 152404380-152404380 on Build GRCh38
rs754217110 Chr.5: 152404721-152404721 on Build GRCh38
rs781590627 Chr.5: 152404569-152404569 on Build GRCh38
rs756384094 Chr.5: 152404656-152404656 on Build GRCh38
rs145273801 Chr.5: 152404645-152404645 on Build GRCh38
rs776787609 Chr.5: 152404354-152404354 on Build GRCh38
rs891893148 Chr.5: 152404619-152404619 on Build GRCh38
rs770057944 Chr.5: 152404600-152404600 on Build GRCh38
rs886131591 Chr.5: 152404729-152404729 on Build GRCh38
rs200145877 Chr.5: 152404696-152404696 on Build GRCh38
rs747399497 Chr.5: 152404482-152404482 on Build GRCh38
rs750095849 Chr.5: 152404633-152404633 on Build GRCh38
rs1043208342 Chr.5: 152404628-152404628 on Build GRCh38
rs17851452 Chr.5: 152404504-152404504 on Build GRCh38
rs766174756 Chr.5: 152404730-152404730 on Build GRCh38
rs764432864 Chr.5: 152404720-152404720 on Build GRCh38
rs1053157647 Chr.5: 152404612-152404612 on Build GRCh38
rs763859624 Chr.5: 152404428-152404428 on Build GRCh38
rs1011768346 Chr.5: 152404478-152404478 on Build GRCh38
rs373673961 Chr.5: 152404459-152404459 on Build GRCh38
rs141055680 Chr.5: 152404607-152404607 on Build GRCh38
rs912549370 Chr.5: 152404732-152404732 on Build GRCh38
rs769060997 Chr.5: 152404486-152404486 on Build GRCh38
rs567937141 Chr.5: 152404487-152404487 on Build GRCh38
rs137937656 Chr.5: 152404629-152404629 on Build GRCh38
rs377153615 Chr.5: 152404654-152404654 on Build GRCh38
rs547914572 Chr.5: 152404611-152404611 on Build GRCh38
rs200602427 Chr.5: 152404460-152404460 on Build GRCh38
rs141741005 Chr.5: 152404761-152404761 on Build GRCh38
rs757558033 Chr.5: 152404727-152404727 on Build GRCh38
rs752104368 Chr.5: 152404558-152404558 on Build GRCh38
rs374625926 Chr.5: 152404630-152404630 on Build GRCh38
rs780499629 Chr.5: 152404481-152404481 on Build GRCh38
rs201003238 Chr.5: 152404533-152404533 on Build GRCh38
rs749512728 Chr.5: 152404661-152404661 on Build GRCh38
rs200730009 Chr.5: 152404512-152404512 on Build GRCh38
rs755930766 Chr.5: 152404420-152404420 on Build GRCh38
rs200336223 Chr.5: 152404464-152404464 on Build GRCh38
rs753673023 Chr.5: 152404515-152404515 on Build GRCh38
rs756612508 Chr.5: 152404718-152404718 on Build GRCh38
rs138543039 Chr.5: 152404675-152404675 on Build GRCh38
rs758601991 Chr.5: 152404733-152404733 on Build GRCh38
rs919913085 Chr.5: 152404338-152404338 on Build GRCh38
rs555065239 Chr.5: 152404771-152404771 on Build GRCh38
rs565548181 Chr.5: 152404506-152404506 on Build GRCh38
rs201041305 Chr.5: 152404557-152404557 on Build GRCh38
rs370782921 Chr.5: 152404659-152404663 on Build GRCh38
rs4623133 Chr.5: 152404778-152404778 on Build GRCh38
rs144629491 Chr.5: 152404705-152404705 on Build GRCh38

View Full Product Details