Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157095171-157095705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAVCR2
Entrez Gene ID
84868
Gene Name
hepatitis A virus cellular receptor 2
Gene Aliases
CD366, HAVcr-2, KIM-3, TIM3, TIMD-3, TIMD3, Tim-3
UniGene
Hs.710500
Gene Chromosome Location
Chr.5: 157085832-157109237 on Build GRCh38
Transcripts
NM_032782.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4851015 c.616G>A p.G206R Chr.5: 157095366-157095366 on Build GRCh38
1209329 c.576A>T p.L192F Chr.5: 157095406-157095406 on Build GRCh38
4585656 c.594C>T p.T198T Chr.5: 157095388-157095388 on Build GRCh38
462024 c.584C>G p.S195C Chr.5: 157095398-157095398 on Build GRCh38
6677782 c.636G>T p.G212G Chr.5: 157095346-157095346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777485384 Chr.5: 157095384-157095384 on Build GRCh38
rs759985431 Chr.5: 157095323-157095323 on Build GRCh38
rs768596805 Chr.5: 157095292-157095292 on Build GRCh38
rs1008866350 Chr.5: 157095651-157095651 on Build GRCh38
rs7727003 Chr.5: 157095367-157095367 on Build GRCh38
rs929135592 Chr.5: 157095325-157095325 on Build GRCh38
rs753941473 Chr.5: 157095281-157095281 on Build GRCh38
rs963056500 Chr.5: 157095611-157095611 on Build GRCh38
rs559449914 Chr.5: 157095388-157095388 on Build GRCh38
rs747959481 Chr.5: 157095380-157095380 on Build GRCh38
rs569173134 Chr.5: 157095206-157095206 on Build GRCh38
rs1002347419 Chr.5: 157095226-157095226 on Build GRCh38
rs149351945 Chr.5: 157095345-157095345 on Build GRCh38
rs769397225 Chr.5: 157095382-157095382 on Build GRCh38
rs780148401 Chr.5: 157095287-157095287 on Build GRCh38
rs140201248 Chr.5: 157095359-157095359 on Build GRCh38
rs776936723 Chr.5: 157095431-157095431 on Build GRCh38
rs956153892 Chr.5: 157095486-157095486 on Build GRCh38
rs1056960316 Chr.5: 157095645-157095645 on Build GRCh38
rs760581555 Chr.5: 157095396-157095397 on Build GRCh38
rs765451458 Chr.5: 157095436-157095436 on Build GRCh38
rs904525884 Chr.5: 157095443-157095443 on Build GRCh38
rs369081388 Chr.5: 157095263-157095263 on Build GRCh38
rs374018243 Chr.5: 157095404-157095404 on Build GRCh38
rs1022933376 Chr.5: 157095419-157095419 on Build GRCh38
rs764310466 Chr.5: 157095420-157095420 on Build GRCh38
rs143098742 Chr.5: 157095369-157095369 on Build GRCh38
rs895733255 Chr.5: 157095361-157095361 on Build GRCh38
rs932919248 Chr.5: 157095199-157095199 on Build GRCh38
rs769464107 Chr.5: 157095311-157095311 on Build GRCh38
rs781096999 Chr.5: 157095293-157095293 on Build GRCh38
rs1030282335 Chr.5: 157095570-157095570 on Build GRCh38
rs548394486 Chr.5: 157095609-157095609 on Build GRCh38
rs973118781 Chr.5: 157095613-157095613 on Build GRCh38
rs761934322 Chr.5: 157095227-157095227 on Build GRCh38
rs772652368 Chr.5: 157095315-157095315 on Build GRCh38
rs762519713 Chr.5: 157095321-157095321 on Build GRCh38
rs757374569 Chr.5: 157095360-157095360 on Build GRCh38
rs770287679 Chr.5: 157095544-157095544 on Build GRCh38
rs998290330 Chr.5: 157095540-157095540 on Build GRCh38
rs764373350 Chr.5: 157095351-157095351 on Build GRCh38
rs745379542 Chr.5: 157095506-157095506 on Build GRCh38
rs369829515 Chr.5: 157095505-157095505 on Build GRCh38
rs755018618 Chr.5: 157095453-157095453 on Build GRCh38
rs929081529 Chr.5: 157095640-157095640 on Build GRCh38
rs541982134 Chr.5: 157095324-157095324 on Build GRCh38
rs753084941 Chr.5: 157095327-157095327 on Build GRCh38
rs773810276 Chr.5: 157095394-157095394 on Build GRCh38
rs537669831 Chr.5: 157095212-157095212 on Build GRCh38
rs377660703 Chr.5: 157095405-157095405 on Build GRCh38
rs749703633 Chr.5: 157095574-157095574 on Build GRCh38
rs13170556 Chr.5: 157095577-157095577 on Build GRCh38
rs756330537 Chr.5: 157095262-157095262 on Build GRCh38
rs758335369 Chr.5: 157095286-157095286 on Build GRCh38
rs767283571 Chr.5: 157095307-157095308 on Build GRCh38
rs747792545 Chr.5: 157095295-157095295 on Build GRCh38
rs186992535 Chr.5: 157095633-157095633 on Build GRCh38
rs777383129 Chr.5: 157095504-157095504 on Build GRCh38
rs752509667 Chr.5: 157095461-157095461 on Build GRCh38
rs770241205 Chr.5: 157095678-157095678 on Build GRCh38
rs573207701 Chr.5: 157095447-157095447 on Build GRCh38
rs145201178 Chr.5: 157095458-157095458 on Build GRCh38
rs765341404 Chr.5: 157095326-157095330 on Build GRCh38
rs918994608 Chr.5: 157095625-157095625 on Build GRCh38
rs941723566 Chr.5: 157095290-157095290 on Build GRCh38
rs551267016 Chr.5: 157095201-157095201 on Build GRCh38
rs762090318 Chr.5: 157095432-157095432 on Build GRCh38
rs781177209 Chr.5: 157095375-157095375 on Build GRCh38
rs183238604 Chr.5: 157095322-157095322 on Build GRCh38
rs767714424 Chr.5: 157095365-157095365 on Build GRCh38
rs910177390 Chr.5: 157095585-157095585 on Build GRCh38
rs982688767 Chr.5: 157095451-157095451 on Build GRCh38
rs746889903 Chr.5: 157095289-157095289 on Build GRCh38
rs755949696 Chr.5: 157095466-157095466 on Build GRCh38
rs201195681 Chr.5: 157095402-157095402 on Build GRCh38
rs890451858 Chr.5: 157095632-157095632 on Build GRCh38
rs1009831782 Chr.5: 157095269-157095270 on Build GRCh38
rs748833026 Chr.5: 157095387-157095387 on Build GRCh38

View Full Product Details